Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_031283.2 | NP_112573.1 | 4, 5 | 743 | |
XM_006712109.2 | XP_006712172.1 | 4, 5 | 533 | ||
GenBank mRNA | AB031046.1 | - | 4, 5 | 592 | |
BC058894.1 | - | 4, 5 | 752 | 6141641 |