Gene Transcripts
- Gene Symbol
- FAM188B
- Entrez Gene ID
- 84182
- Gene Name
- family with sequence similarity 188 member B
- Gene Aliases
- C7orf67
- Chromosome Location
- Chr. 7: 30771417 - 30892387 on Build GRCh38
- UniGene ID
- Hs.660192
- Species
- Homo sapiens
- Gene Symbol
- INMT-FAM188B
- Entrez Gene ID
- 100526825
- Gene Name
- INMT-FAM188B readthrough (NMD candidate)
- Gene Aliases
- Chromosome Location
- Chr. 7: 30752135 - 30892387 on Build GRCh38
- UniGene ID
- Hs.660192
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037598.1 |
|
7 |
1345 |
|
GenBank mRNA |
AK308762.1 |
- |
7 |
1343 |
|
View Full Product Details