Gene Transcripts
- Gene Symbol
- THOC3
- Entrez Gene ID
- 84321
- Gene Name
- THO complex 3
- Gene Aliases
- THO3, hTREX45
- Chromosome Location
- Chr. 5: 175959531 - 176034905 on Build GRCh38
- UniGene ID
- Hs.484227
- Species
- Homo sapiens
- Gene Symbol
- LOC728554
- Entrez Gene ID
- 728554
- Gene Name
- THO complex 3 pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 5: 177875261 - 177884268 on Build GRCh38
- UniGene ID
- Hs.535769
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003615.2 |
|
6 |
1238 |
|
View Full Product Details