Gene Transcripts

Gene Symbol
THOC3
Entrez Gene ID
84321
Gene Name
THO complex 3
Gene Aliases
THO3, hTREX45
Chromosome Location
Chr. 5: 175959531 - 176034905 on Build GRCh38
UniGene ID
Hs.484227
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032361.3 NP_115737.1 6 1093
XM_017009985.1 XP_016865474.1 7 960
GenBank mRNA BC006849.1 - 6 1055
BC068499.1 - 6 1012 5262880
BU634063.1 - 1 521

Gene Symbol
LOC728554
Entrez Gene ID
728554
Gene Name
THO complex 3 pseudogene
Gene Aliases
Chromosome Location
Chr. 5: 177875261 - 177884268 on Build GRCh38
UniGene ID
Hs.535769
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003615.2 6 1319

View Full Product Details