Gene Transcripts

Gene Symbol
LZTS2
Entrez Gene ID
84445
Gene Name
leucine zipper tumor suppressor 2
Gene Aliases
LAPSER1
Chromosome Location
Chr. 10: 100997065 - 101007836 on Build GRCh38
UniGene ID
Hs.523221
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318099.1 NP_001305028.1 2 416
NM_001318100.1 NP_001305029.1 2 493
NM_001318101.1 NP_001305030.1 1 354
NM_032429.3 NP_115805.1 2 523
XM_005270222.4 XP_005270279.1 2 755
XM_017016780.1 XP_016872269.1 1 2884
XM_017016781.1 XP_016872270.1 2 524
GenBank mRNA AB058716.1 - 1 3375
AK097997.1 - 1 146
AK298208.1 - 2 381
AY029201.1 - 1 237
BC006212.2 - 2 381 3161855
BC058938.1 - 2 456 3913470
BM544815.1 - 482
CN365632.1 - 2 493
CV803721.1 - 522

View Full Product Details