Gene Transcripts

Gene Symbol
MUM1
Entrez Gene ID
84939
Gene Name
melanoma associated antigen (mutated) 1
Gene Aliases
EXPAND1, HSPC211, MUM-1
Chromosome Location
Chr. 19: 1354977 - 1378431 on Build GRCh38
UniGene ID
Hs.515016
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032853.3 NP_116242.2 5, 6 1222
NR_024247.1 5, 6 1291
XM_011528380.2 XP_011526682.1 5, 6 1212
XM_011528382.2 XP_011526684.1 5, 6 1212
XM_011528383.1 XP_011526685.1 5, 6 1212
XM_011528384.2 XP_011526686.1 5, 6 1212
XM_011528385.2 XP_011526687.1 6, 7 1413
XM_017027401.1 XP_016882890.1 5, 6 1212
GenBank mRNA AK309092.1 - 4, 5 1110
AK310721.1 - 6, 7 1325
BC110874.1 - 4, 5 1188 4933735
BC130443.1 - 5, 6 1251 40146474
BC144138.1 - 4, 5 1138 9052658
BC144147.1 - 5, 6 1251
BQ071619.1 - 1, 2 253
U20897.1 - 4, 5 1192

View Full Product Details