Gene Transcripts
- Gene Symbol
- KRT37
- Entrez Gene ID
- 8688
- Gene Name
- keratin 37
- Gene Aliases
- HA7, K37, KRTHA7
- Chromosome Location
- Chr. 17: 41420557 - 41424570 on Build GRCh38
- UniGene ID
- Hs.673852
- Species
- Homo sapiens
- Gene Symbol
- LOC100505782
- Entrez Gene ID
- 100505782
- Gene Name
- uncharacterized LOC100505782
- Gene Aliases
- Chromosome Location
- Chr. 17: 41402416 - 41412588 on Build GRCh38
- UniGene ID
- Hs.463024
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK123843.1 |
- |
4 |
3112 |
|
View Full Product Details