Gene Transcripts

Gene Symbol
RABEP1
Entrez Gene ID
9135
Gene Name
rabaptin, RAB GTPase binding effector protein 1
Gene Aliases
RAB5EP, RABPT5
Chromosome Location
Chr. 17: 5282263 - 5386340 on Build GRCh38
UniGene ID
Hs.584784
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083585.2 NP_001077054.1 9 1615
NM_001291581.1 NP_001278510.1 8 1486
NM_004703.5 NP_004694.2 9 1615
XM_011524060.1 XP_011522362.1 10 1681
XM_017025294.1 XP_016880783.1 8 1208
XM_017025295.1 XP_016880784.1 7 1142
GenBank mRNA AF098638.1 - 9 1391
AK297535.1 - 8 1475
AK310932.1 - 8 1427
AK314183.1 - 9 1606
BC032435.1 - 9 1569
BC041700.1 - 9 1581 6046320
X77723.1 - 9 1464
X91141.1 - 9 1579

View Full Product Details