Gene Transcripts

Gene Symbol
SLK
Entrez Gene ID
9748
Gene Name
STE20 like kinase
Gene Aliases
LOSK, STK2, bA16H23.1, se20-9
Chromosome Location
Chr. 10: 103967173 - 104029233 on Build GRCh38
UniGene ID
Hs.591922
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304743.1 NP_001291672.1 9 2569
NM_014720.3 NP_055535.2 9 2569
GenBank mRNA AB002804.1 - 9 2240
AF273048.1 - 9 2042
AK302855.1 - 9 2002
BC111565.1 - 9 2039 40009070
D86959.1 - 9 2519

View Full Product Details