Gene Transcripts

Gene Symbol
AP5Z1
Entrez Gene ID
9907
Gene Name
adaptor related protein complex 5 zeta 1 subunit
Gene Aliases
KIAA0415, SPG48, zeta
Chromosome Location
Chr. 7: 4775617 - 4794395 on Build GRCh38
UniGene ID
Hs.558440
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014855.2 NP_055670.1 2, 3 269
GenBank mRNA AB007875.1 - 2, 3 248
BI766915.1 - 2, 3 269

View Full Product Details