Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 69703843-69704350 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF19
Entrez Gene ID
9965
Gene Name
fibroblast growth factor 19
Gene Aliases
-
UniGene
Hs.249200
Gene Chromosome Location
Chr.11: 69698238-69704338 on Build GRCh38
Transcripts
NM_005117.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3935612 c.33G>T p.W11C Chr.11: 69703844-69703844 on Build GRCh38
4407689 c.28G>A p.V10I Chr.11: 69703849-69703849 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs982366457 Chr.11: 69704214-69704214 on Build GRCh38
rs987180972 Chr.11: 69703938-69703938 on Build GRCh38
rs1039699217 Chr.11: 69704048-69704048 on Build GRCh38
rs988405881 Chr.11: 69704173-69704173 on Build GRCh38
rs528044141 Chr.11: 69703918-69703918 on Build GRCh38
rs890345163 Chr.11: 69704265-69704266 on Build GRCh38
rs988825791 Chr.11: 69703955-69703955 on Build GRCh38
rs999746180 Chr.11: 69704144-69704144 on Build GRCh38
rs914116927 Chr.11: 69704175-69704176 on Build GRCh38
rs951602386 Chr.11: 69704254-69704254 on Build GRCh38
rs186277229 Chr.11: 69704045-69704045 on Build GRCh38
rs569437357 Chr.11: 69703959-69703959 on Build GRCh38
rs1010548268 Chr.11: 69704182-69704182 on Build GRCh38
rs557621967 Chr.11: 69703954-69703954 on Build GRCh38
rs928104252 Chr.11: 69703967-69703967 on Build GRCh38
rs969622553 Chr.11: 69704204-69704204 on Build GRCh38
rs926634610 Chr.11: 69704183-69704183 on Build GRCh38
rs900421094 Chr.11: 69704050-69704050 on Build GRCh38
rs571953909 Chr.11: 69703952-69703952 on Build GRCh38
rs938096940 Chr.11: 69704202-69704202 on Build GRCh38
rs1023219863 Chr.11: 69704191-69704191 on Build GRCh38
rs573163952 Chr.11: 69704094-69704094 on Build GRCh38
rs1047564836 Chr.11: 69704241-69704241 on Build GRCh38
rs753330387 Chr.11: 69703907-69703907 on Build GRCh38
rs978940656 Chr.11: 69704124-69704124 on Build GRCh38
rs993421618 Chr.11: 69704044-69704044 on Build GRCh38
rs890333879 Chr.11: 69704080-69704080 on Build GRCh38
rs188042408 Chr.11: 69704308-69704308 on Build GRCh38
rs1046435798 Chr.11: 69703966-69703966 on Build GRCh38
rs958479648 Chr.11: 69704162-69704163 on Build GRCh38
rs1017238401 Chr.11: 69704101-69704101 on Build GRCh38
rs369012502 Chr.11: 69704001-69704001 on Build GRCh38
rs983342517 Chr.11: 69704256-69704256 on Build GRCh38
rs1030777052 Chr.11: 69704176-69704176 on Build GRCh38
rs1008486071 Chr.11: 69704092-69704092 on Build GRCh38
rs899059742 Chr.11: 69703908-69703908 on Build GRCh38
rs1008790224 Chr.11: 69704271-69704271 on Build GRCh38
rs867572698 Chr.11: 69703986-69703986 on Build GRCh38
rs1001998520 Chr.11: 69703873-69703873 on Build GRCh38
rs1035232907 Chr.11: 69704234-69704234 on Build GRCh38
rs566945997 Chr.11: 69703896-69703896 on Build GRCh38
rs755021668 Chr.11: 69704034-69704034 on Build GRCh38
rs1017270999 Chr.11: 69703941-69703941 on Build GRCh38
rs538941127 Chr.11: 69703953-69703953 on Build GRCh38
rs964407806 Chr.11: 69704115-69704115 on Build GRCh38
rs546268599 Chr.11: 69703949-69703949 on Build GRCh38
rs975418348 Chr.11: 69704323-69704323 on Build GRCh38
rs909982187 Chr.11: 69704291-69704291 on Build GRCh38
rs931851735 Chr.11: 69704064-69704064 on Build GRCh38
rs768424122 Chr.11: 69703965-69703965 on Build GRCh38
rs893516556 Chr.11: 69704179-69704179 on Build GRCh38
rs993846387 Chr.11: 69703912-69703912 on Build GRCh38
rs540664671 Chr.11: 69704162-69704162 on Build GRCh38
rs954412146 Chr.11: 69703927-69703927 on Build GRCh38
rs1032497136 Chr.11: 69704127-69704127 on Build GRCh38
rs558678896 Chr.11: 69704232-69704232 on Build GRCh38
rs1051674274 Chr.11: 69704067-69704067 on Build GRCh38

View Full Product Details