Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 111299504-111300044 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COLCA1
Entrez Gene ID
399948
Gene Name
colorectal cancer associated 1
Gene Aliases
C11orf92, CASC12, LOH11CR1F
UniGene
Hs.729225
Gene Chromosome Location
Chr.11: 111293389-111305048 on Build GRCh38
Transcripts
NM_001302644.1 NM_001302645.1 NM_001302647.1 NM_001302648.1 NM_207429.3 NM_001136105.2 NM_001271457.1 NM_001271458.1 XM_017017196.1 XM_017017198.1 XM_017017199.1

Gene Symbol
COLCA2
Entrez Gene ID
120376
Gene Name
colorectal cancer associated 2
Gene Aliases
C11orf93, CASC13, LOH11CR1G
UniGene
Hs.31409
Gene Chromosome Location
Chr.11: 111298517-111308735 on Build GRCh38
Transcripts
NM_001302644.1 NM_001302645.1 NM_001302647.1 NM_001302648.1 NM_207429.3 NM_001136105.2 NM_001271457.1 NM_001271458.1 XM_017017196.1 XM_017017198.1 XM_017017199.1

SNP Detail

SNP ID Chromosome Location
rs918047867 Chr.11: 111299841-111299841 on Build GRCh38
rs903587061 Chr.11: 111299763-111299763 on Build GRCh38
rs563763640 Chr.11: 111299949-111299949 on Build GRCh38
rs1039212406 Chr.11: 111299805-111299805 on Build GRCh38
rs888800785 Chr.11: 111299656-111299656 on Build GRCh38
rs538172136 Chr.11: 111299803-111299803 on Build GRCh38
rs908402822 Chr.11: 111299563-111299563 on Build GRCh38
rs757601705 Chr.11: 111299769-111299769 on Build GRCh38
rs1019109786 Chr.11: 111299944-111299944 on Build GRCh38
rs188147167 Chr.11: 111299870-111299870 on Build GRCh38
rs7105857 Chr.11: 111300019-111300019 on Build GRCh38
rs949915760 Chr.11: 111299554-111299554 on Build GRCh38
rs553211030 Chr.11: 111299878-111299878 on Build GRCh38
rs888750113 Chr.11: 111299892-111299892 on Build GRCh38
rs900793712 Chr.11: 111299810-111299810 on Build GRCh38
rs192433626 Chr.11: 111299755-111299755 on Build GRCh38
rs926580845 Chr.11: 111299779-111299779 on Build GRCh38
rs926578426 Chr.11: 111299674-111299674 on Build GRCh38
rs139339657 Chr.11: 111299891-111299891 on Build GRCh38
rs994052273 Chr.11: 111300013-111300013 on Build GRCh38
rs1033435866 Chr.11: 111299886-111299886 on Build GRCh38
rs997746056 Chr.11: 111299812-111299812 on Build GRCh38
rs868229775 Chr.11: 111299691-111299691 on Build GRCh38
rs936402352 Chr.11: 111299951-111299951 on Build GRCh38
rs999805935 Chr.11: 111299627-111299627 on Build GRCh38
rs1005274597 Chr.11: 111299867-111299867 on Build GRCh38
rs886418123 Chr.11: 111299818-111299818 on Build GRCh38
rs769430763 Chr.11: 111299927-111299927 on Build GRCh38
rs1032662585 Chr.11: 111299649-111299649 on Build GRCh38
rs1042023054 Chr.11: 111299580-111299580 on Build GRCh38
rs903143448 Chr.11: 111299612-111299612 on Build GRCh38
rs79414772 Chr.11: 111299869-111299869 on Build GRCh38
rs556921080 Chr.11: 111299724-111299724 on Build GRCh38
rs555495534 Chr.11: 111299711-111299711 on Build GRCh38
rs917590248 Chr.11: 111299908-111299908 on Build GRCh38
rs150883566 Chr.11: 111299880-111299880 on Build GRCh38
rs1007231745 Chr.11: 111299924-111299924 on Build GRCh38
rs1026552755 Chr.11: 111299745-111299745 on Build GRCh38
rs546878910 Chr.11: 111299694-111299694 on Build GRCh38
rs983527412 Chr.11: 111299545-111299545 on Build GRCh38
rs973866707 Chr.11: 111299642-111299642 on Build GRCh38
rs962542702 Chr.11: 111299564-111299564 on Build GRCh38
rs10891246 Chr.11: 111299815-111299815 on Build GRCh38
rs1018707274 Chr.11: 111299698-111299698 on Build GRCh38
rs945083932 Chr.11: 111299926-111299926 on Build GRCh38
rs535588239 Chr.11: 111299722-111299722 on Build GRCh38
rs1042113453 Chr.11: 111299759-111299760 on Build GRCh38
rs540233579 Chr.11: 111299871-111299871 on Build GRCh38
rs952295114 Chr.11: 111299748-111299748 on Build GRCh38
rs117460636 Chr.11: 111299719-111299719 on Build GRCh38
rs374806919 Chr.11: 111299970-111299970 on Build GRCh38

View Full Product Details