Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1392588-1393085 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNL2
Entrez Gene ID
81669
Gene Name
cyclin L2
Gene Aliases
ANIA-6B, CCNM, CCNS, HCLA-ISO, HLA-ISO, PCEE, SB138
UniGene
Hs.515704
Gene Chromosome Location
Chr.1: 1385711-1399338 on Build GRCh38
Transcripts
NM_001039577.3 NM_001320153.1 NM_001320155.1 NM_030937.4 NR_135154.1 XM_011542216.2 XM_011542219.2 XM_011542221.2 XM_017002420.1 XM_017002421.1 XM_017002422.1 XM_017002423.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3687044 c.675T>C p.G225G Chr.1: 1392788-1392788 on Build GRCh38
6327376 c.661G>A p.V221I Chr.1: 1392802-1392802 on Build GRCh38
1472506 c.670G>A p.E224K Chr.1: 1392793-1392793 on Build GRCh38
7133932 c.670G>C p.E224Q Chr.1: 1392793-1392793 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs894565099 Chr.1: 1393008-1393008 on Build GRCh38
rs903076820 Chr.1: 1392951-1392951 on Build GRCh38
rs947822827 Chr.1: 1392618-1392620 on Build GRCh38
rs941498751 Chr.1: 1392676-1392676 on Build GRCh38
rs762667799 Chr.1: 1392930-1392938 on Build GRCh38
rs567015990 Chr.1: 1393043-1393044 on Build GRCh38
rs769240589 Chr.1: 1392969-1392969 on Build GRCh38
rs754137598 Chr.1: 1392675-1392675 on Build GRCh38
rs942420287 Chr.1: 1392726-1392726 on Build GRCh38
rs1024513698 Chr.1: 1393060-1393060 on Build GRCh38
rs773132694 Chr.1: 1392911-1392911 on Build GRCh38
rs751183142 Chr.1: 1392781-1392781 on Build GRCh38
rs927720094 Chr.1: 1393048-1393048 on Build GRCh38
rs769130486 Chr.1: 1392795-1392796 on Build GRCh38
rs762691112 Chr.1: 1392756-1392756 on Build GRCh38
rs369862332 Chr.1: 1392641-1392641 on Build GRCh38
rs1038505662 Chr.1: 1392902-1392902 on Build GRCh38
rs749748831 Chr.1: 1392767-1392769 on Build GRCh38
rs573995576 Chr.1: 1392755-1392755 on Build GRCh38
rs996975763 Chr.1: 1392926-1392926 on Build GRCh38
rs542807050 Chr.1: 1392891-1392891 on Build GRCh38
rs927679802 Chr.1: 1392639-1392639 on Build GRCh38
rs780501949 Chr.1: 1392840-1392840 on Build GRCh38
rs113515239 Chr.1: 1392857-1392857 on Build GRCh38
rs750017885 Chr.1: 1392849-1392849 on Build GRCh38
rs756433379 Chr.1: 1392796-1392796 on Build GRCh38
rs956619196 Chr.1: 1392990-1392990 on Build GRCh38
rs1045116117 Chr.1: 1392631-1392631 on Build GRCh38
rs933952225 Chr.1: 1392905-1392905 on Build GRCh38
rs1054421986 Chr.1: 1392973-1392973 on Build GRCh38
rs748150902 Chr.1: 1392736-1392736 on Build GRCh38
rs749688174 Chr.1: 1392915-1392915 on Build GRCh38
rs969529504 Chr.1: 1393040-1393040 on Build GRCh38
rs1013002122 Chr.1: 1393034-1393034 on Build GRCh38
rs756983775 Chr.1: 1392816-1392816 on Build GRCh38
rs989308333 Chr.1: 1393030-1393030 on Build GRCh38
rs763911917 Chr.1: 1392762-1392762 on Build GRCh38
rs755590709 Chr.1: 1392852-1392852 on Build GRCh38
rs1047286302 Chr.1: 1392659-1392659 on Build GRCh38
rs557189440 Chr.1: 1392712-1392712 on Build GRCh38
rs939579103 Chr.1: 1392656-1392656 on Build GRCh38
rs1030064924 Chr.1: 1392927-1392927 on Build GRCh38
rs537476101 Chr.1: 1392610-1392610 on Build GRCh38

View Full Product Details