Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 8136669-8137205 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC4A
Entrez Gene ID
50856
Gene Name
C-type lectin domain family 4 member A
Gene Aliases
CD367, CLECSF6, DCIR, DDB27, HDCGC13P, LLIR
UniGene
Hs.504657
Gene Chromosome Location
Chr.12: 8102932-8138607 on Build GRCh38
Transcripts
NM_016184.3 NM_194447.2 NM_194448.2 NM_194450.2 XM_011520684.2 XM_017019382.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1993827 c.520C>T p.R174* Chr.12: 8136857-8136857 on Build GRCh38
5923959 c.508C>T p.P170S Chr.12: 8136845-8136845 on Build GRCh38
1993828 c.527G>T p.W176L Chr.12: 8136864-8136864 on Build GRCh38
6073910 c.533G>T p.W178L Chr.12: 8136870-8136870 on Build GRCh38
943962 c.463C>A p.Q155K Chr.12: 8136800-8136800 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755332351 Chr.12: 8136937-8136937 on Build GRCh38
rs769747860 Chr.12: 8136867-8136867 on Build GRCh38
rs755010184 Chr.12: 8137000-8137000 on Build GRCh38
rs761777038 Chr.12: 8136754-8136754 on Build GRCh38
rs1028492263 Chr.12: 8136824-8136824 on Build GRCh38
rs369161603 Chr.12: 8137164-8137164 on Build GRCh38
rs901128185 Chr.12: 8137088-8137088 on Build GRCh38
rs780431971 Chr.12: 8136853-8136853 on Build GRCh38
rs773020133 Chr.12: 8136773-8136773 on Build GRCh38
rs770473273 Chr.12: 8136770-8136772 on Build GRCh38
rs751960808 Chr.12: 8136926-8136926 on Build GRCh38
rs754708753 Chr.12: 8136789-8136789 on Build GRCh38
rs952138191 Chr.12: 8136889-8136889 on Build GRCh38
rs1028857983 Chr.12: 8137144-8137144 on Build GRCh38
rs115176426 Chr.12: 8136865-8136865 on Build GRCh38
rs763519457 Chr.12: 8136910-8136910 on Build GRCh38
rs776714056 Chr.12: 8136745-8136745 on Build GRCh38
rs774305756 Chr.12: 8136888-8136888 on Build GRCh38
rs4573763 Chr.12: 8136704-8136704 on Build GRCh38
rs762803801 Chr.12: 8136870-8136870 on Build GRCh38
rs1048862922 Chr.12: 8136969-8136969 on Build GRCh38
rs984854023 Chr.12: 8137020-8137020 on Build GRCh38
rs939124811 Chr.12: 8136908-8136908 on Build GRCh38
rs759380542 Chr.12: 8136891-8136891 on Build GRCh38
rs375780871 Chr.12: 8137055-8137055 on Build GRCh38
rs930506324 Chr.12: 8136955-8136955 on Build GRCh38
rs150108595 Chr.12: 8136868-8136868 on Build GRCh38
rs751331987 Chr.12: 8136782-8136782 on Build GRCh38
rs370567213 Chr.12: 8136945-8136945 on Build GRCh38
rs762861802 Chr.12: 8136774-8136774 on Build GRCh38
rs370356837 Chr.12: 8136766-8136766 on Build GRCh38
rs886376493 Chr.12: 8136970-8136970 on Build GRCh38
rs781238527 Chr.12: 8136911-8136911 on Build GRCh38
rs1050566558 Chr.12: 8136723-8136723 on Build GRCh38
rs1013931980 Chr.12: 8136850-8136850 on Build GRCh38
rs767315232 Chr.12: 8136903-8136903 on Build GRCh38
rs201586495 Chr.12: 8136858-8136858 on Build GRCh38
rs758876638 Chr.12: 8136843-8136843 on Build GRCh38
rs747325810 Chr.12: 8136856-8136856 on Build GRCh38
rs976478355 Chr.12: 8137106-8137106 on Build GRCh38
rs150982146 Chr.12: 8136842-8136842 on Build GRCh38
rs1000615281 Chr.12: 8136797-8136797 on Build GRCh38
rs145478742 Chr.12: 8136874-8136874 on Build GRCh38
rs1015524617 Chr.12: 8136799-8136799 on Build GRCh38
rs758854703 Chr.12: 8136920-8136920 on Build GRCh38
rs1037000364 Chr.12: 8137087-8137087 on Build GRCh38
rs903646008 Chr.12: 8136777-8136778 on Build GRCh38
rs775106643 Chr.12: 8136777-8136777 on Build GRCh38
rs200598348 Chr.12: 8136830-8136830 on Build GRCh38
rs755545607 Chr.12: 8136823-8136823 on Build GRCh38
rs748330306 Chr.12: 8136948-8136948 on Build GRCh38
rs767182062 Chr.12: 8136769-8136769 on Build GRCh38
rs755736857 Chr.12: 8136904-8136904 on Build GRCh38
rs189836326 Chr.12: 8137095-8137095 on Build GRCh38
rs1026766833 Chr.12: 8136885-8136885 on Build GRCh38
rs201965264 Chr.12: 8136815-8136815 on Build GRCh38
rs748313787 Chr.12: 8137161-8137161 on Build GRCh38
rs761602379 Chr.12: 8137056-8137056 on Build GRCh38
rs764432384 Chr.12: 8136857-8136857 on Build GRCh38
rs932221344 Chr.12: 8137170-8137170 on Build GRCh38
rs145194356 Chr.12: 8136808-8136808 on Build GRCh38
rs1011517646 Chr.12: 8137181-8137181 on Build GRCh38
rs923448281 Chr.12: 8136692-8136692 on Build GRCh38
rs774208929 Chr.12: 8136877-8136878 on Build GRCh38

View Full Product Details