Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 9913514-9914056 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC2A
Entrez Gene ID
387836
Gene Name
C-type lectin domain family 2 member A
Gene Aliases
INPE5792, KACL, PILAR, UNQ5792
UniGene
Hs.527665
Gene Chromosome Location
Chr.12: 9882733-9932430 on Build GRCh38
Transcripts
NM_001130711.1 NM_207375.2 XM_011520654.2 XM_011520656.2 XM_011520657.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5766499 c.429C>T p.N143N Chr.12: 9913662-9913662 on Build GRCh38
5794170 c.485A>T p.D162V Chr.12: 9913606-9913606 on Build GRCh38
5448352 c.445T>G p.L149V Chr.12: 9913646-9913646 on Build GRCh38
1476020 c.484G>C p.D162H Chr.12: 9913607-9913607 on Build GRCh38
6335356 c.443T>A p.F148Y Chr.12: 9913648-9913648 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112000446 Chr.12: 9913543-9913543 on Build GRCh38
rs937075409 Chr.12: 9913638-9913638 on Build GRCh38
rs924090120 Chr.12: 9913791-9913791 on Build GRCh38
rs759958618 Chr.12: 9913713-9913713 on Build GRCh38
rs1030226592 Chr.12: 9913737-9913737 on Build GRCh38
rs138914180 Chr.12: 9913606-9913606 on Build GRCh38
rs923104902 Chr.12: 9913540-9913540 on Build GRCh38
rs1026091708 Chr.12: 9913668-9913668 on Build GRCh38
rs978311246 Chr.12: 9913773-9913773 on Build GRCh38
rs117059636 Chr.12: 9913943-9913943 on Build GRCh38
rs945952402 Chr.12: 9913955-9913955 on Build GRCh38
rs73255422 Chr.12: 9913956-9913956 on Build GRCh38
rs556205647 Chr.12: 9913717-9913717 on Build GRCh38
rs377339235 Chr.12: 9913662-9913662 on Build GRCh38
rs373749464 Chr.12: 9913564-9913564 on Build GRCh38
rs566853865 Chr.12: 9913833-9913833 on Build GRCh38
rs527521433 Chr.12: 9913870-9913870 on Build GRCh38
rs978187216 Chr.12: 9913549-9913549 on Build GRCh38
rs112806397 Chr.12: 9913988-9913988 on Build GRCh38
rs373859719 Chr.12: 9913625-9913625 on Build GRCh38
rs994265226 Chr.12: 9913661-9913661 on Build GRCh38
rs755360875 Chr.12: 9913639-9913639 on Build GRCh38
rs913165952 Chr.12: 9913601-9913601 on Build GRCh38
rs974813122 Chr.12: 9913932-9913932 on Build GRCh38
rs541328410 Chr.12: 9913918-9913918 on Build GRCh38
rs186609507 Chr.12: 9913786-9913786 on Build GRCh38
rs935973571 Chr.12: 9913592-9913592 on Build GRCh38
rs1046591493 Chr.12: 9913629-9913629 on Build GRCh38
rs542623105 Chr.12: 9913701-9913701 on Build GRCh38
rs561168803 Chr.12: 9913935-9913935 on Build GRCh38
rs1016353539 Chr.12: 9913682-9913682 on Build GRCh38
rs964336867 Chr.12: 9913720-9913720 on Build GRCh38
rs142033208 Chr.12: 9913680-9913680 on Build GRCh38
rs766082778 Chr.12: 9913861-9913861 on Build GRCh38
rs913221876 Chr.12: 9913946-9913946 on Build GRCh38
rs924008907 Chr.12: 9913553-9913553 on Build GRCh38
rs886726738 Chr.12: 9913755-9913755 on Build GRCh38
rs117696017 Chr.12: 9913600-9913600 on Build GRCh38
rs906728088 Chr.12: 9913636-9913636 on Build GRCh38
rs767801006 Chr.12: 9913915-9913916 on Build GRCh38
rs936744511 Chr.12: 9913794-9913794 on Build GRCh38
rs867813462 Chr.12: 9913642-9913642 on Build GRCh38
rs571746731 Chr.12: 9914006-9914006 on Build GRCh38
rs115598241 Chr.12: 9913676-9913676 on Build GRCh38
rs376979852 Chr.12: 9914031-9914031 on Build GRCh38
rs776059249 Chr.12: 9913683-9913683 on Build GRCh38
rs954556623 Chr.12: 9913772-9913772 on Build GRCh38
rs545512017 Chr.12: 9913727-9913727 on Build GRCh38
rs745563664 Chr.12: 9913683-9913684 on Build GRCh38
rs1005058885 Chr.12: 9913831-9913831 on Build GRCh38
rs532096847 Chr.12: 9913981-9913981 on Build GRCh38
rs114146548 Chr.12: 9913721-9913721 on Build GRCh38
rs1019074946 Chr.12: 9913854-9913854 on Build GRCh38
rs79629724 Chr.12: 9913941-9913941 on Build GRCh38
rs916582855 Chr.12: 9913851-9913851 on Build GRCh38
rs945941055 Chr.12: 9913605-9913605 on Build GRCh38
rs966161019 Chr.12: 9913929-9913929 on Build GRCh38
rs906779258 Chr.12: 9913928-9913928 on Build GRCh38
rs866596057 Chr.12: 9913775-9913775 on Build GRCh38
rs764475870 Chr.12: 9913596-9913596 on Build GRCh38
rs921975677 Chr.12: 9913566-9913566 on Build GRCh38

View Full Product Details