Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 18738407-18738912 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPZA3
Entrez Gene ID
93661
Gene Name
capping actin protein of muscle Z-line alpha subunit 3
Gene Aliases
CAPPA3, Gsg3, HEL-S-86
UniGene
Hs.131288
Gene Chromosome Location
Chr.12: 18738111-18739188 on Build GRCh38
Transcripts
NM_033328.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
348621 c.627G>A p.L209L Chr.12: 18738895-18738895 on Build GRCh38
2003962 c.521G>A p.W174* Chr.12: 18738789-18738789 on Build GRCh38
2003958 c.245G>A p.R82Q Chr.12: 18738513-18738513 on Build GRCh38
4882509 c.414C>A p.C138* Chr.12: 18738682-18738682 on Build GRCh38
6479457 c.402A>G p.P134P Chr.12: 18738670-18738670 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755845015 Chr.12: 18738730-18738730 on Build GRCh38
rs777481523 Chr.12: 18738524-18738524 on Build GRCh38
rs764491379 Chr.12: 18738812-18738812 on Build GRCh38
rs749632201 Chr.12: 18738522-18738522 on Build GRCh38
rs769365098 Chr.12: 18738513-18738513 on Build GRCh38
rs753268001 Chr.12: 18738825-18738825 on Build GRCh38
rs748554082 Chr.12: 18738621-18738621 on Build GRCh38
rs143679033 Chr.12: 18738583-18738583 on Build GRCh38
rs922984227 Chr.12: 18738733-18738733 on Build GRCh38
rs577490193 Chr.12: 18738447-18738447 on Build GRCh38
rs199538736 Chr.12: 18738640-18738640 on Build GRCh38
rs868010711 Chr.12: 18738778-18738778 on Build GRCh38
rs201586615 Chr.12: 18738724-18738724 on Build GRCh38
rs370193589 Chr.12: 18738448-18738448 on Build GRCh38
rs751977734 Chr.12: 18738818-18738818 on Build GRCh38
rs760833481 Chr.12: 18738760-18738760 on Build GRCh38
rs1046422350 Chr.12: 18738484-18738484 on Build GRCh38
rs151069259 Chr.12: 18738506-18738506 on Build GRCh38
rs755787082 Chr.12: 18738822-18738822 on Build GRCh38
rs745337795 Chr.12: 18738636-18738636 on Build GRCh38
rs111403135 Chr.12: 18738625-18738626 on Build GRCh38
rs199684920 Chr.12: 18738726-18738726 on Build GRCh38
rs146799170 Chr.12: 18738729-18738729 on Build GRCh38
rs368077520 Chr.12: 18738823-18738823 on Build GRCh38
rs776700468 Chr.12: 18738479-18738479 on Build GRCh38
rs868744411 Chr.12: 18738675-18738675 on Build GRCh38
rs747865955 Chr.12: 18738750-18738750 on Build GRCh38
rs779638442 Chr.12: 18738731-18738731 on Build GRCh38
rs753969233 Chr.12: 18738674-18738674 on Build GRCh38
rs142906283 Chr.12: 18738463-18738463 on Build GRCh38
rs751382294 Chr.12: 18738562-18738562 on Build GRCh38
rs369388250 Chr.12: 18738505-18738505 on Build GRCh38
rs777482385 Chr.12: 18738856-18738856 on Build GRCh38
rs767705185 Chr.12: 18738660-18738660 on Build GRCh38
rs755360500 Chr.12: 18738614-18738614 on Build GRCh38
rs766521488 Chr.12: 18738732-18738732 on Build GRCh38
rs775605917 Chr.12: 18738549-18738549 on Build GRCh38
rs375648441 Chr.12: 18738746-18738746 on Build GRCh38
rs201991985 Chr.12: 18738644-18738644 on Build GRCh38
rs765807471 Chr.12: 18738688-18738688 on Build GRCh38
rs749122388 Chr.12: 18738638-18738638 on Build GRCh38
rs780507786 Chr.12: 18738595-18738595 on Build GRCh38
rs761872921 Chr.12: 18738566-18738566 on Build GRCh38
rs868817065 Chr.12: 18738789-18738789 on Build GRCh38
rs1035766955 Chr.12: 18738619-18738619 on Build GRCh38
rs753311519 Chr.12: 18738792-18738792 on Build GRCh38
rs370457183 Chr.12: 18738826-18738826 on Build GRCh38
rs750504721 Chr.12: 18738518-18738518 on Build GRCh38
rs376092094 Chr.12: 18738519-18738519 on Build GRCh38
rs140506801 Chr.12: 18738738-18738738 on Build GRCh38
rs778206137 Chr.12: 18738514-18738515 on Build GRCh38
rs774397273 Chr.12: 18738747-18738747 on Build GRCh38
rs757346159 Chr.12: 18738512-18738512 on Build GRCh38
rs61740731 Chr.12: 18738833-18738833 on Build GRCh38
rs752979803 Chr.12: 18738719-18738719 on Build GRCh38
rs759527168 Chr.12: 18738873-18738873 on Build GRCh38
rs372878425 Chr.12: 18738511-18738511 on Build GRCh38
rs763156262 Chr.12: 18738465-18738465 on Build GRCh38
rs1002995382 Chr.12: 18738563-18738563 on Build GRCh38
rs765628337 Chr.12: 18738770-18738770 on Build GRCh38
rs781037697 Chr.12: 18738634-18738634 on Build GRCh38
rs746840009 Chr.12: 18738860-18738860 on Build GRCh38
rs865845647 Chr.12: 18738737-18738737 on Build GRCh38
rs377652683 Chr.12: 18738712-18738712 on Build GRCh38
rs374422807 Chr.12: 18738685-18738685 on Build GRCh38
rs867346742 Chr.12: 18738515-18738515 on Build GRCh38
rs902648501 Chr.12: 18738854-18738854 on Build GRCh38
rs770419225 Chr.12: 18738464-18738464 on Build GRCh38
rs867785228 Chr.12: 18738827-18738827 on Build GRCh38
rs149089246 Chr.12: 18738831-18738831 on Build GRCh38
rs376550649 Chr.12: 18738481-18738481 on Build GRCh38
rs752977813 Chr.12: 18738459-18738459 on Build GRCh38
rs145563800 Chr.12: 18738753-18738753 on Build GRCh38
rs758818877 Chr.12: 18738807-18738807 on Build GRCh38
rs369230199 Chr.12: 18738532-18738532 on Build GRCh38
rs769828898 Chr.12: 18738547-18738547 on Build GRCh38
rs371563189 Chr.12: 18738670-18738670 on Build GRCh38
rs11547467 Chr.12: 18738551-18738551 on Build GRCh38
rs765272253 Chr.12: 18738495-18738495 on Build GRCh38
rs941964125 Chr.12: 18738801-18738801 on Build GRCh38
rs866412879 Chr.12: 18738498-18738498 on Build GRCh38
rs761692565 Chr.12: 18738651-18738651 on Build GRCh38
rs773409877 Chr.12: 18738759-18738759 on Build GRCh38
rs776217578 Chr.12: 18738536-18738536 on Build GRCh38
rs143115929 Chr.12: 18738841-18738841 on Build GRCh38
rs779092742 Chr.12: 18738617-18738617 on Build GRCh38
rs976893520 Chr.12: 18738607-18738607 on Build GRCh38
rs1021106187 Chr.12: 18738848-18738848 on Build GRCh38
rs780562766 Chr.12: 18738521-18738521 on Build GRCh38
rs773464750 Chr.12: 18738669-18738669 on Build GRCh38
rs756237408 Chr.12: 18738589-18738589 on Build GRCh38
rs183467731 Chr.12: 18738597-18738597 on Build GRCh38
rs200625390 Chr.12: 18738637-18738637 on Build GRCh38
rs762800056 Chr.12: 18738550-18738550 on Build GRCh38
rs768941814 Chr.12: 18738523-18738523 on Build GRCh38
rs756823096 Chr.12: 18738624-18738624 on Build GRCh38
rs766596930 Chr.12: 18738765-18738765 on Build GRCh38
rs759136702 Chr.12: 18738537-18738537 on Build GRCh38
rs918403821 Chr.12: 18738757-18738757 on Build GRCh38
rs750455997 Chr.12: 18738585-18738585 on Build GRCh38
rs755377414 Chr.12: 18738679-18738679 on Build GRCh38
rs953806092 Chr.12: 18738671-18738671 on Build GRCh38
rs775763789 Chr.12: 18738442-18738442 on Build GRCh38
rs34456980 Chr.12: 18738769-18738769 on Build GRCh38
rs754370889 Chr.12: 18738600-18738600 on Build GRCh38
rs562040987 Chr.12: 18738477-18738477 on Build GRCh38
rs776949051 Chr.12: 18738862-18738862 on Build GRCh38

View Full Product Details