Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 26954065-26954578 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGFR1OP2
Entrez Gene ID
26127
Gene Name
FGFR1 oncogene partner 2
Gene Aliases
HSPC123-like, WIT3.0
UniGene
Hs.591162
Gene Chromosome Location
Chr.12: 26938372-26966648 on Build GRCh38
Transcripts
NM_001171887.1 NM_001171888.1 NM_015633.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4041106 c.70G>A p.D24N Chr.12: 26954228-26954228 on Build GRCh38
6445757 c.85C>G p.L29V Chr.12: 26954243-26954243 on Build GRCh38
6871547 c.5G>A p.S2N Chr.12: 26954163-26954163 on Build GRCh38
3954584 c.46G>T p.V16F Chr.12: 26954204-26954204 on Build GRCh38
4684569 c.109A>G p.N37D Chr.12: 26954267-26954267 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755685647 Chr.12: 26954172-26954172 on Build GRCh38
rs933781107 Chr.12: 26954551-26954551 on Build GRCh38
rs571282814 Chr.12: 26954404-26954404 on Build GRCh38
rs140186991 Chr.12: 26954189-26954189 on Build GRCh38
rs1027539484 Chr.12: 26954240-26954240 on Build GRCh38
rs116940418 Chr.12: 26954215-26954215 on Build GRCh38
rs142918295 Chr.12: 26954518-26954518 on Build GRCh38
rs761639868 Chr.12: 26954151-26954151 on Build GRCh38
rs191831172 Chr.12: 26954228-26954228 on Build GRCh38
rs751464736 Chr.12: 26954148-26954148 on Build GRCh38
rs960432142 Chr.12: 26954322-26954322 on Build GRCh38
rs1001853313 Chr.12: 26954306-26954306 on Build GRCh38
rs768554982 Chr.12: 26954110-26954114 on Build GRCh38
rs1037111658 Chr.12: 26954543-26954543 on Build GRCh38
rs373552993 Chr.12: 26954339-26954339 on Build GRCh38
rs767507489 Chr.12: 26954156-26954156 on Build GRCh38
rs747913774 Chr.12: 26954316-26954316 on Build GRCh38
rs2129092 Chr.12: 26954335-26954335 on Build GRCh38
rs978821357 Chr.12: 26954416-26954416 on Build GRCh38
rs1014012690 Chr.12: 26954364-26954364 on Build GRCh38
rs767322129 Chr.12: 26954321-26954321 on Build GRCh38
rs770227979 Chr.12: 26954190-26954190 on Build GRCh38
rs751885263 Chr.12: 26954471-26954471 on Build GRCh38
rs759883908 Chr.12: 26954112-26954112 on Build GRCh38
rs1037968880 Chr.12: 26954248-26954248 on Build GRCh38
rs891183685 Chr.12: 26954119-26954119 on Build GRCh38
rs975199675 Chr.12: 26954127-26954127 on Build GRCh38
rs772391363 Chr.12: 26954201-26954201 on Build GRCh38
rs975252085 Chr.12: 26954508-26954508 on Build GRCh38
rs921503674 Chr.12: 26954421-26954421 on Build GRCh38
rs780847728 Chr.12: 26954186-26954187 on Build GRCh38
rs375537398 Chr.12: 26954300-26954300 on Build GRCh38
rs932911238 Chr.12: 26954457-26954457 on Build GRCh38
rs759825448 Chr.12: 26954273-26954273 on Build GRCh38
rs899400122 Chr.12: 26954269-26954269 on Build GRCh38
rs922060464 Chr.12: 26954139-26954139 on Build GRCh38
rs767304440 Chr.12: 26954094-26954094 on Build GRCh38
rs776887161 Chr.12: 26954230-26954230 on Build GRCh38
rs914068018 Chr.12: 26954355-26954355 on Build GRCh38
rs761676055 Chr.12: 26954316-26954316 on Build GRCh38
rs770195742 Chr.12: 26954274-26954274 on Build GRCh38
rs922449902 Chr.12: 26954522-26954522 on Build GRCh38
rs376849887 Chr.12: 26954161-26954161 on Build GRCh38
rs753467930 Chr.12: 26954188-26954188 on Build GRCh38
rs746155998 Chr.12: 26954174-26954174 on Build GRCh38
rs531579559 Chr.12: 26954144-26954144 on Build GRCh38
rs764388090 Chr.12: 26954459-26954459 on Build GRCh38
rs980015237 Chr.12: 26954392-26954392 on Build GRCh38
rs369056114 Chr.12: 26954191-26954191 on Build GRCh38
rs747011880 Chr.12: 26954227-26954227 on Build GRCh38
rs375968651 Chr.12: 26954176-26954176 on Build GRCh38
rs1004285985 Chr.12: 26954126-26954126 on Build GRCh38
rs968277290 Chr.12: 26954383-26954383 on Build GRCh38
rs895601941 Chr.12: 26954347-26954347 on Build GRCh38
rs750400178 Chr.12: 26954334-26954334 on Build GRCh38

View Full Product Details