Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 53308871-53309381 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AAAS
Entrez Gene ID
8086
Gene Name
aladin WD repeat nucleoporin
Gene Aliases
AAA, AAASb, ADRACALA, ADRACALIN, ALADIN, GL003
UniGene
Hs.369144
Gene Chromosome Location
Chr.12: 53307456-53321628 on Build GRCh38
Transcripts
NM_001173466.1 NM_015665.5 XM_011538778.1 XM_011538780.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5361411 c.902A>G p.K301R Chr.12: 53309190-53309190 on Build GRCh38
3871944 c.919C>T p.P307S Chr.12: 53309173-53309173 on Build GRCh38
1932893 c.896G>A p.G299D Chr.12: 53309196-53309196 on Build GRCh38
940974 c.989G>A p.R330H Chr.12: 53308967-53308967 on Build GRCh38
6228354 c.880C>T p.L294F Chr.12: 53309212-53309212 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs560069464 Chr.12: 53308968-53308968 on Build GRCh38
rs748521896 Chr.12: 53309299-53309299 on Build GRCh38
rs1029967361 Chr.12: 53309157-53309157 on Build GRCh38
rs565082206 Chr.12: 53308988-53308988 on Build GRCh38
rs1001407309 Chr.12: 53309292-53309292 on Build GRCh38
rs749264112 Chr.12: 53309028-53309029 on Build GRCh38
rs760745488 Chr.12: 53309203-53309203 on Build GRCh38
rs200312077 Chr.12: 53308948-53308948 on Build GRCh38
rs879241808 Chr.12: 53309310-53309310 on Build GRCh38
rs200423748 Chr.12: 53309312-53309312 on Build GRCh38
rs748177964 Chr.12: 53309002-53309002 on Build GRCh38
rs1012756707 Chr.12: 53308984-53308984 on Build GRCh38
rs755685527 Chr.12: 53308940-53308941 on Build GRCh38
rs369629444 Chr.12: 53309021-53309021 on Build GRCh38
rs372459440 Chr.12: 53309267-53309267 on Build GRCh38
rs750478250 Chr.12: 53308955-53308955 on Build GRCh38
rs773601814 Chr.12: 53309018-53309018 on Build GRCh38
rs765338554 Chr.12: 53309235-53309235 on Build GRCh38
rs764523065 Chr.12: 53308919-53308919 on Build GRCh38
rs149487112 Chr.12: 53309273-53309273 on Build GRCh38
rs140360792 Chr.12: 53309197-53309197 on Build GRCh38
rs904042274 Chr.12: 53309070-53309070 on Build GRCh38
rs772890213 Chr.12: 53309006-53309006 on Build GRCh38
rs1016290579 Chr.12: 53308994-53308994 on Build GRCh38
rs199636211 Chr.12: 53309198-53309198 on Build GRCh38
rs552736260 Chr.12: 53309210-53309210 on Build GRCh38
rs776289940 Chr.12: 53309047-53309047 on Build GRCh38
rs756135188 Chr.12: 53309130-53309130 on Build GRCh38
rs777507615 Chr.12: 53308953-53308954 on Build GRCh38
rs900728531 Chr.12: 53308917-53308917 on Build GRCh38
rs766520045 Chr.12: 53309133-53309133 on Build GRCh38
rs201441867 Chr.12: 53309112-53309112 on Build GRCh38
rs150173795 Chr.12: 53308982-53308982 on Build GRCh38
rs774817639 Chr.12: 53309191-53309191 on Build GRCh38
rs754159917 Chr.12: 53309141-53309141 on Build GRCh38
rs753037128 Chr.12: 53308912-53308912 on Build GRCh38
rs1036729906 Chr.12: 53308899-53308899 on Build GRCh38
rs373652342 Chr.12: 53309090-53309090 on Build GRCh38
rs956959048 Chr.12: 53309143-53309143 on Build GRCh38
rs766542823 Chr.12: 53309207-53309207 on Build GRCh38
rs774564593 Chr.12: 53309069-53309069 on Build GRCh38
rs764585946 Chr.12: 53309061-53309061 on Build GRCh38
rs755270528 Chr.12: 53309216-53309216 on Build GRCh38
rs762121998 Chr.12: 53309193-53309193 on Build GRCh38
rs548425176 Chr.12: 53309152-53309152 on Build GRCh38
rs777806731 Chr.12: 53309169-53309169 on Build GRCh38
rs763230323 Chr.12: 53309052-53309052 on Build GRCh38
rs184264999 Chr.12: 53309123-53309123 on Build GRCh38
rs527404721 Chr.12: 53309189-53309189 on Build GRCh38
rs1039325300 Chr.12: 53308907-53308907 on Build GRCh38
rs387906326 Chr.12: 53308975-53308976 on Build GRCh38
rs770621532 Chr.12: 53309106-53309107 on Build GRCh38
rs79881935 Chr.12: 53309017-53309017 on Build GRCh38
rs138749872 Chr.12: 53309180-53309180 on Build GRCh38
rs762355009 Chr.12: 53309107-53309107 on Build GRCh38
rs778571464 Chr.12: 53308994-53308994 on Build GRCh38
rs774953260 Chr.12: 53309294-53309294 on Build GRCh38
rs938596715 Chr.12: 53309213-53309213 on Build GRCh38
rs746531962 Chr.12: 53309176-53309176 on Build GRCh38
rs909046941 Chr.12: 53309181-53309181 on Build GRCh38
rs372465790 Chr.12: 53309274-53309274 on Build GRCh38
rs769513649 Chr.12: 53309291-53309291 on Build GRCh38
rs189123555 Chr.12: 53309327-53309327 on Build GRCh38
rs768886793 Chr.12: 53309286-53309287 on Build GRCh38
rs773970701 Chr.12: 53309229-53309229 on Build GRCh38
rs121918547 Chr.12: 53309158-53309158 on Build GRCh38
rs145196232 Chr.12: 53309249-53309249 on Build GRCh38
rs1546808 Chr.12: 53309237-53309237 on Build GRCh38
rs747008621 Chr.12: 53309266-53309266 on Build GRCh38
rs145446970 Chr.12: 53309236-53309236 on Build GRCh38
rs745592391 Chr.12: 53309183-53309183 on Build GRCh38
rs755146590 Chr.12: 53309146-53309146 on Build GRCh38
rs756366173 Chr.12: 53308967-53308967 on Build GRCh38
rs547399432 Chr.12: 53309076-53309076 on Build GRCh38
rs111986621 Chr.12: 53308940-53308940 on Build GRCh38
rs772116799 Chr.12: 53309003-53309003 on Build GRCh38
rs377673792 Chr.12: 53309005-53309005 on Build GRCh38
rs868408682 Chr.12: 53309357-53309357 on Build GRCh38
rs200674403 Chr.12: 53308934-53308934 on Build GRCh38
rs371424478 Chr.12: 53308916-53308916 on Build GRCh38
rs772352555 Chr.12: 53309317-53309317 on Build GRCh38

View Full Product Details