Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 54182319-54182829 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMUG1
Entrez Gene ID
23583
Gene Name
single-strand-selective monofunctional uracil-DNA glycosylase 1
Gene Aliases
FDG, HMUDG, UNG3
UniGene
Hs.632721
Gene Chromosome Location
Chr.12: 54171815-54188994 on Build GRCh38
Transcripts
NM_001243787.1 NM_001243788.1 NM_001243789.1 NM_001243790.1 NM_001243791.1 NM_014311.2 NR_045039.1 XM_006719319.3 XM_006719320.3 XM_006719321.3 XM_006719322.3 XM_011538109.2 XM_011538110.2 XM_011538111.2 XM_011538112.2 XM_011538113.2 XM_011538114.2 XM_011538115.2 XM_011538116.2 XM_011538117.2 XM_011538118.2 XM_011538119.2 XM_011538120.2 XM_011538121.2 XM_011538122.2 XM_011538123.2 XM_017019113.1 XM_017019114.1 XM_017019115.1 XM_017019116.1 XM_017019117.1 XM_017019118.1 XM_017019119.1 XM_017019120.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5933157 c.514C>T p.P172S Chr.12: 54182395-54182395 on Build GRCh38
3704250 c.380T>G p.L127R Chr.12: 54182529-54182529 on Build GRCh38
3368867 c.398A>T p.Q133L Chr.12: 54182511-54182511 on Build GRCh38
1606334 c.380T>G p.L127R Chr.12: 54182529-54182529 on Build GRCh38
941094 c.562G>T p.E188* Chr.12: 54182347-54182347 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774126329 Chr.12: 54182755-54182755 on Build GRCh38
rs35971604 Chr.12: 54182429-54182429 on Build GRCh38
rs765345163 Chr.12: 54182663-54182663 on Build GRCh38
rs777128921 Chr.12: 54182643-54182643 on Build GRCh38
rs779147167 Chr.12: 54182388-54182388 on Build GRCh38
rs370518402 Chr.12: 54182472-54182472 on Build GRCh38
rs758324719 Chr.12: 54182499-54182499 on Build GRCh38
rs901745764 Chr.12: 54182724-54182724 on Build GRCh38
rs182375610 Chr.12: 54182805-54182805 on Build GRCh38
rs750929214 Chr.12: 54182383-54182383 on Build GRCh38
rs759602126 Chr.12: 54182489-54182489 on Build GRCh38
rs777923361 Chr.12: 54182392-54182392 on Build GRCh38
rs375365910 Chr.12: 54182582-54182582 on Build GRCh38
rs572090065 Chr.12: 54182796-54182796 on Build GRCh38
rs372577448 Chr.12: 54182771-54182771 on Build GRCh38
rs777682048 Chr.12: 54182510-54182510 on Build GRCh38
rs113375463 Chr.12: 54182671-54182671 on Build GRCh38
rs11538362 Chr.12: 54182531-54182531 on Build GRCh38
rs145157119 Chr.12: 54182790-54182793 on Build GRCh38
rs200031028 Chr.12: 54182360-54182360 on Build GRCh38
rs774605415 Chr.12: 54182359-54182359 on Build GRCh38
rs189765782 Chr.12: 54182675-54182675 on Build GRCh38
rs769345683 Chr.12: 54182573-54182573 on Build GRCh38
rs977548848 Chr.12: 54182635-54182635 on Build GRCh38
rs746922626 Chr.12: 54182401-54182401 on Build GRCh38
rs377427651 Chr.12: 54182435-54182435 on Build GRCh38
rs369292276 Chr.12: 54182691-54182691 on Build GRCh38
rs566454607 Chr.12: 54182549-54182549 on Build GRCh38
rs200527979 Chr.12: 54182436-54182436 on Build GRCh38
rs758198036 Chr.12: 54182390-54182390 on Build GRCh38
rs960521556 Chr.12: 54182377-54182377 on Build GRCh38
rs760798951 Chr.12: 54182384-54182384 on Build GRCh38
rs777098038 Chr.12: 54182447-54182447 on Build GRCh38
rs111953762 Chr.12: 54182645-54182645 on Build GRCh38
rs934112301 Chr.12: 54182530-54182530 on Build GRCh38
rs370951715 Chr.12: 54182628-54182628 on Build GRCh38
rs994231944 Chr.12: 54182602-54182602 on Build GRCh38
rs1023356208 Chr.12: 54182681-54182681 on Build GRCh38
rs1038254163 Chr.12: 54182590-54182590 on Build GRCh38
rs769447985 Chr.12: 54182350-54182350 on Build GRCh38
rs775158364 Chr.12: 54182491-54182491 on Build GRCh38
rs35326992 Chr.12: 54182553-54182554 on Build GRCh38
rs984545727 Chr.12: 54182512-54182512 on Build GRCh38
rs775799057 Chr.12: 54182376-54182377 on Build GRCh38
rs745501752 Chr.12: 54182468-54182468 on Build GRCh38
rs752448480 Chr.12: 54182389-54182389 on Build GRCh38
rs538667087 Chr.12: 54182733-54182733 on Build GRCh38
rs866561490 Chr.12: 54182503-54182503 on Build GRCh38
rs896206542 Chr.12: 54182646-54182646 on Build GRCh38
rs199767286 Chr.12: 54182473-54182473 on Build GRCh38
rs1040313539 Chr.12: 54182702-54182702 on Build GRCh38
rs186896912 Chr.12: 54182616-54182616 on Build GRCh38
rs201341981 Chr.12: 54182598-54182598 on Build GRCh38
rs747321776 Chr.12: 54182627-54182627 on Build GRCh38
rs368087027 Chr.12: 54182708-54182708 on Build GRCh38
rs755183387 Chr.12: 54182386-54182386 on Build GRCh38
rs775146467 Chr.12: 54182443-54182443 on Build GRCh38
rs112691523 Chr.12: 54182796-54182799 on Build GRCh38
rs139760820 Chr.12: 54182539-54182539 on Build GRCh38
rs772464266 Chr.12: 54182608-54182608 on Build GRCh38
rs771198309 Chr.12: 54182438-54182438 on Build GRCh38
rs781126578 Chr.12: 54182553-54182553 on Build GRCh38
rs943024969 Chr.12: 54182701-54182701 on Build GRCh38
rs1033669626 Chr.12: 54182801-54182801 on Build GRCh38
rs762234299 Chr.12: 54182445-54182445 on Build GRCh38
rs753881647 Chr.12: 54182481-54182481 on Build GRCh38
rs906018947 Chr.12: 54182704-54182704 on Build GRCh38
rs752957942 Chr.12: 54182494-54182494 on Build GRCh38
rs3136389 Chr.12: 54182596-54182596 on Build GRCh38
rs768020028 Chr.12: 54182380-54182380 on Build GRCh38
rs372461198 Chr.12: 54182421-54182421 on Build GRCh38
rs769373939 Chr.12: 54182471-54182471 on Build GRCh38
rs3136388 Chr.12: 54182795-54182798 on Build GRCh38
rs773755041 Chr.12: 54182456-54182456 on Build GRCh38
rs745378076 Chr.12: 54182555-54182555 on Build GRCh38
rs369762317 Chr.12: 54182664-54182664 on Build GRCh38
rs941192010 Chr.12: 54182587-54182587 on Build GRCh38
rs762184563 Chr.12: 54182376-54182376 on Build GRCh38
rs541683930 Chr.12: 54182385-54182385 on Build GRCh38
rs773593836 Chr.12: 54182621-54182621 on Build GRCh38
rs541473644 Chr.12: 54182536-54182536 on Build GRCh38
rs778086758 Chr.12: 54182349-54182349 on Build GRCh38

View Full Product Details