Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 94305899-94306398 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP83
Entrez Gene ID
51134
Gene Name
centrosomal protein 83
Gene Aliases
CCDC41, NPHP18, NY-REN-58
UniGene
Hs.279209
Gene Chromosome Location
Chr.12: 94268832-94459991 on Build GRCh38
Transcripts
XM_006719437.2 XM_017019385.1 XM_017019386.1 NM_005761.2 NR_037687.1 XM_006719186.3

Gene Symbol
PLXNC1
Entrez Gene ID
10154
Gene Name
plexin C1
Gene Aliases
CD232, PLXN-C1, VESPR
UniGene
Hs.584845
Gene Chromosome Location
Chr.12: 94148696-94307675 on Build GRCh38
Transcripts
XM_006719437.2 XM_017019385.1 XM_017019386.1 NM_005761.2 NR_037687.1 XM_006719186.3

SNP Detail

SNP ID Chromosome Location
rs1035357970 Chr.12: 94306138-94306138 on Build GRCh38
rs959671112 Chr.12: 94306148-94306148 on Build GRCh38
rs1527321 Chr.12: 94306254-94306254 on Build GRCh38
rs920688993 Chr.12: 94306297-94306297 on Build GRCh38
rs551025136 Chr.12: 94306110-94306110 on Build GRCh38
rs546988549 Chr.12: 94306041-94306041 on Build GRCh38
rs192925347 Chr.12: 94306334-94306334 on Build GRCh38
rs769581490 Chr.12: 94306300-94306300 on Build GRCh38
rs934935751 Chr.12: 94306229-94306229 on Build GRCh38
rs142423166 Chr.12: 94306214-94306214 on Build GRCh38
rs1014917863 Chr.12: 94306092-94306092 on Build GRCh38
rs978949260 Chr.12: 94306215-94306215 on Build GRCh38
rs987641062 Chr.12: 94306237-94306240 on Build GRCh38
rs1029178659 Chr.12: 94306130-94306130 on Build GRCh38
rs765513159 Chr.12: 94305964-94305964 on Build GRCh38
rs573843614 Chr.12: 94306308-94306308 on Build GRCh38
rs769891544 Chr.12: 94306211-94306212 on Build GRCh38
rs953502912 Chr.12: 94306211-94306211 on Build GRCh38
rs781166513 Chr.12: 94306242-94306242 on Build GRCh38
rs1042780918 Chr.12: 94306356-94306356 on Build GRCh38
rs754464197 Chr.12: 94306094-94306094 on Build GRCh38
rs886904779 Chr.12: 94306331-94306331 on Build GRCh38
rs578219459 Chr.12: 94305981-94305981 on Build GRCh38
rs1039070492 Chr.12: 94306030-94306030 on Build GRCh38
rs746566092 Chr.12: 94306080-94306080 on Build GRCh38
rs947156550 Chr.12: 94305931-94305931 on Build GRCh38
rs887025299 Chr.12: 94306067-94306067 on Build GRCh38
rs1022709704 Chr.12: 94306205-94306205 on Build GRCh38
rs1027358400 Chr.12: 94306063-94306063 on Build GRCh38
rs988924086 Chr.12: 94306258-94306258 on Build GRCh38
rs1005320315 Chr.12: 94306083-94306083 on Build GRCh38
rs149396943 Chr.12: 94305924-94305927 on Build GRCh38
rs1039121574 Chr.12: 94306291-94306292 on Build GRCh38
rs994892513 Chr.12: 94306118-94306118 on Build GRCh38
rs912178429 Chr.12: 94306222-94306222 on Build GRCh38
rs943457418 Chr.12: 94305982-94305982 on Build GRCh38
rs1012568046 Chr.12: 94306367-94306367 on Build GRCh38
rs1001417095 Chr.12: 94306001-94306001 on Build GRCh38
rs924891593 Chr.12: 94306227-94306227 on Build GRCh38
rs187835052 Chr.12: 94306280-94306280 on Build GRCh38
rs539051159 Chr.12: 94306226-94306227 on Build GRCh38
rs931174224 Chr.12: 94306301-94306301 on Build GRCh38
rs376674905 Chr.12: 94305926-94305929 on Build GRCh38
rs148765234 Chr.12: 94306060-94306060 on Build GRCh38
rs991251191 Chr.12: 94306170-94306170 on Build GRCh38
rs11107502 Chr.12: 94306043-94306043 on Build GRCh38
rs35061130 Chr.12: 94306208-94306209 on Build GRCh38
rs895513335 Chr.12: 94306348-94306348 on Build GRCh38

View Full Product Details