Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 94306256-94306787 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP83
Entrez Gene ID
51134
Gene Name
centrosomal protein 83
Gene Aliases
CCDC41, NPHP18, NY-REN-58
UniGene
Hs.279209
Gene Chromosome Location
Chr.12: 94268832-94459991 on Build GRCh38
Transcripts
XM_006719437.2 XM_017019385.1 XM_017019386.1 NM_005761.2 NR_037687.1 XM_006719186.3

Gene Symbol
PLXNC1
Entrez Gene ID
10154
Gene Name
plexin C1
Gene Aliases
CD232, PLXN-C1, VESPR
UniGene
Hs.584845
Gene Chromosome Location
Chr.12: 94148696-94307675 on Build GRCh38
Transcripts
XM_006719437.2 XM_017019385.1 XM_017019386.1 NM_005761.2 NR_037687.1 XM_006719186.3

SNP Detail

SNP ID Chromosome Location
rs1036774028 Chr.12: 94306502-94306502 on Build GRCh38
rs762021914 Chr.12: 94306545-94306545 on Build GRCh38
rs939824173 Chr.12: 94306625-94306625 on Build GRCh38
rs927145770 Chr.12: 94306376-94306376 on Build GRCh38
rs920688993 Chr.12: 94306297-94306297 on Build GRCh38
rs140244222 Chr.12: 94306562-94306562 on Build GRCh38
rs769581490 Chr.12: 94306300-94306300 on Build GRCh38
rs192925347 Chr.12: 94306334-94306334 on Build GRCh38
rs1012621200 Chr.12: 94306681-94306681 on Build GRCh38
rs904146642 Chr.12: 94306693-94306693 on Build GRCh38
rs570732919 Chr.12: 94306569-94306569 on Build GRCh38
rs1019122204 Chr.12: 94306683-94306683 on Build GRCh38
rs573843614 Chr.12: 94306308-94306308 on Build GRCh38
rs988378115 Chr.12: 94306697-94306697 on Build GRCh38
rs1042780918 Chr.12: 94306356-94306356 on Build GRCh38
rs886904779 Chr.12: 94306331-94306331 on Build GRCh38
rs895732852 Chr.12: 94306630-94306630 on Build GRCh38
rs912130739 Chr.12: 94306524-94306524 on Build GRCh38
rs545441695 Chr.12: 94306552-94306552 on Build GRCh38
rs1057095717 Chr.12: 94306627-94306627 on Build GRCh38
rs772248322 Chr.12: 94306542-94306544 on Build GRCh38
rs542845120 Chr.12: 94306600-94306600 on Build GRCh38
rs773373203 Chr.12: 94306443-94306444 on Build GRCh38
rs889634118 Chr.12: 94306619-94306638 on Build GRCh38
rs956377714 Chr.12: 94306497-94306497 on Build GRCh38
rs775748141 Chr.12: 94306577-94306577 on Build GRCh38
rs968490168 Chr.12: 94306443-94306443 on Build GRCh38
rs1039121574 Chr.12: 94306291-94306292 on Build GRCh38
rs777208772 Chr.12: 94306489-94306489 on Build GRCh38
rs957181974 Chr.12: 94306688-94306688 on Build GRCh38
rs1031806545 Chr.12: 94306749-94306749 on Build GRCh38
rs987695193 Chr.12: 94306516-94306516 on Build GRCh38
rs1012568046 Chr.12: 94306367-94306367 on Build GRCh38
rs11558081 Chr.12: 94306703-94306703 on Build GRCh38
rs1056879799 Chr.12: 94306416-94306416 on Build GRCh38
rs772144618 Chr.12: 94306568-94306568 on Build GRCh38
rs553314454 Chr.12: 94306380-94306380 on Build GRCh38
rs150903094 Chr.12: 94306458-94306458 on Build GRCh38
rs187835052 Chr.12: 94306280-94306280 on Build GRCh38
rs931174224 Chr.12: 94306301-94306301 on Build GRCh38
rs908384219 Chr.12: 94306620-94306620 on Build GRCh38
rs760885557 Chr.12: 94306704-94306704 on Build GRCh38
rs964722411 Chr.12: 94306534-94306534 on Build GRCh38
rs561577708 Chr.12: 94306722-94306722 on Build GRCh38
rs895513335 Chr.12: 94306348-94306348 on Build GRCh38
rs968386921 Chr.12: 94306708-94306708 on Build GRCh38

View Full Product Details