Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 109933907-109934453 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIT2
Entrez Gene ID
9815
Gene Name
GIT ArfGAP 2
Gene Aliases
CAT-2, CAT2, PKL
UniGene
Hs.434996
Gene Chromosome Location
Chr.12: 109929792-109996389 on Build GRCh38
Transcripts
NM_001135213.1 NM_001135214.1 NM_014776.3 NM_057169.3 NM_057170.3 XM_005253997.3 XM_005254000.3 XM_006719707.3 XM_006719708.3 XM_006719709.3 XM_006719712.3 XM_017020258.1 XM_017020259.1 XM_017020260.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6671105 c.2018C>T p.S673L Chr.12: 109934071-109934071 on Build GRCh38
6671107 c.1688C>T p.S563L Chr.12: 109934071-109934071 on Build GRCh38
6671106 c.1784C>T p.S595L Chr.12: 109934071-109934071 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1050400522 Chr.12: 109933942-109933942 on Build GRCh38
rs1024080489 Chr.12: 109934076-109934076 on Build GRCh38
rs539945346 Chr.12: 109934251-109934251 on Build GRCh38
rs1019570310 Chr.12: 109934254-109934254 on Build GRCh38
rs778246114 Chr.12: 109934119-109934119 on Build GRCh38
rs952594911 Chr.12: 109934369-109934369 on Build GRCh38
rs574148587 Chr.12: 109934246-109934246 on Build GRCh38
rs867608861 Chr.12: 109934065-109934065 on Build GRCh38
rs1034035301 Chr.12: 109934195-109934195 on Build GRCh38
rs984344277 Chr.12: 109934374-109934374 on Build GRCh38
rs1043336801 Chr.12: 109934183-109934183 on Build GRCh38
rs940557746 Chr.12: 109934317-109934317 on Build GRCh38
rs16940703 Chr.12: 109934092-109934092 on Build GRCh38
rs1031634680 Chr.12: 109934006-109934006 on Build GRCh38
rs947516472 Chr.12: 109934177-109934177 on Build GRCh38
rs890020486 Chr.12: 109934209-109934209 on Build GRCh38
rs756670903 Chr.12: 109934115-109934115 on Build GRCh38
rs182386350 Chr.12: 109934419-109934419 on Build GRCh38
rs764117262 Chr.12: 109934046-109934046 on Build GRCh38
rs189701508 Chr.12: 109934178-109934178 on Build GRCh38
rs7954192 Chr.12: 109934132-109934132 on Build GRCh38
rs377331684 Chr.12: 109934375-109934375 on Build GRCh38
rs757503016 Chr.12: 109934130-109934130 on Build GRCh38
rs146384144 Chr.12: 109934057-109934057 on Build GRCh38
rs984068855 Chr.12: 109933934-109933934 on Build GRCh38
rs751683768 Chr.12: 109934078-109934078 on Build GRCh38
rs745466425 Chr.12: 109934135-109934135 on Build GRCh38
rs891979346 Chr.12: 109934158-109934158 on Build GRCh38
rs778647308 Chr.12: 109934133-109934133 on Build GRCh38
rs775595755 Chr.12: 109933977-109933977 on Build GRCh38
rs7970705 Chr.12: 109933967-109933967 on Build GRCh38
rs951658778 Chr.12: 109934210-109934210 on Build GRCh38
rs903841979 Chr.12: 109933946-109933946 on Build GRCh38
rs766639068 Chr.12: 109934060-109934060 on Build GRCh38
rs145147322 Chr.12: 109934297-109934297 on Build GRCh38
rs16939769 Chr.12: 109934405-109934405 on Build GRCh38
rs7954196 Chr.12: 109934144-109934144 on Build GRCh38
rs373918020 Chr.12: 109933975-109933975 on Build GRCh38
rs780337150 Chr.12: 109934058-109934058 on Build GRCh38
rs749666530 Chr.12: 109934127-109934127 on Build GRCh38
rs760748585 Chr.12: 109933991-109933991 on Build GRCh38
rs901141645 Chr.12: 109934295-109934295 on Build GRCh38
rs544371063 Chr.12: 109934339-109934339 on Build GRCh38
rs573902552 Chr.12: 109934302-109934302 on Build GRCh38
rs7974577 Chr.12: 109934204-109934204 on Build GRCh38
rs1006925610 Chr.12: 109934252-109934252 on Build GRCh38
rs7971068 Chr.12: 109934194-109934194 on Build GRCh38
rs1025655729 Chr.12: 109934341-109934341 on Build GRCh38
rs923077491 Chr.12: 109934404-109934404 on Build GRCh38
rs927618978 Chr.12: 109934421-109934421 on Build GRCh38
rs572943123 Chr.12: 109934220-109934220 on Build GRCh38
rs979820396 Chr.12: 109934102-109934102 on Build GRCh38

View Full Product Details