Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 46833662-46834161 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HTR2A
Entrez Gene ID
3356
Gene Name
5-hydroxytryptamine receptor 2A
Gene Aliases
5-HT2A, HTR2
UniGene
Hs.72630
Gene Chromosome Location
Chr.13: 46831542-46897076 on Build GRCh38
Transcripts
NM_000621.4 NM_001165947.2

SNP Detail

SNP ID Chromosome Location
rs77274377 Chr.13: 46834091-46834091 on Build GRCh38
rs1014491709 Chr.13: 46833870-46833870 on Build GRCh38
rs760491424 Chr.13: 46833960-46833960 on Build GRCh38
rs143950250 Chr.13: 46833807-46833807 on Build GRCh38
rs539553503 Chr.13: 46833993-46833993 on Build GRCh38
rs905711864 Chr.13: 46833981-46833981 on Build GRCh38
rs914591173 Chr.13: 46833831-46833831 on Build GRCh38
rs148625875 Chr.13: 46834114-46834114 on Build GRCh38
rs75604221 Chr.13: 46834042-46834042 on Build GRCh38
rs542054192 Chr.13: 46834129-46834129 on Build GRCh38
rs983166852 Chr.13: 46833718-46833718 on Build GRCh38
rs932526639 Chr.13: 46833858-46833858 on Build GRCh38
rs1036851852 Chr.13: 46833910-46833910 on Build GRCh38
rs528765432 Chr.13: 46833799-46833799 on Build GRCh38
rs149887481 Chr.13: 46833806-46833806 on Build GRCh38
rs776309770 Chr.13: 46834016-46834016 on Build GRCh38
rs976973504 Chr.13: 46833748-46833749 on Build GRCh38
rs190839539 Chr.13: 46834134-46834134 on Build GRCh38
rs912507917 Chr.13: 46833869-46833869 on Build GRCh38
rs896889559 Chr.13: 46833928-46833928 on Build GRCh38
rs574217885 Chr.13: 46833882-46833882 on Build GRCh38
rs995237387 Chr.13: 46833956-46833956 on Build GRCh38
rs965792307 Chr.13: 46834095-46834095 on Build GRCh38
rs1023214751 Chr.13: 46834019-46834019 on Build GRCh38
rs950368602 Chr.13: 46833711-46833711 on Build GRCh38
rs557169652 Chr.13: 46833903-46833903 on Build GRCh38
rs552886187 Chr.13: 46834111-46834111 on Build GRCh38
rs1027302440 Chr.13: 46833684-46833684 on Build GRCh38
rs994719735 Chr.13: 46833999-46833999 on Build GRCh38
rs943865185 Chr.13: 46833877-46833877 on Build GRCh38
rs775248657 Chr.13: 46833735-46833735 on Build GRCh38
rs557694225 Chr.13: 46834018-46834018 on Build GRCh38
rs911681185 Chr.13: 46833734-46833734 on Build GRCh38
rs575813702 Chr.13: 46833890-46833890 on Build GRCh38
rs1049668227 Chr.13: 46833865-46833865 on Build GRCh38
rs1053130512 Chr.13: 46833836-46833836 on Build GRCh38
rs894819801 Chr.13: 46834012-46834012 on Build GRCh38
rs766444467 Chr.13: 46833997-46833997 on Build GRCh38
rs924682901 Chr.13: 46833769-46833769 on Build GRCh38
rs547343985 Chr.13: 46833801-46833801 on Build GRCh38
rs934694804 Chr.13: 46833822-46833822 on Build GRCh38
rs1003029566 Chr.13: 46834059-46834059 on Build GRCh38
rs1004573817 Chr.13: 46834062-46834062 on Build GRCh38
rs569285914 Chr.13: 46833913-46833913 on Build GRCh38
rs932895209 Chr.13: 46833796-46833796 on Build GRCh38
rs981646447 Chr.13: 46833745-46833745 on Build GRCh38

View Full Product Details