Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 52703178-52703676 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LECT1
Entrez Gene ID
11061
Gene Name
leukocyte cell derived chemotaxin 1
Gene Aliases
BRICD3, CHM-I, CHM1, MYETS1
UniGene
Hs.421391
Gene Chromosome Location
Chr.13: 52703264-52739812 on Build GRCh38
Transcripts
NM_001011705.1 NM_007015.2 XM_011534897.2 XM_011534898.2 XM_011534899.2 XM_011534900.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5023607 c.990C>T p.I330I Chr.13: 52703610-52703610 on Build GRCh38
5967728 c.973T>G p.W325G Chr.13: 52703627-52703627 on Build GRCh38
6042753 c.948C>A p.A316A Chr.13: 52703652-52703652 on Build GRCh38
6700361 c.931C>A p.Q311K Chr.13: 52703669-52703669 on Build GRCh38
371736 c.979G>A p.V327M Chr.13: 52703621-52703621 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770828040 Chr.13: 52703641-52703641 on Build GRCh38
rs1035986975 Chr.13: 52703636-52703636 on Build GRCh38
rs762304496 Chr.13: 52703547-52703547 on Build GRCh38
rs201954506 Chr.13: 52703605-52703605 on Build GRCh38
rs780788020 Chr.13: 52703649-52703649 on Build GRCh38
rs766477684 Chr.13: 52703576-52703576 on Build GRCh38
rs17541412 Chr.13: 52703537-52703537 on Build GRCh38
rs201786976 Chr.13: 52703276-52703277 on Build GRCh38
rs939939790 Chr.13: 52703365-52703365 on Build GRCh38
rs71772282 Chr.13: 52703277-52703278 on Build GRCh38
rs201144209 Chr.13: 52703275-52703276 on Build GRCh38
rs757979532 Chr.13: 52703302-52703302 on Build GRCh38
rs959966857 Chr.13: 52703646-52703646 on Build GRCh38
rs777466607 Chr.13: 52703637-52703637 on Build GRCh38
rs769580305 Chr.13: 52703544-52703544 on Build GRCh38
rs767864633 Chr.13: 52703549-52703549 on Build GRCh38
rs530692435 Chr.13: 52703398-52703398 on Build GRCh38
rs9536243 Chr.13: 52703504-52703504 on Build GRCh38
rs758188708 Chr.13: 52703633-52703633 on Build GRCh38
rs941337661 Chr.13: 52703391-52703391 on Build GRCh38
rs991196489 Chr.13: 52703204-52703204 on Build GRCh38
rs952535622 Chr.13: 52703274-52703274 on Build GRCh38
rs9536244 Chr.13: 52703511-52703511 on Build GRCh38
rs557654998 Chr.13: 52703312-52703312 on Build GRCh38
rs78575404 Chr.13: 52703655-52703655 on Build GRCh38
rs761127119 Chr.13: 52703556-52703556 on Build GRCh38
rs999518543 Chr.13: 52703550-52703550 on Build GRCh38
rs371116344 Chr.13: 52703616-52703616 on Build GRCh38
rs201204111 Chr.13: 52703231-52703234 on Build GRCh38
rs970216485 Chr.13: 52703231-52703232 on Build GRCh38
rs545526489 Chr.13: 52703360-52703362 on Build GRCh38
rs775724574 Chr.13: 52703215-52703215 on Build GRCh38
rs774793940 Chr.13: 52703546-52703546 on Build GRCh38
rs184520481 Chr.13: 52703405-52703405 on Build GRCh38
rs766094871 Chr.13: 52703450-52703450 on Build GRCh38
rs772880438 Chr.13: 52703630-52703631 on Build GRCh38
rs750923573 Chr.13: 52703552-52703552 on Build GRCh38
rs983943402 Chr.13: 52703281-52703281 on Build GRCh38
rs1052307 Chr.13: 52703577-52703577 on Build GRCh38
rs1035948227 Chr.13: 52703254-52703254 on Build GRCh38
rs779135748 Chr.13: 52703615-52703615 on Build GRCh38
rs746958626 Chr.13: 52703638-52703638 on Build GRCh38
rs566918510 Chr.13: 52703522-52703522 on Build GRCh38
rs753976938 Chr.13: 52703614-52703614 on Build GRCh38
rs903403001 Chr.13: 52703543-52703543 on Build GRCh38
rs555908353 Chr.13: 52703248-52703249 on Build GRCh38
rs531221574 Chr.13: 52703215-52703216 on Build GRCh38
rs570303464 Chr.13: 52703469-52703469 on Build GRCh38
rs909160549 Chr.13: 52703282-52703282 on Build GRCh38
rs563834252 Chr.13: 52703258-52703258 on Build GRCh38
rs891271512 Chr.13: 52703499-52703499 on Build GRCh38
rs1036325231 Chr.13: 52703384-52703384 on Build GRCh38
rs932739851 Chr.13: 52703466-52703466 on Build GRCh38
rs769459626 Chr.13: 52703612-52703613 on Build GRCh38
rs374765429 Chr.13: 52703493-52703493 on Build GRCh38
rs1052308 Chr.13: 52703342-52703342 on Build GRCh38
rs750854123 Chr.13: 52703255-52703255 on Build GRCh38
rs561667817 Chr.13: 52703395-52703395 on Build GRCh38
rs765460181 Chr.13: 52703227-52703227 on Build GRCh38
rs1132184 Chr.13: 52703287-52703287 on Build GRCh38

View Full Product Details