Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 110462186-110462695 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL4A2
Entrez Gene ID
1284
Gene Name
collagen type IV alpha 2
Gene Aliases
ICH, POREN2
UniGene
Hs.508716
Gene Chromosome Location
Chr.13: 110307284-110513027 on Build GRCh38
Transcripts
NM_001267044.1

Gene Symbol
COL4A2-AS2
Entrez Gene ID
100129836
Gene Name
COL4A2 antisense RNA 2
Gene Aliases
-
UniGene
Hs.640035
Gene Chromosome Location
Chr.13: 110456397-110463287 on Build GRCh38
Transcripts
NM_001267044.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6138545 c.1720G>A p.D574N Chr.13: 110462328-110462328 on Build GRCh38
6138546 c.1676G>T p.R559L Chr.13: 110462284-110462284 on Build GRCh38
3467225 c.1755C>T p.F585F Chr.13: 110462363-110462363 on Build GRCh38
6633170 c.1687G>A p.G563S Chr.13: 110462295-110462295 on Build GRCh38
2263101 c.1704C>T p.P568P Chr.13: 110462312-110462312 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749031666 Chr.13: 110462403-110462403 on Build GRCh38
rs532970219 Chr.13: 110462482-110462482 on Build GRCh38
rs1054784239 Chr.13: 110462540-110462540 on Build GRCh38
rs760848044 Chr.13: 110462355-110462355 on Build GRCh38
rs780963953 Chr.13: 110462333-110462333 on Build GRCh38
rs7992086 Chr.13: 110462573-110462573 on Build GRCh38
rs770923283 Chr.13: 110462507-110462507 on Build GRCh38
rs751105211 Chr.13: 110462328-110462328 on Build GRCh38
rs142749554 Chr.13: 110462582-110462582 on Build GRCh38
rs1050643583 Chr.13: 110462647-110462647 on Build GRCh38
rs767094122 Chr.13: 110462384-110462384 on Build GRCh38
rs1032622522 Chr.13: 110462649-110462649 on Build GRCh38
rs772023099 Chr.13: 110462329-110462330 on Build GRCh38
rs948529464 Chr.13: 110462291-110462291 on Build GRCh38
rs886049973 Chr.13: 110462314-110462314 on Build GRCh38
rs757806507 Chr.13: 110462217-110462217 on Build GRCh38
rs942259650 Chr.13: 110462514-110462514 on Build GRCh38
rs148511298 Chr.13: 110462412-110462412 on Build GRCh38
rs368047200 Chr.13: 110462246-110462246 on Build GRCh38
rs886049974 Chr.13: 110462380-110462380 on Build GRCh38
rs752922363 Chr.13: 110462542-110462542 on Build GRCh38
rs1056934875 Chr.13: 110462589-110462589 on Build GRCh38
rs937803017 Chr.13: 110462520-110462520 on Build GRCh38
rs759668716 Chr.13: 110462277-110462277 on Build GRCh38
rs199574157 Chr.13: 110462284-110462284 on Build GRCh38
rs374304814 Chr.13: 110462298-110462298 on Build GRCh38
rs765791292 Chr.13: 110462313-110462313 on Build GRCh38
rs772031454 Chr.13: 110462415-110462415 on Build GRCh38
rs757092411 Chr.13: 110462559-110462559 on Build GRCh38
rs762299284 Chr.13: 110462362-110462362 on Build GRCh38
rs367811110 Chr.13: 110462427-110462427 on Build GRCh38
rs368207474 Chr.13: 110462356-110462356 on Build GRCh38
rs113219763 Chr.13: 110462386-110462386 on Build GRCh38
rs754604565 Chr.13: 110462330-110462330 on Build GRCh38
rs747751731 Chr.13: 110462338-110462338 on Build GRCh38
rs897838617 Chr.13: 110462316-110462316 on Build GRCh38
rs371324929 Chr.13: 110462263-110462263 on Build GRCh38
rs905241418 Chr.13: 110462295-110462295 on Build GRCh38
rs777538816 Chr.13: 110462398-110462398 on Build GRCh38
rs765662785 Chr.13: 110462366-110462366 on Build GRCh38
rs886039602 Chr.13: 110462385-110462385 on Build GRCh38
rs1049098842 Chr.13: 110462633-110462633 on Build GRCh38
rs551461632 Chr.13: 110462488-110462488 on Build GRCh38
rs781053670 Chr.13: 110462231-110462231 on Build GRCh38
rs754261920 Chr.13: 110462310-110462310 on Build GRCh38
rs374871266 Chr.13: 110462208-110462208 on Build GRCh38
rs7992707 Chr.13: 110462616-110462616 on Build GRCh38
rs777360782 Chr.13: 110462344-110462344 on Build GRCh38
rs924275183 Chr.13: 110462430-110462430 on Build GRCh38
rs755661629 Chr.13: 110462389-110462389 on Build GRCh38
rs770760523 Chr.13: 110462345-110462345 on Build GRCh38
rs768370216 Chr.13: 110462228-110462228 on Build GRCh38
rs948623490 Chr.13: 110462524-110462524 on Build GRCh38
rs112264778 Chr.13: 110462659-110462660 on Build GRCh38
rs779393349 Chr.13: 110462219-110462219 on Build GRCh38
rs770902143 Chr.13: 110462269-110462269 on Build GRCh38
rs775569965 Chr.13: 110462354-110462354 on Build GRCh38
rs745758837 Chr.13: 110462413-110462413 on Build GRCh38
rs1044293547 Chr.13: 110462294-110462294 on Build GRCh38
rs9588174 Chr.13: 110462666-110462666 on Build GRCh38
rs745914600 Chr.13: 110462348-110462348 on Build GRCh38
rs41275110 Chr.13: 110462207-110462207 on Build GRCh38
rs766981913 Chr.13: 110462346-110462346 on Build GRCh38
rs776804161 Chr.13: 110462359-110462359 on Build GRCh38
rs914491654 Chr.13: 110462499-110462499 on Build GRCh38
rs747956626 Chr.13: 110462232-110462232 on Build GRCh38
rs772187018 Chr.13: 110462351-110462351 on Build GRCh38
rs774236731 Chr.13: 110462274-110462274 on Build GRCh38
rs561349636 Chr.13: 110462367-110462367 on Build GRCh38
rs534913617 Chr.13: 110462657-110462657 on Build GRCh38
rs927459902 Chr.13: 110462518-110462518 on Build GRCh38
rs754354411 Chr.13: 110462212-110462212 on Build GRCh38
rs765587454 Chr.13: 110462511-110462511 on Build GRCh38
rs908141502 Chr.13: 110462504-110462512 on Build GRCh38
rs755820436 Chr.13: 110462343-110462343 on Build GRCh38
rs757638360 Chr.13: 110462312-110462312 on Build GRCh38
rs1000755427 Chr.13: 110462646-110462646 on Build GRCh38
rs769757328 Chr.13: 110462254-110462254 on Build GRCh38
rs761007313 Chr.13: 110462297-110462297 on Build GRCh38
rs955878003 Chr.13: 110462487-110462487 on Build GRCh38
rs887991199 Chr.13: 110462648-110462648 on Build GRCh38
rs973989117 Chr.13: 110462515-110462516 on Build GRCh38
rs867029393 Chr.13: 110462283-110462283 on Build GRCh38
rs780042932 Chr.13: 110462416-110462416 on Build GRCh38
rs541679551 Chr.13: 110462607-110462607 on Build GRCh38
rs746522177 Chr.13: 110462222-110462222 on Build GRCh38
rs7992330 Chr.13: 110462404-110462404 on Build GRCh38
rs752255907 Chr.13: 110462388-110462388 on Build GRCh38
rs761151294 Chr.13: 110462260-110462260 on Build GRCh38
rs189622970 Chr.13: 110462209-110462209 on Build GRCh38
rs567388740 Chr.13: 110462656-110462656 on Build GRCh38
rs767721159 Chr.13: 110462279-110462279 on Build GRCh38
rs1017730027 Chr.13: 110462382-110462382 on Build GRCh38

View Full Product Details