Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 110627432-110627926 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAXD
Entrez Gene ID
55739
Gene Name
NAD(P)HX dehydratase
Gene Aliases
CARKD, LP3298
UniGene
Hs.408324
Gene Chromosome Location
Chr.13: 110615460-110639996 on Build GRCh38
Transcripts
NM_001242881.1 NM_001242882.1 NM_001242883.1 NM_018210.3 NR_040103.1 NR_040104.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
344238 c.414G>A p.V138V Chr.13: 110627466-110627466 on Build GRCh38
4611505 c.445_447delGTC p.V150delV Chr.13: 110627497-110627499 on Build GRCh38
945385 c.404T>C p.V135A Chr.13: 110627456-110627456 on Build GRCh38
6402970 c.449T>C p.V150A Chr.13: 110627501-110627501 on Build GRCh38
7271627 c.417G>A p.E139E Chr.13: 110627469-110627469 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1044112 Chr.13: 110627506-110627506 on Build GRCh38
rs750729449 Chr.13: 110627502-110627502 on Build GRCh38
rs145070867 Chr.13: 110627597-110627597 on Build GRCh38
rs546818831 Chr.13: 110627742-110627742 on Build GRCh38
rs1048937957 Chr.13: 110627859-110627859 on Build GRCh38
rs767422095 Chr.13: 110627479-110627479 on Build GRCh38
rs750632866 Chr.13: 110627593-110627593 on Build GRCh38
rs988929666 Chr.13: 110627894-110627894 on Build GRCh38
rs909122190 Chr.13: 110627875-110627875 on Build GRCh38
rs1044130080 Chr.13: 110627555-110627555 on Build GRCh38
rs41306678 Chr.13: 110627500-110627500 on Build GRCh38
rs553217045 Chr.13: 110627469-110627469 on Build GRCh38
rs759336239 Chr.13: 110627562-110627562 on Build GRCh38
rs3742194 Chr.13: 110627731-110627731 on Build GRCh38
rs764210701 Chr.13: 110627490-110627490 on Build GRCh38
rs1009785187 Chr.13: 110627757-110627757 on Build GRCh38
rs3742191 Chr.13: 110627470-110627470 on Build GRCh38
rs976332651 Chr.13: 110627478-110627478 on Build GRCh38
rs965852687 Chr.13: 110627685-110627685 on Build GRCh38
rs758717285 Chr.13: 110627516-110627516 on Build GRCh38
rs188583354 Chr.13: 110627626-110627626 on Build GRCh38
rs969724949 Chr.13: 110627828-110627828 on Build GRCh38
rs549417346 Chr.13: 110627690-110627690 on Build GRCh38
rs779036921 Chr.13: 110627670-110627670 on Build GRCh38
rs909150519 Chr.13: 110627824-110627824 on Build GRCh38
rs767943607 Chr.13: 110627762-110627763 on Build GRCh38
rs992241950 Chr.13: 110627804-110627804 on Build GRCh38
rs960479232 Chr.13: 110627771-110627771 on Build GRCh38
rs767262971 Chr.13: 110627603-110627603 on Build GRCh38
rs541251719 Chr.13: 110627614-110627614 on Build GRCh38
rs576045093 Chr.13: 110627528-110627528 on Build GRCh38
rs76068264 Chr.13: 110627623-110627623 on Build GRCh38
rs939036163 Chr.13: 110627638-110627638 on Build GRCh38
rs201831524 Chr.13: 110627483-110627483 on Build GRCh38
rs773880857 Chr.13: 110627557-110627557 on Build GRCh38
rs888537853 Chr.13: 110627476-110627476 on Build GRCh38
rs146504568 Chr.13: 110627482-110627482 on Build GRCh38
rs972573485 Chr.13: 110627648-110627648 on Build GRCh38
rs765455122 Chr.13: 110627592-110627592 on Build GRCh38
rs766274714 Chr.13: 110627467-110627467 on Build GRCh38
rs770196268 Chr.13: 110627761-110627761 on Build GRCh38
rs561062257 Chr.13: 110627723-110627723 on Build GRCh38
rs749047531 Chr.13: 110627589-110627589 on Build GRCh38
rs938468147 Chr.13: 110627891-110627891 on Build GRCh38
rs929242766 Chr.13: 110627833-110627833 on Build GRCh38
rs545067600 Chr.13: 110627640-110627640 on Build GRCh38
rs543549091 Chr.13: 110627581-110627581 on Build GRCh38
rs984729085 Chr.13: 110627780-110627780 on Build GRCh38
rs3742192 Chr.13: 110627497-110627497 on Build GRCh38
rs866367234 Chr.13: 110627462-110627462 on Build GRCh38
rs964362259 Chr.13: 110627488-110627488 on Build GRCh38
rs377595355 Chr.13: 110627454-110627454 on Build GRCh38
rs968234841 Chr.13: 110627760-110627760 on Build GRCh38
rs371103258 Chr.13: 110627552-110627552 on Build GRCh38
rs764143271 Chr.13: 110627579-110627579 on Build GRCh38
rs918889996 Chr.13: 110627805-110627805 on Build GRCh38
rs373944589 Chr.13: 110627580-110627580 on Build GRCh38
rs771788978 Chr.13: 110627566-110627566 on Build GRCh38
rs746257501 Chr.13: 110627743-110627743 on Build GRCh38
rs887586991 Chr.13: 110627662-110627662 on Build GRCh38
rs377430322 Chr.13: 110627521-110627521 on Build GRCh38
rs3742193 Chr.13: 110627655-110627655 on Build GRCh38
rs757530909 Chr.13: 110627499-110627499 on Build GRCh38
rs957186890 Chr.13: 110627736-110627736 on Build GRCh38
rs1056811855 Chr.13: 110627468-110627468 on Build GRCh38
rs751918279 Chr.13: 110627751-110627752 on Build GRCh38
rs113263042 Chr.13: 110627596-110627596 on Build GRCh38
rs534898507 Chr.13: 110627459-110627459 on Build GRCh38
rs775478477 Chr.13: 110627568-110627568 on Build GRCh38
rs775804662 Chr.13: 110627907-110627907 on Build GRCh38
rs148946345 Chr.13: 110627650-110627650 on Build GRCh38
rs12184786 Chr.13: 110627728-110627728 on Build GRCh38
rs1048872804 Chr.13: 110627472-110627472 on Build GRCh38
rs998633657 Chr.13: 110627724-110627724 on Build GRCh38
rs928388068 Chr.13: 110627571-110627571 on Build GRCh38
rs1005605646 Chr.13: 110627683-110627683 on Build GRCh38
rs417714 Chr.13: 110627843-110627843 on Build GRCh38
rs762733351 Chr.13: 110627460-110627460 on Build GRCh38
rs929205379 Chr.13: 110627652-110627652 on Build GRCh38
rs376375207 Chr.13: 110627666-110627670 on Build GRCh38
rs1039793448 Chr.13: 110627696-110627696 on Build GRCh38
rs928366019 Chr.13: 110627836-110627836 on Build GRCh38
rs777490219 Chr.13: 110627529-110627529 on Build GRCh38
rs926403023 Chr.13: 110627769-110627769 on Build GRCh38
rs760647801 Chr.13: 110627575-110627575 on Build GRCh38
rs1022482236 Chr.13: 110627758-110627758 on Build GRCh38

View Full Product Details