Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 52439072-52439604 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TXNDC16
Entrez Gene ID
57544
Gene Name
thioredoxin domain containing 16
Gene Aliases
ERp90, KIAA1344
UniGene
Hs.532609
Gene Chromosome Location
Chr.14: 52430590-52552583 on Build GRCh38
Transcripts
NM_001160047.1 NM_020784.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
956224 c.2065C>A p.P689T Chr.14: 52439333-52439333 on Build GRCh38
4150807 c.2004-9C>G p.? Chr.14: 52439403-52439403 on Build GRCh38
3814930 c.2163G>T p.K721N Chr.14: 52439235-52439235 on Build GRCh38
6001154 c.2048A>T p.D683V Chr.14: 52439350-52439350 on Build GRCh38
7326861 c.2179C>A p.L727I Chr.14: 52439219-52439219 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766548813 Chr.14: 52439198-52439198 on Build GRCh38
rs199628626 Chr.14: 52439311-52439311 on Build GRCh38
rs28475854 Chr.14: 52439541-52439541 on Build GRCh38
rs1026731548 Chr.14: 52439354-52439354 on Build GRCh38
rs749284011 Chr.14: 52439542-52439542 on Build GRCh38
rs949101979 Chr.14: 52439475-52439475 on Build GRCh38
rs1020625418 Chr.14: 52439118-52439118 on Build GRCh38
rs913192064 Chr.14: 52439533-52439533 on Build GRCh38
rs763954697 Chr.14: 52439178-52439178 on Build GRCh38
rs760811900 Chr.14: 52439358-52439358 on Build GRCh38
rs903646885 Chr.14: 52439263-52439263 on Build GRCh38
rs927500929 Chr.14: 52439411-52439411 on Build GRCh38
rs927393810 Chr.14: 52439546-52439546 on Build GRCh38
rs1057482854 Chr.14: 52439455-52439455 on Build GRCh38
rs767805499 Chr.14: 52439339-52439339 on Build GRCh38
rs781048334 Chr.14: 52439398-52439398 on Build GRCh38
rs775467421 Chr.14: 52439172-52439172 on Build GRCh38
rs746571909 Chr.14: 52439381-52439381 on Build GRCh38
rs913336402 Chr.14: 52439159-52439159 on Build GRCh38
rs537398672 Chr.14: 52439149-52439149 on Build GRCh38
rs574451141 Chr.14: 52439273-52439273 on Build GRCh38
rs760095210 Chr.14: 52439352-52439352 on Build GRCh38
rs779545975 Chr.14: 52439308-52439308 on Build GRCh38
rs374110782 Chr.14: 52439371-52439371 on Build GRCh38
rs752842679 Chr.14: 52439341-52439341 on Build GRCh38
rs758078938 Chr.14: 52439372-52439372 on Build GRCh38
rs61740050 Chr.14: 52439324-52439324 on Build GRCh38
rs988843137 Chr.14: 52439462-52439462 on Build GRCh38
rs769693236 Chr.14: 52439491-52439491 on Build GRCh38
rs756716657 Chr.14: 52439235-52439235 on Build GRCh38
rs202242219 Chr.14: 52439180-52439180 on Build GRCh38
rs2249922 Chr.14: 52439363-52439363 on Build GRCh38
rs2296720 Chr.14: 52439152-52439152 on Build GRCh38
rs1013296475 Chr.14: 52439116-52439116 on Build GRCh38
rs745524837 Chr.14: 52439282-52439282 on Build GRCh38
rs568846820 Chr.14: 52439134-52439134 on Build GRCh38
rs138718323 Chr.14: 52439335-52439335 on Build GRCh38
rs1000765653 Chr.14: 52439183-52439183 on Build GRCh38
rs1002036100 Chr.14: 52439299-52439299 on Build GRCh38
rs779014538 Chr.14: 52439232-52439232 on Build GRCh38
rs376721513 Chr.14: 52439312-52439312 on Build GRCh38
rs540063493 Chr.14: 52439553-52439553 on Build GRCh38
rs769617436 Chr.14: 52439331-52439331 on Build GRCh38
rs775244593 Chr.14: 52439195-52439195 on Build GRCh38
rs771657526 Chr.14: 52439301-52439301 on Build GRCh38
rs558944957 Chr.14: 52439362-52439362 on Build GRCh38
rs767715971 Chr.14: 52439221-52439221 on Build GRCh38
rs779633860 Chr.14: 52439378-52439378 on Build GRCh38
rs985713847 Chr.14: 52439099-52439099 on Build GRCh38
rs138138518 Chr.14: 52439213-52439213 on Build GRCh38
rs761620835 Chr.14: 52439189-52439189 on Build GRCh38
rs757317866 Chr.14: 52439204-52439204 on Build GRCh38
rs755069094 Chr.14: 52439220-52439220 on Build GRCh38
rs866086281 Chr.14: 52439405-52439405 on Build GRCh38
rs1013281593 Chr.14: 52439329-52439329 on Build GRCh38
rs778372157 Chr.14: 52439245-52439245 on Build GRCh38
rs369684872 Chr.14: 52439316-52439316 on Build GRCh38
rs2249925 Chr.14: 52439461-52439461 on Build GRCh38
rs377005085 Chr.14: 52439406-52439406 on Build GRCh38
rs561060070 Chr.14: 52439508-52439508 on Build GRCh38
rs746484184 Chr.14: 52439310-52439310 on Build GRCh38
rs780335534 Chr.14: 52439417-52439417 on Build GRCh38
rs372638200 Chr.14: 52439559-52439559 on Build GRCh38
rs763751089 Chr.14: 52439398-52439399 on Build GRCh38
rs757614421 Chr.14: 52439270-52439270 on Build GRCh38
rs1032267818 Chr.14: 52439560-52439560 on Build GRCh38
rs760621129 Chr.14: 52439176-52439176 on Build GRCh38
rs768150891 Chr.14: 52439388-52439388 on Build GRCh38
rs942312339 Chr.14: 52439352-52439352 on Build GRCh38
rs912805829 Chr.14: 52439105-52439105 on Build GRCh38
rs1032651674 Chr.14: 52439207-52439207 on Build GRCh38
rs115175934 Chr.14: 52439337-52439337 on Build GRCh38
rs142488571 Chr.14: 52439247-52439247 on Build GRCh38

View Full Product Details