Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 74763623-74764126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YLPM1
Entrez Gene ID
56252
Gene Name
YLP motif containing 1
Gene Aliases
C14orf170, PPP1R169, ZAP113, ZAP3
UniGene
Hs.531111
Gene Chromosome Location
Chr.14: 74763322-74837310 on Build GRCh38
Transcripts
NM_019589.2 XM_005267860.3 XM_011536967.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3793859 c.569C>G p.S190C Chr.14: 74764058-74764058 on Build GRCh38
6344394 c.347G>T p.S116I Chr.14: 74763836-74763836 on Build GRCh38
4614746 c.625_627delTCC p.S214delS Chr.14: 74764114-74764116 on Build GRCh38
4623020 c.138C>A p.S46S Chr.14: 74763627-74763627 on Build GRCh38
6328160 c.556C>T p.P186S Chr.14: 74764045-74764045 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760241306 Chr.14: 74764003-74764003 on Build GRCh38
rs767840541 Chr.14: 74763709-74763709 on Build GRCh38
rs764442899 Chr.14: 74764032-74764032 on Build GRCh38
rs372131920 Chr.14: 74763910-74763910 on Build GRCh38
rs560854168 Chr.14: 74764085-74764085 on Build GRCh38
rs763664957 Chr.14: 74763799-74763799 on Build GRCh38
rs143987825 Chr.14: 74764083-74764083 on Build GRCh38
rs768085986 Chr.14: 74764038-74764038 on Build GRCh38
rs766067966 Chr.14: 74763922-74763922 on Build GRCh38
rs781018053 Chr.14: 74763750-74763750 on Build GRCh38
rs763071273 Chr.14: 74764030-74764030 on Build GRCh38
rs764988374 Chr.14: 74764022-74764022 on Build GRCh38
rs760520326 Chr.14: 74763967-74763967 on Build GRCh38
rs756351026 Chr.14: 74763753-74763753 on Build GRCh38
rs749483295 Chr.14: 74763825-74763825 on Build GRCh38
rs747735348 Chr.14: 74763998-74763998 on Build GRCh38
rs960401134 Chr.14: 74763737-74763737 on Build GRCh38
rs753694448 Chr.14: 74763927-74763927 on Build GRCh38
rs199939020 Chr.14: 74763959-74763959 on Build GRCh38
rs368367178 Chr.14: 74763868-74763868 on Build GRCh38
rs368003492 Chr.14: 74763654-74763654 on Build GRCh38
rs778954948 Chr.14: 74763843-74763843 on Build GRCh38
rs369288881 Chr.14: 74763963-74763963 on Build GRCh38
rs750347405 Chr.14: 74763919-74763919 on Build GRCh38
rs982192582 Chr.14: 74763728-74763728 on Build GRCh38
rs746297298 Chr.14: 74763948-74763948 on Build GRCh38
rs752717104 Chr.14: 74763937-74763937 on Build GRCh38
rs760709881 Chr.14: 74763921-74763921 on Build GRCh38
rs765397089 Chr.14: 74763982-74763982 on Build GRCh38
rs767428055 Chr.14: 74763913-74763913 on Build GRCh38
rs748962521 Chr.14: 74763792-74763793 on Build GRCh38
rs61978907 Chr.14: 74763691-74763691 on Build GRCh38
rs758431783 Chr.14: 74763938-74763938 on Build GRCh38
rs200773284 Chr.14: 74763793-74763793 on Build GRCh38
rs1003156778 Chr.14: 74763720-74763720 on Build GRCh38
rs777175177 Chr.14: 74763877-74763877 on Build GRCh38
rs780555267 Chr.14: 74764060-74764060 on Build GRCh38
rs766374065 Chr.14: 74763970-74763970 on Build GRCh38
rs775621619 Chr.14: 74763651-74763651 on Build GRCh38
rs757422623 Chr.14: 74764035-74764035 on Build GRCh38
rs774864106 Chr.14: 74763675-74763675 on Build GRCh38
rs749305771 Chr.14: 74763756-74763756 on Build GRCh38
rs760952640 Chr.14: 74763782-74763782 on Build GRCh38
rs139231018 Chr.14: 74763951-74763951 on Build GRCh38
rs770797200 Chr.14: 74764007-74764007 on Build GRCh38
rs141202274 Chr.14: 74763798-74763798 on Build GRCh38
rs771228986 Chr.14: 74763874-74763874 on Build GRCh38
rs755193062 Chr.14: 74763928-74763928 on Build GRCh38
rs773756062 Chr.14: 74763945-74763947 on Build GRCh38
rs773058360 Chr.14: 74763767-74763768 on Build GRCh38
rs373775190 Chr.14: 74764034-74764034 on Build GRCh38
rs951562997 Chr.14: 74763900-74763900 on Build GRCh38
rs866012396 Chr.14: 74763725-74763725 on Build GRCh38
rs759341342 Chr.14: 74763977-74763977 on Build GRCh38
rs201818656 Chr.14: 74763940-74763940 on Build GRCh38
rs762127963 Chr.14: 74763707-74763707 on Build GRCh38
rs912465364 Chr.14: 74763831-74763831 on Build GRCh38
rs775028830 Chr.14: 74763795-74763795 on Build GRCh38
rs865863304 Chr.14: 74763660-74763660 on Build GRCh38
rs34816316 Chr.14: 74764061-74764062 on Build GRCh38
rs771515706 Chr.14: 74763941-74763941 on Build GRCh38
rs770407893 Chr.14: 74763922-74763923 on Build GRCh38
rs772831206 Chr.14: 74763966-74763966 on Build GRCh38
rs769566297 Chr.14: 74763747-74763753 on Build GRCh38
rs747352630 Chr.14: 74763871-74763871 on Build GRCh38
rs774184639 Chr.14: 74763741-74763741 on Build GRCh38
rs763896644 Chr.14: 74763888-74763888 on Build GRCh38
rs759213738 Chr.14: 74763724-74763724 on Build GRCh38
rs879247052 Chr.14: 74763759-74763759 on Build GRCh38
rs773623708 Chr.14: 74763781-74763781 on Build GRCh38
rs755872108 Chr.14: 74763821-74763821 on Build GRCh38
rs759288330 Chr.14: 74764017-74764017 on Build GRCh38
rs772517359 Chr.14: 74763775-74763775 on Build GRCh38
rs757699857 Chr.14: 74763987-74763987 on Build GRCh38
rs1001780756 Chr.14: 74763792-74763792 on Build GRCh38
rs781764577 Chr.14: 74763988-74763988 on Build GRCh38
rs749732011 Chr.14: 74764064-74764064 on Build GRCh38
rs868506723 Chr.14: 74763690-74763690 on Build GRCh38
rs1025699077 Chr.14: 74763891-74763891 on Build GRCh38
rs369631818 Chr.14: 74764086-74764086 on Build GRCh38
rs764865724 Chr.14: 74763734-74763734 on Build GRCh38
rs532465116 Chr.14: 74763711-74763711 on Build GRCh38
rs762672098 Chr.14: 74763887-74763887 on Build GRCh38
rs772597134 Chr.14: 74764002-74764002 on Build GRCh38
rs780286180 Chr.14: 74763823-74763823 on Build GRCh38
rs753564375 Chr.14: 74763976-74763976 on Build GRCh38
rs867768250 Chr.14: 74763685-74763685 on Build GRCh38
rs781679324 Chr.14: 74763947-74763947 on Build GRCh38
rs776572971 Chr.14: 74764015-74764015 on Build GRCh38
rs774457725 Chr.14: 74763760-74763760 on Build GRCh38
rs757926327 Chr.14: 74764075-74764075 on Build GRCh38
rs779112462 Chr.14: 74763929-74763929 on Build GRCh38
rs919906035 Chr.14: 74763876-74763876 on Build GRCh38
rs746173855 Chr.14: 74763991-74763991 on Build GRCh38
rs767416412 Chr.14: 74763810-74763810 on Build GRCh38
rs865945775 Chr.14: 74763932-74763932 on Build GRCh38
rs778666327 Chr.14: 74763992-74763992 on Build GRCh38
rs1023323150 Chr.14: 74764018-74764018 on Build GRCh38
rs745704435 Chr.14: 74763751-74763751 on Build GRCh38
rs758686163 Chr.14: 74763985-74763985 on Build GRCh38
rs200074627 Chr.14: 74763852-74763852 on Build GRCh38
rs757300464 Chr.14: 74763748-74763748 on Build GRCh38
rs376981714 Chr.14: 74764077-74764077 on Build GRCh38
rs775402634 Chr.14: 74764028-74764028 on Build GRCh38
rs923819539 Chr.14: 74763777-74763777 on Build GRCh38
rs887317811 Chr.14: 74763965-74763965 on Build GRCh38
rs780141684 Chr.14: 74763754-74763754 on Build GRCh38
rs1036345996 Chr.14: 74763730-74763730 on Build GRCh38
rs750995498 Chr.14: 74764054-74764054 on Build GRCh38
rs757997357 Chr.14: 74763743-74763743 on Build GRCh38
rs751717978 Chr.14: 74763986-74763986 on Build GRCh38
rs748636491 Chr.14: 74763768-74763768 on Build GRCh38
rs531538260 Chr.14: 74763855-74763855 on Build GRCh38
rs771818064 Chr.14: 74764000-74764000 on Build GRCh38
rs370889119 Chr.14: 74763820-74763820 on Build GRCh38
rs757243353 Chr.14: 74763800-74763800 on Build GRCh38
rs756736824 Chr.14: 74764056-74764056 on Build GRCh38
rs867987108 Chr.14: 74763746-74763746 on Build GRCh38
rs755246366 Chr.14: 74763837-74763837 on Build GRCh38
rs371353683 Chr.14: 74763952-74763952 on Build GRCh38

View Full Product Details