Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 81262439-81262953 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STON2
Entrez Gene ID
85439
Gene Name
stonin 2
Gene Aliases
STN2, STNB, STNB2
UniGene
Hs.14248
Gene Chromosome Location
Chr.14: 81260650-81427404 on Build GRCh38
Transcripts
NM_001256430.1 NM_033104.3 XM_006720279.1 XM_011537240.1 XM_011537242.1

SNP Detail

SNP ID Chromosome Location
rs899712315 Chr.14: 81262666-81262666 on Build GRCh38
rs1003118049 Chr.14: 81262822-81262822 on Build GRCh38
rs748762351 Chr.14: 81262682-81262682 on Build GRCh38
rs913663162 Chr.14: 81262603-81262603 on Build GRCh38
rs540476121 Chr.14: 81262588-81262588 on Build GRCh38
rs1006460199 Chr.14: 81262924-81262924 on Build GRCh38
rs934621463 Chr.14: 81262690-81262690 on Build GRCh38
rs771889492 Chr.14: 81262782-81262782 on Build GRCh38
rs904987069 Chr.14: 81262512-81262512 on Build GRCh38
rs896118046 Chr.14: 81262561-81262561 on Build GRCh38
rs188855109 Chr.14: 81262566-81262566 on Build GRCh38
rs555865349 Chr.14: 81262915-81262915 on Build GRCh38
rs937794205 Chr.14: 81262811-81262811 on Build GRCh38
rs540114121 Chr.14: 81262528-81262528 on Build GRCh38
rs1043571754 Chr.14: 81262784-81262784 on Build GRCh38
rs1015665793 Chr.14: 81262736-81262736 on Build GRCh38
rs1047419660 Chr.14: 81262847-81262847 on Build GRCh38
rs372101640 Chr.14: 81262576-81262577 on Build GRCh38
rs796570022 Chr.14: 81262789-81262789 on Build GRCh38
rs1026949943 Chr.14: 81262573-81262573 on Build GRCh38
rs913620891 Chr.14: 81262905-81262905 on Build GRCh38
rs970263242 Chr.14: 81262814-81262814 on Build GRCh38
rs1043804084 Chr.14: 81262635-81262635 on Build GRCh38
rs775909246 Chr.14: 81262783-81262788 on Build GRCh38
rs898588544 Chr.14: 81262753-81262753 on Build GRCh38
rs1004027063 Chr.14: 81262720-81262720 on Build GRCh38
rs770523761 Chr.14: 81262684-81262684 on Build GRCh38
rs745700352 Chr.14: 81262712-81262712 on Build GRCh38
rs956173567 Chr.14: 81262845-81262845 on Build GRCh38
rs563116955 Chr.14: 81262836-81262836 on Build GRCh38
rs946387354 Chr.14: 81262606-81262606 on Build GRCh38
rs1056587863 Chr.14: 81262812-81262812 on Build GRCh38
rs778600445 Chr.14: 81262700-81262700 on Build GRCh38
rs1023390693 Chr.14: 81262760-81262760 on Build GRCh38
rs181310724 Chr.14: 81262617-81262617 on Build GRCh38
rs780600379 Chr.14: 81262470-81262475 on Build GRCh38
rs989411152 Chr.14: 81262897-81262897 on Build GRCh38
rs946630421 Chr.14: 81262758-81262758 on Build GRCh38
rs777442944 Chr.14: 81262651-81262651 on Build GRCh38
rs987904269 Chr.14: 81262569-81262569 on Build GRCh38
rs541943404 Chr.14: 81262783-81262783 on Build GRCh38
rs1023188423 Chr.14: 81262527-81262527 on Build GRCh38
rs896162058 Chr.14: 81262823-81262823 on Build GRCh38
rs928795143 Chr.14: 81262560-81262560 on Build GRCh38
rs868514651 Chr.14: 81262848-81262848 on Build GRCh38
rs993532912 Chr.14: 81262829-81262829 on Build GRCh38
rs984990359 Chr.14: 81262591-81262591 on Build GRCh38
rs536486743 Chr.14: 81262648-81262648 on Build GRCh38
rs952270900 Chr.14: 81262590-81262590 on Build GRCh38
rs888057960 Chr.14: 81262871-81262871 on Build GRCh38
rs184654736 Chr.14: 81262541-81262541 on Build GRCh38
rs530471521 Chr.14: 81262870-81262870 on Build GRCh38
rs964813807 Chr.14: 81262639-81262639 on Build GRCh38
rs138554160 Chr.14: 81262701-81262702 on Build GRCh38
rs17111620 Chr.14: 81262715-81262715 on Build GRCh38
rs995495921 Chr.14: 81262757-81262757 on Build GRCh38

View Full Product Details