Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 88612419-88612966 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZC3H14
Entrez Gene ID
79882
Gene Name
zinc finger CCCH-type containing 14
Gene Aliases
MSUT-2, NY-REN-37, SUT2, UKp68
UniGene
Hs.684046
Gene Chromosome Location
Chr.14: 88562909-88614720 on Build GRCh38
Transcripts
NM_001160103.1 NM_001160104.1 NM_001326295.1 NM_001326296.1 NM_001326297.1 NM_001326298.1 NM_001326299.1 NM_001326300.1 NM_001326301.1 NM_001326302.1 NM_001326303.1 NM_001326304.1 NM_001326305.1 NM_001326306.1 NM_001326307.1 NM_001326308.1 NM_001326309.1 NM_001326310.1 NM_001326311.1 NM_001326312.1 NM_001326313.1 NM_001326314.1 NM_001326315.1 NM_001326316.1 NM_024824.4 NM_207660.3 NM_207661.2 NM_207662.3 NR_136936.1 XM_005268068.4 XM_005268069.4 XM_005268070.4 XM_005268071.4 XM_011537160.2 XM_017021648.1

SNP Detail

SNP ID Chromosome Location
rs1040860636 Chr.14: 88612706-88612706 on Build GRCh38
rs550701278 Chr.14: 88612751-88612751 on Build GRCh38
rs778603557 Chr.14: 88612936-88612937 on Build GRCh38
rs11556106 Chr.14: 88612626-88612626 on Build GRCh38
rs761459934 Chr.14: 88612498-88612498 on Build GRCh38
rs1025158826 Chr.14: 88612787-88612789 on Build GRCh38
rs563454003 Chr.14: 88612890-88612890 on Build GRCh38
rs767350484 Chr.14: 88612511-88612512 on Build GRCh38
rs901817901 Chr.14: 88612716-88612720 on Build GRCh38
rs976735096 Chr.14: 88612899-88612899 on Build GRCh38
rs944028085 Chr.14: 88612471-88612471 on Build GRCh38
rs1055315398 Chr.14: 88612472-88612509 on Build GRCh38
rs552371256 Chr.14: 88612861-88612861 on Build GRCh38
rs976784205 Chr.14: 88612513-88612513 on Build GRCh38
rs921278488 Chr.14: 88612928-88612928 on Build GRCh38
rs908504058 Chr.14: 88612467-88612467 on Build GRCh38
rs921347544 Chr.14: 88612520-88612520 on Build GRCh38
rs149251333 Chr.14: 88612690-88612690 on Build GRCh38
rs932705193 Chr.14: 88612937-88612937 on Build GRCh38
rs1053379010 Chr.14: 88612590-88612590 on Build GRCh38
rs980829768 Chr.14: 88612812-88612813 on Build GRCh38
rs983407601 Chr.14: 88612448-88612448 on Build GRCh38
rs190464296 Chr.14: 88612832-88612832 on Build GRCh38
rs1018866840 Chr.14: 88612788-88612788 on Build GRCh38
rs983740567 Chr.14: 88612781-88612781 on Build GRCh38
rs145276541 Chr.14: 88612752-88612752 on Build GRCh38
rs904708713 Chr.14: 88612582-88612582 on Build GRCh38
rs753438681 Chr.14: 88612938-88612938 on Build GRCh38
rs1043588427 Chr.14: 88612554-88612554 on Build GRCh38
rs773884510 Chr.14: 88612672-88612672 on Build GRCh38
rs990410339 Chr.14: 88612924-88612924 on Build GRCh38
rs533340086 Chr.14: 88612742-88612745 on Build GRCh38
rs969479488 Chr.14: 88612517-88612517 on Build GRCh38
rs961765950 Chr.14: 88612473-88612474 on Build GRCh38
rs376074219 Chr.14: 88612576-88612576 on Build GRCh38
rs200951824 Chr.14: 88612781-88612782 on Build GRCh38
rs776858214 Chr.14: 88612728-88612728 on Build GRCh38
rs545361045 Chr.14: 88612777-88612777 on Build GRCh38
rs1013313971 Chr.14: 88612787-88612787 on Build GRCh38
rs980351883 Chr.14: 88612524-88612524 on Build GRCh38
rs796498483 Chr.14: 88612781-88612782 on Build GRCh38
rs932780197 Chr.14: 88612531-88612531 on Build GRCh38
rs8008847 Chr.14: 88612583-88612583 on Build GRCh38
rs992351350 Chr.14: 88612491-88612491 on Build GRCh38
rs965992012 Chr.14: 88612888-88612888 on Build GRCh38
rs756140251 Chr.14: 88612687-88612688 on Build GRCh38
rs771333779 Chr.14: 88612725-88612725 on Build GRCh38
rs957602429 Chr.14: 88612876-88612877 on Build GRCh38
rs1013872013 Chr.14: 88612686-88612686 on Build GRCh38

View Full Product Details