Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 99512255-99512755 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC85C
Entrez Gene ID
317762
Gene Name
coiled-coil domain containing 85C
Gene Aliases
-
UniGene
Hs.709288
Gene Chromosome Location
Chr.14: 99511266-99604390 on Build GRCh38
Transcripts
NM_001144995.1 XM_011536705.2

SNP Detail

SNP ID Chromosome Location
rs1049131625 Chr.14: 99512379-99512379 on Build GRCh38
rs919829640 Chr.14: 99512670-99512670 on Build GRCh38
rs189880485 Chr.14: 99512431-99512431 on Build GRCh38
rs373138565 Chr.14: 99512314-99512314 on Build GRCh38
rs917739500 Chr.14: 99512456-99512457 on Build GRCh38
rs940779177 Chr.14: 99512590-99512590 on Build GRCh38
rs1040516134 Chr.14: 99512591-99512591 on Build GRCh38
rs956661574 Chr.14: 99512446-99512446 on Build GRCh38
rs969181028 Chr.14: 99512665-99512665 on Build GRCh38
rs923712039 Chr.14: 99512299-99512299 on Build GRCh38
rs575084229 Chr.14: 99512624-99512624 on Build GRCh38
rs917791940 Chr.14: 99512309-99512309 on Build GRCh38
rs779034473 Chr.14: 99512671-99512671 on Build GRCh38
rs947929170 Chr.14: 99512476-99512476 on Build GRCh38
rs768631016 Chr.14: 99512528-99512529 on Build GRCh38
rs1005029921 Chr.14: 99512401-99512401 on Build GRCh38
rs558942238 Chr.14: 99512680-99512680 on Build GRCh38
rs752739920 Chr.14: 99512461-99512463 on Build GRCh38
rs755454721 Chr.14: 99512447-99512450 on Build GRCh38
rs980585303 Chr.14: 99512666-99512666 on Build GRCh38
rs140985730 Chr.14: 99512612-99512612 on Build GRCh38
rs1019126987 Chr.14: 99512407-99512407 on Build GRCh38
rs536211359 Chr.14: 99512289-99512289 on Build GRCh38
rs886534985 Chr.14: 99512399-99512399 on Build GRCh38
rs1030724791 Chr.14: 99512440-99512440 on Build GRCh38
rs979538576 Chr.14: 99512433-99512433 on Build GRCh38
rs376791507 Chr.14: 99512440-99512440 on Build GRCh38
rs370972726 Chr.14: 99512652-99512652 on Build GRCh38
rs984775537 Chr.14: 99512697-99512697 on Build GRCh38
rs1029237615 Chr.14: 99512620-99512620 on Build GRCh38
rs796414757 Chr.14: 99512300-99512300 on Build GRCh38
rs995269018 Chr.14: 99512332-99512332 on Build GRCh38
rs919871674 Chr.14: 99512499-99512499 on Build GRCh38
rs905870054 Chr.14: 99512329-99512329 on Build GRCh38
rs528156315 Chr.14: 99512690-99512690 on Build GRCh38
rs181040875 Chr.14: 99512544-99512544 on Build GRCh38
rs183695757 Chr.14: 99512644-99512644 on Build GRCh38
rs756139706 Chr.14: 99512621-99512621 on Build GRCh38
rs575957110 Chr.14: 99512513-99512513 on Build GRCh38
rs545684223 Chr.14: 99512475-99512476 on Build GRCh38
rs184938586 Chr.14: 99512398-99512398 on Build GRCh38
rs967723154 Chr.14: 99512429-99512432 on Build GRCh38
rs372418154 Chr.14: 99512561-99512561 on Build GRCh38
rs886486839 Chr.14: 99512565-99512565 on Build GRCh38
rs904117570 Chr.14: 99512623-99512623 on Build GRCh38
rs966250375 Chr.14: 99512410-99512410 on Build GRCh38
rs562569012 Chr.14: 99512606-99512606 on Build GRCh38
rs911650299 Chr.14: 99512326-99512339 on Build GRCh38
rs1031091303 Chr.14: 99512626-99512626 on Build GRCh38
rs1029289911 Chr.14: 99512429-99512431 on Build GRCh38
rs3918133 Chr.14: 99512416-99512416 on Build GRCh38
rs980637533 Chr.14: 99512487-99512487 on Build GRCh38
rs1056717546 Chr.14: 99512306-99512306 on Build GRCh38

View Full Product Details