Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 99512642-99513133 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC85C
Entrez Gene ID
317762
Gene Name
coiled-coil domain containing 85C
Gene Aliases
-
UniGene
Hs.709288
Gene Chromosome Location
Chr.14: 99511266-99604390 on Build GRCh38
Transcripts
NM_001144995.1 XM_011536705.2

SNP Detail

SNP ID Chromosome Location
rs566901692 Chr.14: 99512855-99512855 on Build GRCh38
rs919829640 Chr.14: 99512670-99512670 on Build GRCh38
rs901958474 Chr.14: 99512816-99512816 on Build GRCh38
rs758353640 Chr.14: 99513029-99513030 on Build GRCh38
rs865954793 Chr.14: 99512775-99512775 on Build GRCh38
rs1013233308 Chr.14: 99512847-99512847 on Build GRCh38
rs1001515301 Chr.14: 99512837-99512837 on Build GRCh38
rs984775537 Chr.14: 99512697-99512697 on Build GRCh38
rs936876164 Chr.14: 99513105-99513105 on Build GRCh38
rs555170545 Chr.14: 99512954-99512954 on Build GRCh38
rs528156315 Chr.14: 99512690-99512690 on Build GRCh38
rs908482111 Chr.14: 99512922-99512922 on Build GRCh38
rs3918138 Chr.14: 99512862-99512862 on Build GRCh38
rs565821698 Chr.14: 99513006-99513006 on Build GRCh38
rs969181028 Chr.14: 99512665-99512665 on Build GRCh38
rs540957945 Chr.14: 99513014-99513014 on Build GRCh38
rs539864008 Chr.14: 99513062-99513062 on Build GRCh38
rs761925311 Chr.14: 99513015-99513015 on Build GRCh38
rs940741059 Chr.14: 99512780-99512780 on Build GRCh38
rs536569172 Chr.14: 99512846-99512846 on Build GRCh38
rs149721981 Chr.14: 99512867-99512867 on Build GRCh38
rs1040465962 Chr.14: 99512789-99512789 on Build GRCh38
rs779034473 Chr.14: 99512671-99512671 on Build GRCh38
rs188021001 Chr.14: 99512992-99512992 on Build GRCh38
rs148908740 Chr.14: 99512956-99512956 on Build GRCh38
rs538010021 Chr.14: 99512953-99512953 on Build GRCh38
rs551088249 Chr.14: 99512757-99512757 on Build GRCh38
rs546981061 Chr.14: 99512854-99512854 on Build GRCh38
rs3918137 Chr.14: 99512832-99512833 on Build GRCh38
rs1031288607 Chr.14: 99512843-99512843 on Build GRCh38
rs3918139 Chr.14: 99512900-99512900 on Build GRCh38
rs3918135 Chr.14: 99512776-99512776 on Build GRCh38
rs932059675 Chr.14: 99512989-99512989 on Build GRCh38
rs923328955 Chr.14: 99512968-99512968 on Build GRCh38
rs145541190 Chr.14: 99512903-99512903 on Build GRCh38
rs1053102368 Chr.14: 99513109-99513109 on Build GRCh38
rs567401222 Chr.14: 99512821-99512821 on Build GRCh38
rs1027242693 Chr.14: 99512898-99512898 on Build GRCh38
rs3918136 Chr.14: 99512818-99512818 on Build GRCh38
rs962028818 Chr.14: 99512932-99512932 on Build GRCh38
rs772016836 Chr.14: 99512784-99512784 on Build GRCh38
rs373028289 Chr.14: 99512957-99512957 on Build GRCh38
rs558942238 Chr.14: 99512680-99512680 on Build GRCh38
rs980585303 Chr.14: 99512666-99512666 on Build GRCh38
rs550759999 Chr.14: 99512819-99512819 on Build GRCh38
rs577910008 Chr.14: 99512761-99512764 on Build GRCh38

View Full Product Details