Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 99513354-99513867 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC85C
Entrez Gene ID
317762
Gene Name
coiled-coil domain containing 85C
Gene Aliases
-
UniGene
Hs.709288
Gene Chromosome Location
Chr.14: 99511266-99604390 on Build GRCh38
Transcripts
NM_001144995.1 XM_011536705.2

SNP Detail

SNP ID Chromosome Location
rs1048746799 Chr.14: 99513744-99513744 on Build GRCh38
rs1007933313 Chr.14: 99513623-99513623 on Build GRCh38
rs1028214886 Chr.14: 99513608-99513608 on Build GRCh38
rs949595011 Chr.14: 99513514-99513514 on Build GRCh38
rs987287042 Chr.14: 99513617-99513617 on Build GRCh38
rs8015772 Chr.14: 99513519-99513519 on Build GRCh38
rs988680660 Chr.14: 99513500-99513500 on Build GRCh38
rs926907276 Chr.14: 99513517-99513517 on Build GRCh38
rs1032351499 Chr.14: 99513686-99513686 on Build GRCh38
rs957248795 Chr.14: 99513575-99513575 on Build GRCh38
rs957300285 Chr.14: 99513800-99513800 on Build GRCh38
rs1036819724 Chr.14: 99513550-99513550 on Build GRCh38
rs552609772 Chr.14: 99513564-99513564 on Build GRCh38
rs386780676 Chr.14: 99513845-99513849 on Build GRCh38
rs900812545 Chr.14: 99513630-99513630 on Build GRCh38
rs78617458 Chr.14: 99513413-99513413 on Build GRCh38
rs998097640 Chr.14: 99513435-99513435 on Build GRCh38
rs776836083 Chr.14: 99513846-99513848 on Build GRCh38
rs144603938 Chr.14: 99513611-99513611 on Build GRCh38
rs544048801 Chr.14: 99513384-99513384 on Build GRCh38
rs187287918 Chr.14: 99513675-99513675 on Build GRCh38
rs1028265566 Chr.14: 99513436-99513436 on Build GRCh38
rs544557177 Chr.14: 99513832-99513832 on Build GRCh38
rs771323642 Chr.14: 99513748-99513750 on Build GRCh38
rs1048080362 Chr.14: 99513532-99513532 on Build GRCh38
rs181993150 Chr.14: 99513527-99513527 on Build GRCh38
rs35857445 Chr.14: 99513531-99513532 on Build GRCh38
rs776864310 Chr.14: 99513844-99513845 on Build GRCh38
rs769387628 Chr.14: 99513741-99513741 on Build GRCh38
rs4900433 Chr.14: 99513618-99513618 on Build GRCh38
rs191782351 Chr.14: 99513834-99513834 on Build GRCh38
rs1005340557 Chr.14: 99513846-99513846 on Build GRCh38
rs1039845433 Chr.14: 99513713-99513713 on Build GRCh38
rs763861141 Chr.14: 99513606-99513606 on Build GRCh38
rs990171275 Chr.14: 99513593-99513593 on Build GRCh38
rs984161050 Chr.14: 99513764-99513764 on Build GRCh38
rs1004431025 Chr.14: 99513420-99513420 on Build GRCh38
rs551486327 Chr.14: 99513378-99513378 on Build GRCh38
rs368014564 Chr.14: 99513780-99513780 on Build GRCh38
rs1036767208 Chr.14: 99513817-99513817 on Build GRCh38
rs76462303 Chr.14: 99513845-99513847 on Build GRCh38
rs900934450 Chr.14: 99513826-99513826 on Build GRCh38
rs8015735 Chr.14: 99513432-99513432 on Build GRCh38
rs889666906 Chr.14: 99513836-99513836 on Build GRCh38
rs1048607756 Chr.14: 99513394-99513394 on Build GRCh38
rs888399018 Chr.14: 99513845-99513845 on Build GRCh38
rs912608303 Chr.14: 99513619-99513619 on Build GRCh38
rs1046664760 Chr.14: 99513379-99513379 on Build GRCh38
rs538356108 Chr.14: 99513656-99513656 on Build GRCh38
rs550000483 Chr.14: 99513754-99513754 on Build GRCh38
rs920197546 Chr.14: 99513721-99513721 on Build GRCh38
rs568663804 Chr.14: 99513745-99513745 on Build GRCh38
rs868166681 Chr.14: 99513514-99513514 on Build GRCh38
rs577127787 Chr.14: 99513814-99513814 on Build GRCh38
rs371438848 Chr.14: 99513756-99513756 on Build GRCh38
rs986161948 Chr.14: 99513450-99513451 on Build GRCh38
rs958314085 Chr.14: 99513728-99513728 on Build GRCh38
rs914065211 Chr.14: 99513501-99513501 on Build GRCh38
rs931547041 Chr.14: 99513740-99513740 on Build GRCh38
rs752029907 Chr.14: 99513460-99513460 on Build GRCh38
rs1998050 Chr.14: 99513750-99513750 on Build GRCh38
rs566864818 Chr.14: 99513483-99513483 on Build GRCh38
rs1034164605 Chr.14: 99513785-99513785 on Build GRCh38
rs938280513 Chr.14: 99513760-99513760 on Build GRCh38
rs906920828 Chr.14: 99513779-99513779 on Build GRCh38
rs546962778 Chr.14: 99513474-99513474 on Build GRCh38
rs3994976 Chr.14: 99513845-99513848 on Build GRCh38
rs953343580 Chr.14: 99513437-99513437 on Build GRCh38
rs1001849000 Chr.14: 99513385-99513385 on Build GRCh38
rs933788668 Chr.14: 99513827-99513827 on Build GRCh38
rs140831626 Chr.14: 99513518-99513518 on Build GRCh38
rs901038863 Chr.14: 99513433-99513433 on Build GRCh38
rs888155509 Chr.14: 99513537-99513537 on Build GRCh38

View Full Product Details