Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 99514188-99514682 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC85C
Entrez Gene ID
317762
Gene Name
coiled-coil domain containing 85C
Gene Aliases
-
UniGene
Hs.709288
Gene Chromosome Location
Chr.14: 99511266-99604390 on Build GRCh38
Transcripts
NM_001144995.1 XM_011536705.2

SNP Detail

SNP ID Chromosome Location
rs1048361980 Chr.14: 99514610-99514610 on Build GRCh38
rs924461374 Chr.14: 99514575-99514575 on Build GRCh38
rs981309700 Chr.14: 99514581-99514581 on Build GRCh38
rs961931332 Chr.14: 99514218-99514218 on Build GRCh38
rs914342040 Chr.14: 99514533-99514533 on Build GRCh38
rs548707393 Chr.14: 99514234-99514234 on Build GRCh38
rs994418423 Chr.14: 99514486-99514486 on Build GRCh38
rs910992133 Chr.14: 99514321-99514321 on Build GRCh38
rs991123577 Chr.14: 99514508-99514508 on Build GRCh38
rs939381149 Chr.14: 99514384-99514384 on Build GRCh38
rs1055535539 Chr.14: 99514617-99514617 on Build GRCh38
rs982380824 Chr.14: 99514454-99514454 on Build GRCh38
rs952770992 Chr.14: 99514432-99514432 on Build GRCh38
rs1016214406 Chr.14: 99514649-99514649 on Build GRCh38
rs1041550497 Chr.14: 99514345-99514345 on Build GRCh38
rs1008133824 Chr.14: 99514375-99514375 on Build GRCh38
rs955063332 Chr.14: 99514297-99514297 on Build GRCh38
rs570780814 Chr.14: 99514448-99514448 on Build GRCh38
rs926828339 Chr.14: 99514485-99514485 on Build GRCh38
rs957352960 Chr.14: 99514572-99514572 on Build GRCh38
rs766001705 Chr.14: 99514658-99514658 on Build GRCh38
rs1055554706 Chr.14: 99514294-99514294 on Build GRCh38
rs1015747591 Chr.14: 99514442-99514442 on Build GRCh38
rs574589763 Chr.14: 99514625-99514625 on Build GRCh38
rs972357108 Chr.14: 99514259-99514259 on Build GRCh38
rs922255024 Chr.14: 99514269-99514269 on Build GRCh38
rs547586038 Chr.14: 99514381-99514381 on Build GRCh38
rs1045661079 Chr.14: 99514378-99514378 on Build GRCh38
rs754706343 Chr.14: 99514494-99514494 on Build GRCh38
rs116589348 Chr.14: 99514318-99514318 on Build GRCh38
rs200317241 Chr.14: 99514242-99514243 on Build GRCh38
rs899831493 Chr.14: 99514396-99514396 on Build GRCh38
rs985139130 Chr.14: 99514541-99514541 on Build GRCh38
rs888558914 Chr.14: 99514611-99514611 on Build GRCh38
rs576004205 Chr.14: 99514441-99514442 on Build GRCh38
rs1018001365 Chr.14: 99514546-99514546 on Build GRCh38
rs1017526984 Chr.14: 99514450-99514450 on Build GRCh38
rs1015344486 Chr.14: 99514616-99514616 on Build GRCh38
rs903123146 Chr.14: 99514372-99514372 on Build GRCh38
rs575242934 Chr.14: 99514583-99514583 on Build GRCh38
rs879193482 Chr.14: 99514571-99514571 on Build GRCh38
rs190322118 Chr.14: 99514211-99514211 on Build GRCh38
rs371325301 Chr.14: 99514578-99514578 on Build GRCh38
rs112997304 Chr.14: 99514451-99514451 on Build GRCh38
rs77332608 Chr.14: 99514573-99514573 on Build GRCh38
rs990408892 Chr.14: 99514352-99514352 on Build GRCh38
rs937113079 Chr.14: 99514289-99514289 on Build GRCh38
rs982935333 Chr.14: 99514433-99514433 on Build GRCh38
rs913110697 Chr.14: 99514366-99514366 on Build GRCh38
rs943994244 Chr.14: 99514580-99514580 on Build GRCh38
rs947101915 Chr.14: 99514572-99514572 on Build GRCh38
rs527398331 Chr.14: 99514609-99514609 on Build GRCh38
rs537534781 Chr.14: 99514606-99514606 on Build GRCh38
rs996766457 Chr.14: 99514419-99514419 on Build GRCh38
rs947216030 Chr.14: 99514343-99514343 on Build GRCh38
rs895238045 Chr.14: 99514424-99514431 on Build GRCh38
rs534084801 Chr.14: 99514358-99514358 on Build GRCh38
rs1027037301 Chr.14: 99514422-99514422 on Build GRCh38
rs955012802 Chr.14: 99514534-99514534 on Build GRCh38

View Full Product Details