Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 99691695-99692129 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP46A1
Entrez Gene ID
10858
Gene Name
cytochrome P450 family 46 subfamily A member 1
Gene Aliases
CP46, CYP46
UniGene
Hs.25121
Gene Chromosome Location
Chr.14: 99684262-99727321 on Build GRCh38
Transcripts
NM_006668.1 XM_011536364.1 XM_017020933.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1249599 c.258C>T p.I86I Chr.14: 99691837-99691837 on Build GRCh38
949252 c.226C>T p.R76W Chr.14: 99691805-99691805 on Build GRCh38
4960540 c.201G>A p.W67* Chr.14: 99691780-99691780 on Build GRCh38
6639320 c.234C>T p.N78N Chr.14: 99691813-99691813 on Build GRCh38
6253170 c.235G>T p.V79F Chr.14: 99691814-99691814 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs545500131 Chr.14: 99691718-99691718 on Build GRCh38
rs781586799 Chr.14: 99691838-99691838 on Build GRCh38
rs1056712698 Chr.14: 99691807-99691807 on Build GRCh38
rs763781486 Chr.14: 99691831-99691831 on Build GRCh38
rs746361117 Chr.14: 99691845-99691845 on Build GRCh38
rs955586861 Chr.14: 99692084-99692084 on Build GRCh38
rs4900442 Chr.14: 99691904-99691904 on Build GRCh38
rs549791030 Chr.14: 99692085-99692085 on Build GRCh38
rs749591505 Chr.14: 99691891-99691891 on Build GRCh38
rs780458125 Chr.14: 99691856-99691856 on Build GRCh38
rs117491281 Chr.14: 99691973-99691973 on Build GRCh38
rs368649611 Chr.14: 99691761-99691761 on Build GRCh38
rs561755920 Chr.14: 99691966-99691966 on Build GRCh38
rs755640800 Chr.14: 99692039-99692039 on Build GRCh38
rs141032633 Chr.14: 99691798-99691798 on Build GRCh38
rs751135557 Chr.14: 99691837-99691837 on Build GRCh38
rs556161085 Chr.14: 99691801-99691801 on Build GRCh38
rs772474629 Chr.14: 99691760-99691760 on Build GRCh38
rs771713484 Chr.14: 99691717-99691717 on Build GRCh38
rs1047372639 Chr.14: 99691916-99691916 on Build GRCh38
rs748262266 Chr.14: 99691905-99691905 on Build GRCh38
rs995361187 Chr.14: 99691913-99691913 on Build GRCh38
rs749510095 Chr.14: 99691762-99691762 on Build GRCh38
rs761826761 Chr.14: 99691796-99691796 on Build GRCh38
rs781687634 Chr.14: 99691734-99691734 on Build GRCh38
rs760979832 Chr.14: 99691820-99691820 on Build GRCh38
rs757850572 Chr.14: 99691733-99691733 on Build GRCh38
rs956093313 Chr.14: 99692031-99692031 on Build GRCh38
rs746247397 Chr.14: 99691751-99691751 on Build GRCh38
rs759291829 Chr.14: 99691806-99691806 on Build GRCh38
rs762631689 Chr.14: 99691991-99691991 on Build GRCh38
rs748682770 Chr.14: 99691875-99691875 on Build GRCh38
rs796498854 Chr.14: 99691955-99691955 on Build GRCh38
rs533220194 Chr.14: 99692030-99692030 on Build GRCh38
rs754818020 Chr.14: 99691744-99691744 on Build GRCh38
rs996092569 Chr.14: 99692004-99692004 on Build GRCh38
rs563728585 Chr.14: 99692098-99692098 on Build GRCh38
rs113180502 Chr.14: 99691972-99691972 on Build GRCh38
rs917617855 Chr.14: 99691812-99691812 on Build GRCh38
rs752350612 Chr.14: 99691814-99691814 on Build GRCh38
rs979536178 Chr.14: 99692099-99692099 on Build GRCh38
rs774423149 Chr.14: 99691789-99691789 on Build GRCh38
rs765059068 Chr.14: 99691813-99691813 on Build GRCh38
rs758126503 Chr.14: 99691822-99691822 on Build GRCh38
rs772163206 Chr.14: 99691906-99691906 on Build GRCh38
rs564242911 Chr.14: 99692005-99692005 on Build GRCh38
rs756488741 Chr.14: 99691855-99691855 on Build GRCh38
rs1032813141 Chr.14: 99692021-99692021 on Build GRCh38
rs768795276 Chr.14: 99691776-99691776 on Build GRCh38
rs1029643572 Chr.14: 99692036-99692036 on Build GRCh38
rs898877389 Chr.14: 99691919-99691919 on Build GRCh38
rs541995020 Chr.14: 99691952-99691952 on Build GRCh38
rs1010753198 Chr.14: 99692033-99692033 on Build GRCh38
rs1037451871 Chr.14: 99691911-99691911 on Build GRCh38
rs905698656 Chr.14: 99691988-99691988 on Build GRCh38
rs929048868 Chr.14: 99691938-99691938 on Build GRCh38

View Full Product Details