Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 30904433-30904938 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAN1
Entrez Gene ID
22909
Gene Name
FANCD2/FANCI-associated nuclease 1
Gene Aliases
KIAA1018, KMIN, MTMR15, hFAN1
UniGene
Hs.584863
Gene Chromosome Location
Chr.15: 30903695-30943108 on Build GRCh38
Transcripts
NM_001146094.1 NM_001146095.1 NM_001146096.1 NM_014967.4 XM_005254232.4 XM_005254234.4 XM_005254235.3 XM_011521370.2 XM_011521371.1 XM_011521372.2 XM_017022012.1 XM_017022013.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4683330 c.63G>T p.K21N Chr.15: 30904726-30904726 on Build GRCh38
6228652 c.208G>A p.V70I Chr.15: 30904871-30904871 on Build GRCh38
700210 c.3G>A p.M1I Chr.15: 30904666-30904666 on Build GRCh38
1749094 c.15G>A p.G5G Chr.15: 30904678-30904678 on Build GRCh38
2084272 c.95C>T p.S32L Chr.15: 30904758-30904758 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769939083 Chr.15: 30904766-30904766 on Build GRCh38
rs754760573 Chr.15: 30904835-30904835 on Build GRCh38
rs183652083 Chr.15: 30904778-30904778 on Build GRCh38
rs759761370 Chr.15: 30904664-30904666 on Build GRCh38
rs768557388 Chr.15: 30904570-30904570 on Build GRCh38
rs768704240 Chr.15: 30904683-30904683 on Build GRCh38
rs773502379 Chr.15: 30904703-30904703 on Build GRCh38
rs775363349 Chr.15: 30904550-30904550 on Build GRCh38
rs551430119 Chr.15: 30904663-30904663 on Build GRCh38
rs750883961 Chr.15: 30904502-30904502 on Build GRCh38
rs562301610 Chr.15: 30904863-30904863 on Build GRCh38
rs750016168 Chr.15: 30904733-30904733 on Build GRCh38
rs766857188 Chr.15: 30904632-30904632 on Build GRCh38
rs752145511 Chr.15: 30904522-30904522 on Build GRCh38
rs1019800957 Chr.15: 30904617-30904617 on Build GRCh38
rs912196574 Chr.15: 30904567-30904567 on Build GRCh38
rs774760583 Chr.15: 30904650-30904651 on Build GRCh38
rs201860403 Chr.15: 30904881-30904881 on Build GRCh38
rs758119761 Chr.15: 30904672-30904672 on Build GRCh38
rs753859662 Chr.15: 30904895-30904895 on Build GRCh38
rs1032025952 Chr.15: 30904868-30904868 on Build GRCh38
rs747665384 Chr.15: 30904500-30904502 on Build GRCh38
rs752055523 Chr.15: 30904634-30904634 on Build GRCh38
rs963747565 Chr.15: 30904576-30904576 on Build GRCh38
rs143294144 Chr.15: 30904668-30904668 on Build GRCh38
rs1028765504 Chr.15: 30904588-30904588 on Build GRCh38
rs751963671 Chr.15: 30904686-30904686 on Build GRCh38
rs773755093 Chr.15: 30904579-30904581 on Build GRCh38
rs954198296 Chr.15: 30904597-30904597 on Build GRCh38
rs779950676 Chr.15: 30904879-30904879 on Build GRCh38
rs143965941 Chr.15: 30904837-30904837 on Build GRCh38
rs776829777 Chr.15: 30904596-30904596 on Build GRCh38
rs772357075 Chr.15: 30904795-30904795 on Build GRCh38
rs149291568 Chr.15: 30904676-30904676 on Build GRCh38
rs1046469111 Chr.15: 30904665-30904665 on Build GRCh38
rs139353520 Chr.15: 30904743-30904743 on Build GRCh38
rs906464148 Chr.15: 30904492-30904492 on Build GRCh38
rs765535049 Chr.15: 30904604-30904604 on Build GRCh38
rs772421002 Chr.15: 30904866-30904866 on Build GRCh38
rs1007940466 Chr.15: 30904813-30904813 on Build GRCh38
rs148404807 Chr.15: 30904812-30904812 on Build GRCh38
rs951181120 Chr.15: 30904544-30904544 on Build GRCh38
rs771874092 Chr.15: 30904547-30904547 on Build GRCh38
rs139312614 Chr.15: 30904664-30904664 on Build GRCh38
rs756283160 Chr.15: 30904691-30904694 on Build GRCh38
rs753432330 Chr.15: 30904660-30904660 on Build GRCh38
rs965603817 Chr.15: 30904656-30904656 on Build GRCh38
rs916700095 Chr.15: 30904690-30904690 on Build GRCh38
rs758101202 Chr.15: 30904739-30904739 on Build GRCh38
rs766836928 Chr.15: 30904706-30904706 on Build GRCh38
rs778869753 Chr.15: 30904908-30904908 on Build GRCh38
rs376214743 Chr.15: 30904824-30904824 on Build GRCh38
rs936835971 Chr.15: 30904608-30904608 on Build GRCh38
rs774562404 Chr.15: 30904781-30904781 on Build GRCh38
rs371593413 Chr.15: 30904799-30904799 on Build GRCh38
rs751410785 Chr.15: 30904751-30904751 on Build GRCh38
rs916645657 Chr.15: 30904911-30904911 on Build GRCh38
rs753170032 Chr.15: 30904721-30904721 on Build GRCh38
rs757766894 Chr.15: 30904780-30904780 on Build GRCh38
rs777970369 Chr.15: 30904842-30904842 on Build GRCh38
rs773688318 Chr.15: 30904606-30904606 on Build GRCh38
rs746966589 Chr.15: 30904566-30904566 on Build GRCh38
rs755514856 Chr.15: 30904524-30904524 on Build GRCh38
rs748042109 Chr.15: 30904758-30904758 on Build GRCh38
rs763284160 Chr.15: 30904704-30904704 on Build GRCh38
rs756251120 Chr.15: 30904873-30904873 on Build GRCh38
rs748880190 Chr.15: 30904527-30904527 on Build GRCh38
rs745714118 Chr.15: 30904542-30904542 on Build GRCh38
rs986560136 Chr.15: 30904546-30904546 on Build GRCh38
rs749998047 Chr.15: 30904811-30904811 on Build GRCh38
rs766716280 Chr.15: 30904641-30904641 on Build GRCh38
rs367612076 Chr.15: 30904891-30904891 on Build GRCh38
rs750426078 Chr.15: 30904733-30904734 on Build GRCh38
rs764248199 Chr.15: 30904703-30904703 on Build GRCh38
rs200233447 Chr.15: 30904792-30904792 on Build GRCh38
rs770014670 Chr.15: 30904701-30904701 on Build GRCh38
rs533629765 Chr.15: 30904892-30904892 on Build GRCh38
rs763370888 Chr.15: 30904623-30904623 on Build GRCh38
rs762399552 Chr.15: 30904884-30904884 on Build GRCh38
rs146522953 Chr.15: 30904862-30904862 on Build GRCh38
rs369326801 Chr.15: 30904642-30904642 on Build GRCh38
rs369398471 Chr.15: 30904673-30904673 on Build GRCh38
rs958506136 Chr.15: 30904880-30904880 on Build GRCh38
rs764078415 Chr.15: 30904893-30904893 on Build GRCh38
rs767783867 Chr.15: 30904682-30904684 on Build GRCh38
rs756748584 Chr.15: 30904726-30904726 on Build GRCh38
rs745548375 Chr.15: 30904671-30904671 on Build GRCh38
rs376882561 Chr.15: 30904888-30904888 on Build GRCh38
rs1019748777 Chr.15: 30904829-30904830 on Build GRCh38
rs755567701 Chr.15: 30904636-30904636 on Build GRCh38
rs1025373720 Chr.15: 30904526-30904526 on Build GRCh38
rs780753489 Chr.15: 30904509-30904509 on Build GRCh38
rs760735589 Chr.15: 30904582-30904587 on Build GRCh38
rs781134478 Chr.15: 30904756-30904756 on Build GRCh38
rs757342848 Chr.15: 30904904-30904904 on Build GRCh38
rs752820596 Chr.15: 30904690-30904691 on Build GRCh38
rs146249441 Chr.15: 30904770-30904770 on Build GRCh38
rs1001369292 Chr.15: 30904464-30904464 on Build GRCh38
rs766068846 Chr.15: 30904817-30904817 on Build GRCh38
rs764729330 Chr.15: 30904728-30904728 on Build GRCh38
rs759899724 Chr.15: 30904788-30904788 on Build GRCh38
rs763511300 Chr.15: 30904658-30904658 on Build GRCh38
rs199574217 Chr.15: 30904749-30904749 on Build GRCh38
rs746079776 Chr.15: 30904865-30904865 on Build GRCh38
rs776542535 Chr.15: 30904685-30904685 on Build GRCh38
rs771162193 Chr.15: 30904861-30904861 on Build GRCh38
rs761162643 Chr.15: 30904871-30904871 on Build GRCh38
rs144469584 Chr.15: 30904804-30904804 on Build GRCh38
rs759933516 Chr.15: 30904714-30904714 on Build GRCh38
rs749431740 Chr.15: 30904850-30904850 on Build GRCh38
rs756187284 Chr.15: 30904840-30904840 on Build GRCh38
rs765505383 Chr.15: 30904499-30904499 on Build GRCh38
rs775771244 Chr.15: 30904867-30904867 on Build GRCh38
rs898428118 Chr.15: 30904741-30904741 on Build GRCh38
rs962565548 Chr.15: 30904518-30904518 on Build GRCh38
rs774923094 Chr.15: 30904711-30904711 on Build GRCh38
rs113045164 Chr.15: 30904578-30904578 on Build GRCh38
rs1012112818 Chr.15: 30904458-30904458 on Build GRCh38
rs769257347 Chr.15: 30904539-30904540 on Build GRCh38
rs757006087 Chr.15: 30904539-30904539 on Build GRCh38
rs560053916 Chr.15: 30904689-30904689 on Build GRCh38
rs778650689 Chr.15: 30904540-30904540 on Build GRCh38
rs751192352 Chr.15: 30904834-30904834 on Build GRCh38
rs758942143 Chr.15: 30904508-30904508 on Build GRCh38

View Full Product Details