Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 45608607-45609117 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BLOC1S6
Entrez Gene ID
26258
Gene Name
biogenesis of lysosomal organelles complex 1 subunit 6
Gene Aliases
BLOS6, HPS9, PA, PALLID, PLDN
UniGene
Hs.7037
Gene Chromosome Location
Chr.15: 45587123-45609716 on Build GRCh38
Transcripts
NM_001311255.1 NM_001311256.1 NM_012388.3 NR_132350.1 NR_132351.1 NR_132352.1 NR_132353.1 NR_132354.1 NR_132355.1 NR_132356.1 NR_132357.1 NR_132358.1 NR_132359.1

SNP Detail

SNP ID Chromosome Location
rs1056958185 Chr.15: 45608984-45608984 on Build GRCh38
rs995016659 Chr.15: 45608765-45608765 on Build GRCh38
rs1053168339 Chr.15: 45608669-45608669 on Build GRCh38
rs564382292 Chr.15: 45609023-45609023 on Build GRCh38
rs561775048 Chr.15: 45608659-45608659 on Build GRCh38
rs1027198839 Chr.15: 45609046-45609046 on Build GRCh38
rs140243429 Chr.15: 45608809-45608809 on Build GRCh38
rs961181093 Chr.15: 45608641-45608641 on Build GRCh38
rs773617591 Chr.15: 45609054-45609054 on Build GRCh38
rs1026791726 Chr.15: 45608726-45608726 on Build GRCh38
rs7183382 Chr.15: 45608723-45608723 on Build GRCh38
rs532924446 Chr.15: 45608742-45608742 on Build GRCh38
rs924214053 Chr.15: 45608876-45608876 on Build GRCh38
rs935589466 Chr.15: 45608892-45608892 on Build GRCh38
rs549838962 Chr.15: 45608764-45608764 on Build GRCh38
rs1053929992 Chr.15: 45608916-45608916 on Build GRCh38
rs902309741 Chr.15: 45608651-45608651 on Build GRCh38
rs948098050 Chr.15: 45608931-45608931 on Build GRCh38
rs960732113 Chr.15: 45608946-45608946 on Build GRCh38
rs755524457 Chr.15: 45608950-45608950 on Build GRCh38
rs952537891 Chr.15: 45608735-45608735 on Build GRCh38
rs1050899749 Chr.15: 45609074-45609074 on Build GRCh38
rs946404994 Chr.15: 45608983-45608983 on Build GRCh38
rs541084009 Chr.15: 45608715-45608715 on Build GRCh38
rs1027879950 Chr.15: 45608785-45608785 on Build GRCh38
rs1003526598 Chr.15: 45608980-45608980 on Build GRCh38
rs531957344 Chr.15: 45608844-45608844 on Build GRCh38
rs778005816 Chr.15: 45609077-45609077 on Build GRCh38
rs983570317 Chr.15: 45608810-45608810 on Build GRCh38
rs910839992 Chr.15: 45608783-45608783 on Build GRCh38
rs943757567 Chr.15: 45608792-45608792 on Build GRCh38
rs944779796 Chr.15: 45609084-45609084 on Build GRCh38
rs1009007553 Chr.15: 45608747-45608747 on Build GRCh38
rs568517979 Chr.15: 45608988-45608988 on Build GRCh38
rs923868881 Chr.15: 45609008-45609008 on Build GRCh38
rs1018019947 Chr.15: 45608749-45608749 on Build GRCh38
rs879008991 Chr.15: 45608820-45608820 on Build GRCh38
rs13376 Chr.15: 45608918-45608918 on Build GRCh38
rs183813503 Chr.15: 45608949-45608949 on Build GRCh38
rs906521228 Chr.15: 45608964-45608964 on Build GRCh38
rs879518950 Chr.15: 45608630-45608630 on Build GRCh38
rs527490948 Chr.15: 45608703-45608703 on Build GRCh38
rs748461591 Chr.15: 45608973-45608973 on Build GRCh38

View Full Product Details