Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 48341944-48342440 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUT
Entrez Gene ID
1854
Gene Name
deoxyuridine triphosphatase
Gene Aliases
dUTPase
UniGene
Hs.527980
Gene Chromosome Location
Chr.15: 48331018-48343373 on Build GRCh38
Transcripts
NM_001025248.1 NM_001025249.1 NM_001948.3 XM_011521315.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7100678 c.448G>A p.D150N Chr.15: 48342031-48342031 on Build GRCh38
6650419 c.703-2A>G p.? Chr.15: 48342020-48342020 on Build GRCh38
6650420 c.439-2A>G p.? Chr.15: 48342020-48342020 on Build GRCh38
4776377 c.469G>C p.G157R Chr.15: 48342052-48342052 on Build GRCh38
1608446 c.439-9A>G p.? Chr.15: 48342013-48342013 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs932588880 Chr.15: 48342247-48342247 on Build GRCh38
rs189753094 Chr.15: 48342395-48342395 on Build GRCh38
rs568698068 Chr.15: 48342394-48342395 on Build GRCh38
rs754312965 Chr.15: 48341996-48341996 on Build GRCh38
rs976592404 Chr.15: 48342150-48342150 on Build GRCh38
rs751976151 Chr.15: 48342123-48342124 on Build GRCh38
rs780874489 Chr.15: 48342034-48342034 on Build GRCh38
rs966078844 Chr.15: 48342243-48342243 on Build GRCh38
rs766927597 Chr.15: 48342123-48342132 on Build GRCh38
rs760819518 Chr.15: 48342090-48342090 on Build GRCh38
rs913345006 Chr.15: 48342226-48342226 on Build GRCh38
rs760134978 Chr.15: 48342123-48342125 on Build GRCh38
rs764469281 Chr.15: 48342094-48342094 on Build GRCh38
rs941007162 Chr.15: 48342184-48342184 on Build GRCh38
rs1045894173 Chr.15: 48342265-48342265 on Build GRCh38
rs541113636 Chr.15: 48342244-48342244 on Build GRCh38
rs532656179 Chr.15: 48342072-48342072 on Build GRCh38
rs762278544 Chr.15: 48342111-48342111 on Build GRCh38
rs111657285 Chr.15: 48342107-48342107 on Build GRCh38
rs761093797 Chr.15: 48341987-48341987 on Build GRCh38
rs982459956 Chr.15: 48342156-48342156 on Build GRCh38
rs753022633 Chr.15: 48341983-48341983 on Build GRCh38
rs3191517 Chr.15: 48342064-48342064 on Build GRCh38
rs896925886 Chr.15: 48342327-48342327 on Build GRCh38
rs748345998 Chr.15: 48342316-48342316 on Build GRCh38
rs879669862 Chr.15: 48342395-48342395 on Build GRCh38
rs367581813 Chr.15: 48342013-48342013 on Build GRCh38
rs757962290 Chr.15: 48342021-48342021 on Build GRCh38
rs764369536 Chr.15: 48341992-48341992 on Build GRCh38
rs748082058 Chr.15: 48342036-48342036 on Build GRCh38
rs776821799 Chr.15: 48341983-48341983 on Build GRCh38
rs757837796 Chr.15: 48342127-48342127 on Build GRCh38
rs565270265 Chr.15: 48342043-48342043 on Build GRCh38
rs759671140 Chr.15: 48342062-48342062 on Build GRCh38
rs879263317 Chr.15: 48342282-48342282 on Build GRCh38
rs779679093 Chr.15: 48342022-48342022 on Build GRCh38
rs200974717 Chr.15: 48342246-48342247 on Build GRCh38
rs532217216 Chr.15: 48342359-48342359 on Build GRCh38
rs768587963 Chr.15: 48341982-48341983 on Build GRCh38
rs772341797 Chr.15: 48342071-48342071 on Build GRCh38
rs949752097 Chr.15: 48342380-48342381 on Build GRCh38
rs909792980 Chr.15: 48342259-48342259 on Build GRCh38
rs559651438 Chr.15: 48342247-48342247 on Build GRCh38
rs1802838 Chr.15: 48342044-48342044 on Build GRCh38
rs770007620 Chr.15: 48342113-48342113 on Build GRCh38
rs1036680871 Chr.15: 48342294-48342294 on Build GRCh38
rs1007554623 Chr.15: 48342322-48342322 on Build GRCh38
rs768442455 Chr.15: 48342032-48342032 on Build GRCh38
rs750996310 Chr.15: 48342120-48342120 on Build GRCh38
rs1002158260 Chr.15: 48342167-48342167 on Build GRCh38
rs765720628 Chr.15: 48342119-48342119 on Build GRCh38
rs763326450 Chr.15: 48342122-48342123 on Build GRCh38
rs548625600 Chr.15: 48342311-48342311 on Build GRCh38
rs371555609 Chr.15: 48342037-48342037 on Build GRCh38
rs988816828 Chr.15: 48342218-48342218 on Build GRCh38
rs757726281 Chr.15: 48342000-48342000 on Build GRCh38
rs34176493 Chr.15: 48342123-48342123 on Build GRCh38
rs762017160 Chr.15: 48342038-48342038 on Build GRCh38
rs115925970 Chr.15: 48342005-48342005 on Build GRCh38
rs922383188 Chr.15: 48342151-48342151 on Build GRCh38
rs746648206 Chr.15: 48342024-48342024 on Build GRCh38
rs530143957 Chr.15: 48342271-48342271 on Build GRCh38
rs941051665 Chr.15: 48342268-48342271 on Build GRCh38
rs1024453143 Chr.15: 48342128-48342128 on Build GRCh38
rs1033710401 Chr.15: 48342417-48342417 on Build GRCh38
rs763319540 Chr.15: 48342306-48342306 on Build GRCh38
rs957637470 Chr.15: 48342198-48342198 on Build GRCh38
rs929730848 Chr.15: 48342152-48342152 on Build GRCh38
rs950304291 Chr.15: 48342256-48342256 on Build GRCh38
rs909554037 Chr.15: 48342183-48342183 on Build GRCh38

View Full Product Details