Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 49371031-49371529 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM227B
Entrez Gene ID
196951
Gene Name
family with sequence similarity 227 member B
Gene Aliases
C15orf33
UniGene
Hs.712959
Gene Chromosome Location
Chr.15: 49326997-49620931 on Build GRCh38
Transcripts
NM_152647.2 XM_005254213.3 XM_005254214.3 XM_005254215.3 XM_005254216.3 XM_005254219.3 XM_006720423.2 XM_006720424.2 XM_006720426.2 XM_006720427.3 XM_011521319.2 XM_011521320.1 XM_011521321.2 XM_011521322.1 XM_017021990.1 XM_017021991.1 XM_017021992.1 XM_017021993.1 XM_017021994.1 XM_017021995.1 XM_017021996.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6418681 c.1030A>T p.I344L Chr.15: 49371382-49371382 on Build GRCh38
962477 c.1017C>T p.I339I Chr.15: 49371395-49371395 on Build GRCh38
6401353 c.1097G>A p.R366K Chr.15: 49371315-49371315 on Build GRCh38
189949 c.1018G>A p.D340N Chr.15: 49371394-49371394 on Build GRCh38
6425614 c.1039C>A p.L347M Chr.15: 49371373-49371373 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755985230 Chr.15: 49371388-49371388 on Build GRCh38
rs748018624 Chr.15: 49371383-49371383 on Build GRCh38
rs921060123 Chr.15: 49371259-49371259 on Build GRCh38
rs569813053 Chr.15: 49371081-49371083 on Build GRCh38
rs771786867 Chr.15: 49371101-49371101 on Build GRCh38
rs993380954 Chr.15: 49371194-49371194 on Build GRCh38
rs917151734 Chr.15: 49371368-49371368 on Build GRCh38
rs949446992 Chr.15: 49371381-49371381 on Build GRCh38
rs532686804 Chr.15: 49371061-49371061 on Build GRCh38
rs991866761 Chr.15: 49371323-49371323 on Build GRCh38
rs375798990 Chr.15: 49371356-49371356 on Build GRCh38
rs975479239 Chr.15: 49371235-49371235 on Build GRCh38
rs775559315 Chr.15: 49371419-49371419 on Build GRCh38
rs747182684 Chr.15: 49371423-49371423 on Build GRCh38
rs983852324 Chr.15: 49371271-49371271 on Build GRCh38
rs946198123 Chr.15: 49371057-49371057 on Build GRCh38
rs147388904 Chr.15: 49371317-49371317 on Build GRCh38
rs967531341 Chr.15: 49371346-49371346 on Build GRCh38
rs143135128 Chr.15: 49371350-49371350 on Build GRCh38
rs772270991 Chr.15: 49371275-49371275 on Build GRCh38
rs1008904747 Chr.15: 49371129-49371129 on Build GRCh38
rs926260130 Chr.15: 49371082-49371082 on Build GRCh38
rs8030273 Chr.15: 49371212-49371212 on Build GRCh38
rs145082626 Chr.15: 49371395-49371395 on Build GRCh38
rs12903238 Chr.15: 49371208-49371208 on Build GRCh38
rs960366321 Chr.15: 49371311-49371311 on Build GRCh38
rs200397296 Chr.15: 49371337-49371337 on Build GRCh38
rs1040430481 Chr.15: 49371460-49371460 on Build GRCh38
rs768895802 Chr.15: 49371427-49371427 on Build GRCh38
rs746011176 Chr.15: 49371273-49371273 on Build GRCh38
rs772068591 Chr.15: 49371403-49371403 on Build GRCh38
rs774267439 Chr.15: 49371255-49371255 on Build GRCh38
rs1019914312 Chr.15: 49371498-49371498 on Build GRCh38
rs538375897 Chr.15: 49371351-49371351 on Build GRCh38
rs986797053 Chr.15: 49371436-49371436 on Build GRCh38
rs371981118 Chr.15: 49371390-49371390 on Build GRCh38
rs777828804 Chr.15: 49371394-49371394 on Build GRCh38
rs769449330 Chr.15: 49371265-49371265 on Build GRCh38
rs1009727947 Chr.15: 49371461-49371461 on Build GRCh38
rs905273639 Chr.15: 49371097-49371097 on Build GRCh38
rs190202177 Chr.15: 49371169-49371169 on Build GRCh38
rs765648659 Chr.15: 49371444-49371444 on Build GRCh38
rs905159858 Chr.15: 49371389-49371389 on Build GRCh38
rs765813656 Chr.15: 49371316-49371316 on Build GRCh38
rs775753628 Chr.15: 49371284-49371284 on Build GRCh38
rs910515288 Chr.15: 49371437-49371437 on Build GRCh38
rs746925973 Chr.15: 49371105-49371105 on Build GRCh38
rs952569716 Chr.15: 49371265-49371268 on Build GRCh38
rs777073504 Chr.15: 49371300-49371300 on Build GRCh38
rs758038600 Chr.15: 49371319-49371319 on Build GRCh38
rs1045180755 Chr.15: 49371083-49371083 on Build GRCh38
rs1053849441 Chr.15: 49371391-49371391 on Build GRCh38
rs370455293 Chr.15: 49371501-49371501 on Build GRCh38
rs72727271 Chr.15: 49371102-49371102 on Build GRCh38
rs768648254 Chr.15: 49371124-49371124 on Build GRCh38
rs764380459 Chr.15: 49371299-49371299 on Build GRCh38
rs1040438851 Chr.15: 49371163-49371163 on Build GRCh38
rs879236654 Chr.15: 49371491-49371491 on Build GRCh38
rs375286606 Chr.15: 49371435-49371435 on Build GRCh38
rs536659902 Chr.15: 49371186-49371186 on Build GRCh38
rs537970945 Chr.15: 49371487-49371487 on Build GRCh38
rs867905391 Chr.15: 49371059-49371059 on Build GRCh38
rs199971746 Chr.15: 49371309-49371309 on Build GRCh38
rs772941061 Chr.15: 49371413-49371413 on Build GRCh38
rs369064258 Chr.15: 49371386-49371386 on Build GRCh38
rs537748594 Chr.15: 49371218-49371218 on Build GRCh38
rs965073005 Chr.15: 49371220-49371220 on Build GRCh38
rs760888893 Chr.15: 49371292-49371292 on Build GRCh38

View Full Product Details