Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 52046612-52047155 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAPK6
Entrez Gene ID
5597
Gene Name
mitogen-activated protein kinase 6
Gene Aliases
ERK3, HsT17250, PRKM6, p97MAPK
UniGene
Hs.411847
Gene Chromosome Location
Chr.15: 52004265-52066265 on Build GRCh38
Transcripts
NM_002748.3 XM_005254537.2 XM_005254538.2 XM_005254539.3 XM_011521782.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2217209 c.189A>C p.L63L Chr.15: 52046649-52046649 on Build GRCh38
2217208 c.162T>C p.T54T Chr.15: 52046622-52046622 on Build GRCh38
4535326 c.217G>A p.D73N Chr.15: 52046677-52046677 on Build GRCh38
1161125 c.186T>A p.A62A Chr.15: 52046646-52046646 on Build GRCh38
2217211 c.222T>C p.H74H Chr.15: 52046682-52046682 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766419636 Chr.15: 52046740-52046740 on Build GRCh38
rs754545662 Chr.15: 52046826-52046826 on Build GRCh38
rs760824918 Chr.15: 52046742-52046742 on Build GRCh38
rs758693564 Chr.15: 52046774-52046774 on Build GRCh38
rs774091539 Chr.15: 52046741-52046741 on Build GRCh38
rs202143631 Chr.15: 52046905-52046905 on Build GRCh38
rs764364407 Chr.15: 52046874-52046874 on Build GRCh38
rs1003338661 Chr.15: 52046935-52046935 on Build GRCh38
rs773211716 Chr.15: 52046877-52046877 on Build GRCh38
rs779209290 Chr.15: 52046672-52046672 on Build GRCh38
rs776552712 Chr.15: 52046946-52046946 on Build GRCh38
rs763046031 Chr.15: 52046736-52046736 on Build GRCh38
rs371245995 Chr.15: 52046689-52046689 on Build GRCh38
rs966348722 Chr.15: 52047041-52047041 on Build GRCh38
rs775652618 Chr.15: 52046763-52046763 on Build GRCh38
rs752144746 Chr.15: 52047028-52047028 on Build GRCh38
rs376954744 Chr.15: 52047057-52047057 on Build GRCh38
rs780809405 Chr.15: 52047036-52047036 on Build GRCh38
rs760966150 Chr.15: 52046779-52046779 on Build GRCh38
rs762024573 Chr.15: 52047128-52047128 on Build GRCh38
rs144986992 Chr.15: 52046858-52046858 on Build GRCh38
rs769747043 Chr.15: 52046730-52046730 on Build GRCh38
rs748903340 Chr.15: 52047058-52047058 on Build GRCh38
rs747660506 Chr.15: 52047038-52047038 on Build GRCh38
rs773115522 Chr.15: 52046735-52046735 on Build GRCh38
rs374066365 Chr.15: 52047035-52047035 on Build GRCh38
rs765647261 Chr.15: 52046766-52046766 on Build GRCh38
rs763386764 Chr.15: 52046791-52046791 on Build GRCh38
rs267604252 Chr.15: 52046883-52046883 on Build GRCh38
rs1033451433 Chr.15: 52046944-52046944 on Build GRCh38
rs774370270 Chr.15: 52046887-52046887 on Build GRCh38
rs140290030 Chr.15: 52046674-52046674 on Build GRCh38
rs762089050 Chr.15: 52046954-52046954 on Build GRCh38
rs372256056 Chr.15: 52047120-52047120 on Build GRCh38
rs767991240 Chr.15: 52046702-52046702 on Build GRCh38
rs749880752 Chr.15: 52047115-52047115 on Build GRCh38
rs897147276 Chr.15: 52046746-52046746 on Build GRCh38
rs367726654 Chr.15: 52046773-52046773 on Build GRCh38
rs750584368 Chr.15: 52046994-52046994 on Build GRCh38
rs909678077 Chr.15: 52047072-52047072 on Build GRCh38
rs182134495 Chr.15: 52047078-52047078 on Build GRCh38
rs780623512 Chr.15: 52046776-52046776 on Build GRCh38
rs772099610 Chr.15: 52046908-52046909 on Build GRCh38
rs576463529 Chr.15: 52047055-52047055 on Build GRCh38
rs753951278 Chr.15: 52046649-52046649 on Build GRCh38
rs1055616987 Chr.15: 52047092-52047092 on Build GRCh38
rs35058355 Chr.15: 52046851-52046852 on Build GRCh38
rs776366789 Chr.15: 52046720-52046720 on Build GRCh38
rs200525670 Chr.15: 52046932-52046932 on Build GRCh38
rs989230305 Chr.15: 52047023-52047023 on Build GRCh38
rs757641589 Chr.15: 52046668-52046668 on Build GRCh38
rs766609842 Chr.15: 52047018-52047018 on Build GRCh38
rs149935282 Chr.15: 52046778-52046778 on Build GRCh38
rs773987345 Chr.15: 52047063-52047063 on Build GRCh38
rs769560084 Chr.15: 52046868-52046868 on Build GRCh38
rs757440608 Chr.15: 52046765-52046765 on Build GRCh38
rs147851662 Chr.15: 52046739-52046739 on Build GRCh38
rs780533110 Chr.15: 52046845-52046845 on Build GRCh38
rs746532599 Chr.15: 52046696-52046696 on Build GRCh38
rs1038749513 Chr.15: 52046703-52046703 on Build GRCh38
rs765450216 Chr.15: 52046987-52046987 on Build GRCh38
rs761329295 Chr.15: 52046723-52046723 on Build GRCh38
rs554991448 Chr.15: 52046756-52046756 on Build GRCh38
rs759353376 Chr.15: 52046919-52046919 on Build GRCh38
rs749102467 Chr.15: 52046878-52046878 on Build GRCh38
rs55946048 Chr.15: 52046992-52046992 on Build GRCh38
rs754003394 Chr.15: 52047113-52047113 on Build GRCh38
rs147161626 Chr.15: 52047082-52047082 on Build GRCh38
rs374313292 Chr.15: 52046757-52046757 on Build GRCh38
rs779496309 Chr.15: 52046691-52046691 on Build GRCh38
rs758783352 Chr.15: 52047010-52047010 on Build GRCh38

View Full Product Details