Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 62686554-62687068 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TLN2
Entrez Gene ID
83660
Gene Name
talin 2
Gene Aliases
ILWEQ
UniGene
Hs.569438
Gene Chromosome Location
Chr.15: 62390527-62844631 on Build GRCh38
Transcripts
NM_015059.2 XM_005254708.4 XM_005254710.4 XM_005254711.4 XM_005254712.4 XM_005254713.4 XM_005254714.3 XM_005254715.2 XM_006720717.3 XM_017022665.1 XM_017022666.1 XM_017022667.1 XM_017022668.1 XM_017022669.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2254617 c.1096C>T p.P366S Chr.15: 62686779-62686779 on Build GRCh38
5478945 c.1025G>A p.R342H Chr.15: 62686708-62686708 on Build GRCh38
7323172 c.1086G>A p.W362* Chr.15: 62686769-62686769 on Build GRCh38
1664572 c.1057G>A p.E353K Chr.15: 62686740-62686740 on Build GRCh38
1129159 c.987G>T p.V329V Chr.15: 62686670-62686670 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779996365 Chr.15: 62686779-62686779 on Build GRCh38
rs904868501 Chr.15: 62686856-62686856 on Build GRCh38
rs369934703 Chr.15: 62686670-62686670 on Build GRCh38
rs200219832 Chr.15: 62686818-62686818 on Build GRCh38
rs570665348 Chr.15: 62686624-62686624 on Build GRCh38
rs768210728 Chr.15: 62686691-62686691 on Build GRCh38
rs750629080 Chr.15: 62686778-62686778 on Build GRCh38
rs764145223 Chr.15: 62686736-62686736 on Build GRCh38
rs1014802951 Chr.15: 62686617-62686617 on Build GRCh38
rs756386098 Chr.15: 62686758-62686758 on Build GRCh38
rs752032192 Chr.15: 62686619-62686619 on Build GRCh38
rs369483377 Chr.15: 62686830-62686830 on Build GRCh38
rs776342599 Chr.15: 62686696-62686696 on Build GRCh38
rs369853815 Chr.15: 62686911-62686911 on Build GRCh38
rs774303999 Chr.15: 62686853-62686853 on Build GRCh38
rs967552963 Chr.15: 62686968-62686968 on Build GRCh38
rs962054403 Chr.15: 62687018-62687018 on Build GRCh38
rs765261008 Chr.15: 62686831-62686831 on Build GRCh38
rs778666187 Chr.15: 62686813-62686813 on Build GRCh38
rs1014482817 Chr.15: 62686917-62686917 on Build GRCh38
rs767806424 Chr.15: 62686709-62686709 on Build GRCh38
rs751444528 Chr.15: 62686847-62686847 on Build GRCh38
rs746971452 Chr.15: 62686690-62686690 on Build GRCh38
rs778567163 Chr.15: 62686674-62686674 on Build GRCh38
rs534122605 Chr.15: 62686613-62686613 on Build GRCh38
rs1008656270 Chr.15: 62686595-62686595 on Build GRCh38
rs773117332 Chr.15: 62686978-62686978 on Build GRCh38
rs746881731 Chr.15: 62686788-62686788 on Build GRCh38
rs767415744 Chr.15: 62686818-62686819 on Build GRCh38
rs761166609 Chr.15: 62686819-62686819 on Build GRCh38
rs781494562 Chr.15: 62686621-62686621 on Build GRCh38
rs200781214 Chr.15: 62686630-62686630 on Build GRCh38
rs28624818 Chr.15: 62686825-62686825 on Build GRCh38
rs11634784 Chr.15: 62686702-62686702 on Build GRCh38
rs980293939 Chr.15: 62686587-62686587 on Build GRCh38
rs946324236 Chr.15: 62686753-62686753 on Build GRCh38
rs201477913 Chr.15: 62686834-62686834 on Build GRCh38
rs779989050 Chr.15: 62686680-62686680 on Build GRCh38
rs372498240 Chr.15: 62686625-62686625 on Build GRCh38
rs768724291 Chr.15: 62686836-62686836 on Build GRCh38
rs753985153 Chr.15: 62686739-62686739 on Build GRCh38
rs759897878 Chr.15: 62686708-62686708 on Build GRCh38
rs112279831 Chr.15: 62686622-62686622 on Build GRCh38
rs775909178 Chr.15: 62686721-62686721 on Build GRCh38
rs765543453 Chr.15: 62686764-62686764 on Build GRCh38
rs1034768157 Chr.15: 62686894-62686894 on Build GRCh38
rs925971469 Chr.15: 62686800-62686800 on Build GRCh38
rs761185045 Chr.15: 62686731-62686731 on Build GRCh38
rs758243412 Chr.15: 62686677-62686677 on Build GRCh38
rs12917611 Chr.15: 62686817-62686817 on Build GRCh38
rs774560404 Chr.15: 62686701-62686701 on Build GRCh38
rs376693088 Chr.15: 62686810-62686810 on Build GRCh38
rs557491872 Chr.15: 62686635-62686635 on Build GRCh38
rs979443244 Chr.15: 62686972-62686972 on Build GRCh38
rs926198256 Chr.15: 62686981-62686981 on Build GRCh38
rs973405314 Chr.15: 62687021-62687021 on Build GRCh38
rs940702292 Chr.15: 62686632-62686632 on Build GRCh38
rs745713228 Chr.15: 62686675-62686675 on Build GRCh38
rs960483146 Chr.15: 62686910-62686910 on Build GRCh38
rs777834275 Chr.15: 62686633-62686633 on Build GRCh38
rs747746097 Chr.15: 62686812-62686812 on Build GRCh38
rs756599332 Chr.15: 62686859-62686859 on Build GRCh38
rs1001699013 Chr.15: 62686851-62686851 on Build GRCh38
rs527506662 Chr.15: 62687026-62687026 on Build GRCh38
rs1050453013 Chr.15: 62686584-62686584 on Build GRCh38
rs1001834722 Chr.15: 62686870-62686870 on Build GRCh38
rs968854146 Chr.15: 62686581-62686581 on Build GRCh38
rs761558540 Chr.15: 62686596-62686596 on Build GRCh38
rs757388322 Chr.15: 62686752-62686752 on Build GRCh38
rs759224534 Chr.15: 62686607-62686616 on Build GRCh38
rs769213283 Chr.15: 62686814-62686814 on Build GRCh38
rs577034181 Chr.15: 62686939-62686939 on Build GRCh38
rs747801249 Chr.15: 62686700-62686700 on Build GRCh38
rs115874792 Chr.15: 62686839-62686839 on Build GRCh38

View Full Product Details