Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 263896-264395 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM234A
Entrez Gene ID
83986
Gene Name
family with sequence similarity 234 member A
Gene Aliases
C16orf9, ITFG3, gs19
UniGene
Hs.513225
Gene Chromosome Location
Chr.16: 234546-269943 on Build GRCh38
Transcripts
NM_001284497.1 NM_032039.3 NR_104317.1 XM_005255622.1 XM_005255623.2 XM_006720957.1 XM_011522691.1 XM_011522692.1 XM_017023760.1 XM_017023761.1 XM_017023762.1 XM_017023763.1 XM_017023764.1 XM_017023765.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3888460 c.1315C>T p.L439F Chr.16: 264142-264142 on Build GRCh38
5792567 c.1304T>C p.F435S Chr.16: 264131-264131 on Build GRCh38
5820252 c.1311G>A p.W437* Chr.16: 264138-264138 on Build GRCh38
6687734 c.1344G>A p.T448T Chr.16: 264171-264171 on Build GRCh38
5850812 c.1235C>T p.A412V Chr.16: 264062-264062 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1014596201 Chr.16: 264178-264178 on Build GRCh38
rs753075754 Chr.16: 264158-264158 on Build GRCh38
rs1036583090 Chr.16: 264353-264353 on Build GRCh38
rs773791056 Chr.16: 264200-264200 on Build GRCh38
rs770985694 Chr.16: 264146-264146 on Build GRCh38
rs948326784 Chr.16: 264180-264180 on Build GRCh38
rs372545837 Chr.16: 263974-263974 on Build GRCh38
rs890147799 Chr.16: 263961-263961 on Build GRCh38
rs373393536 Chr.16: 264108-264108 on Build GRCh38
rs912188524 Chr.16: 264284-264284 on Build GRCh38
rs149439168 Chr.16: 264144-264144 on Build GRCh38
rs756719253 Chr.16: 264175-264175 on Build GRCh38
rs769542419 Chr.16: 264083-264083 on Build GRCh38
rs758150908 Chr.16: 264109-264109 on Build GRCh38
rs980726234 Chr.16: 264082-264082 on Build GRCh38
rs777606548 Chr.16: 264115-264115 on Build GRCh38
rs748567451 Chr.16: 264352-264352 on Build GRCh38
rs776479223 Chr.16: 264076-264076 on Build GRCh38
rs774493137 Chr.16: 264011-264011 on Build GRCh38
rs372124961 Chr.16: 264019-264019 on Build GRCh38
rs767537349 Chr.16: 264089-264089 on Build GRCh38
rs897509262 Chr.16: 264367-264367 on Build GRCh38
rs766948808 Chr.16: 263960-263960 on Build GRCh38
rs528590536 Chr.16: 264209-264209 on Build GRCh38
rs1034972542 Chr.16: 264224-264224 on Build GRCh38
rs757917953 Chr.16: 264063-264063 on Build GRCh38
rs746493202 Chr.16: 264118-264118 on Build GRCh38
rs62032213 Chr.16: 264176-264176 on Build GRCh38
rs201255499 Chr.16: 264166-264166 on Build GRCh38
rs755350819 Chr.16: 264048-264048 on Build GRCh38
rs577965772 Chr.16: 263973-263982 on Build GRCh38
rs759978569 Chr.16: 264148-264148 on Build GRCh38
rs779501759 Chr.16: 264322-264322 on Build GRCh38
rs747826873 Chr.16: 264057-264057 on Build GRCh38
rs994598336 Chr.16: 264207-264207 on Build GRCh38
rs748843510 Chr.16: 263972-263972 on Build GRCh38
rs376116131 Chr.16: 264120-264120 on Build GRCh38
rs962145079 Chr.16: 264300-264303 on Build GRCh38
rs996442777 Chr.16: 263942-263942 on Build GRCh38
rs1048827192 Chr.16: 264375-264375 on Build GRCh38
rs1054168734 Chr.16: 264196-264196 on Build GRCh38
rs373250620 Chr.16: 264069-264069 on Build GRCh38
rs369204524 Chr.16: 264025-264025 on Build GRCh38
rs532807951 Chr.16: 264302-264302 on Build GRCh38
rs926013987 Chr.16: 264251-264251 on Build GRCh38
rs770958044 Chr.16: 264075-264075 on Build GRCh38
rs192659169 Chr.16: 264090-264090 on Build GRCh38
rs759287594 Chr.16: 264008-264008 on Build GRCh38
rs766691351 Chr.16: 264040-264040 on Build GRCh38
rs913235487 Chr.16: 264239-264239 on Build GRCh38
rs745661765 Chr.16: 264079-264079 on Build GRCh38
rs376809582 Chr.16: 263983-263983 on Build GRCh38
rs978827318 Chr.16: 264246-264246 on Build GRCh38
rs769736523 Chr.16: 264126-264128 on Build GRCh38
rs992826517 Chr.16: 264236-264236 on Build GRCh38
rs371900572 Chr.16: 264004-264004 on Build GRCh38
rs769572816 Chr.16: 264125-264125 on Build GRCh38
rs772252493 Chr.16: 264139-264139 on Build GRCh38
rs114724570 Chr.16: 264210-264210 on Build GRCh38
rs575166109 Chr.16: 264369-264369 on Build GRCh38
rs367863094 Chr.16: 263984-263984 on Build GRCh38
rs370418122 Chr.16: 264104-264104 on Build GRCh38
rs148499142 Chr.16: 264374-264374 on Build GRCh38
rs1044058454 Chr.16: 264012-264012 on Build GRCh38
rs751322019 Chr.16: 264170-264170 on Build GRCh38
rs1008962748 Chr.16: 263964-263964 on Build GRCh38
rs756038282 Chr.16: 264188-264188 on Build GRCh38
rs375509833 Chr.16: 264046-264046 on Build GRCh38
rs754958364 Chr.16: 264235-264235 on Build GRCh38
rs777969465 Chr.16: 264213-264213 on Build GRCh38
rs561772756 Chr.16: 264253-264253 on Build GRCh38
rs754362951 Chr.16: 264094-264094 on Build GRCh38
rs765527749 Chr.16: 264105-264105 on Build GRCh38
rs1045371166 Chr.16: 264185-264185 on Build GRCh38
rs778473386 Chr.16: 263977-263977 on Build GRCh38
rs542200430 Chr.16: 264141-264141 on Build GRCh38
rs770796648 Chr.16: 263998-263998 on Build GRCh38
rs756636372 Chr.16: 264026-264026 on Build GRCh38
rs200473849 Chr.16: 264068-264068 on Build GRCh38
rs904695916 Chr.16: 264132-264132 on Build GRCh38
rs201125551 Chr.16: 264165-264165 on Build GRCh38
rs780411385 Chr.16: 263971-263971 on Build GRCh38
rs778616905 Chr.16: 264049-264049 on Build GRCh38
rs71378508 Chr.16: 264126-264126 on Build GRCh38
rs765594311 Chr.16: 264150-264150 on Build GRCh38
rs537717641 Chr.16: 264189-264189 on Build GRCh38
rs375362136 Chr.16: 264093-264093 on Build GRCh38
rs368799634 Chr.16: 264262-264262 on Build GRCh38
rs930724851 Chr.16: 263939-263939 on Build GRCh38
rs750326172 Chr.16: 264186-264186 on Build GRCh38
rs768717459 Chr.16: 264062-264062 on Build GRCh38
rs75571507 Chr.16: 264256-264256 on Build GRCh38
rs552345956 Chr.16: 264259-264259 on Build GRCh38
rs779736114 Chr.16: 264128-264128 on Build GRCh38
rs373363223 Chr.16: 264240-264240 on Build GRCh38
rs944943415 Chr.16: 264338-264338 on Build GRCh38
rs747675180 Chr.16: 263980-263980 on Build GRCh38
rs144851886 Chr.16: 264267-264267 on Build GRCh38
rs756884001 Chr.16: 264119-264119 on Build GRCh38
rs777133130 Chr.16: 264147-264147 on Build GRCh38
rs774566081 Chr.16: 264088-264088 on Build GRCh38
rs748329887 Chr.16: 264137-264137 on Build GRCh38

View Full Product Details