Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1828804-1829266 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAHD1
Entrez Gene ID
81889
Gene Name
fumarylacetoacetate hydrolase domain containing 1
Gene Aliases
C16orf36, YISKL
UniGene
Hs.513265
Gene Chromosome Location
Chr.16: 1827224-1840207 on Build GRCh38
Transcripts
NM_001018104.2 NM_001142398.1 NM_031208.3

SNP Detail

SNP ID Chromosome Location
rs756917339 Chr.16: 1828847-1828847 on Build GRCh38
rs998117562 Chr.16: 1829058-1829058 on Build GRCh38
rs984235977 Chr.16: 1828928-1828928 on Build GRCh38
rs554461252 Chr.16: 1829089-1829089 on Build GRCh38
rs555264076 Chr.16: 1828995-1828995 on Build GRCh38
rs915744517 Chr.16: 1828864-1828864 on Build GRCh38
rs916979563 Chr.16: 1829120-1829120 on Build GRCh38
rs562935781 Chr.16: 1829209-1829209 on Build GRCh38
rs1000004031 Chr.16: 1828829-1828829 on Build GRCh38
rs959761864 Chr.16: 1829091-1829091 on Build GRCh38
rs1010067797 Chr.16: 1829030-1829030 on Build GRCh38
rs238683 Chr.16: 1829064-1829064 on Build GRCh38
rs909665178 Chr.16: 1828930-1828930 on Build GRCh38
rs927227105 Chr.16: 1829128-1829128 on Build GRCh38
rs1007300553 Chr.16: 1828949-1828949 on Build GRCh38
rs958095021 Chr.16: 1828843-1828843 on Build GRCh38
rs961530651 Chr.16: 1829105-1829105 on Build GRCh38
rs747110789 Chr.16: 1829238-1829238 on Build GRCh38
rs540951453 Chr.16: 1829070-1829070 on Build GRCh38
rs1054280332 Chr.16: 1828980-1828980 on Build GRCh38
rs965771505 Chr.16: 1829032-1829032 on Build GRCh38
rs11643316 Chr.16: 1828976-1828976 on Build GRCh38
rs566571759 Chr.16: 1828946-1828946 on Build GRCh38
rs1010042645 Chr.16: 1829195-1829195 on Build GRCh38
rs182436430 Chr.16: 1828935-1828935 on Build GRCh38
rs373345255 Chr.16: 1829187-1829187 on Build GRCh38
rs150751353 Chr.16: 1829124-1829124 on Build GRCh38
rs754503125 Chr.16: 1829200-1829200 on Build GRCh38
rs959315487 Chr.16: 1828909-1828919 on Build GRCh38
rs1065590 Chr.16: 1828851-1828851 on Build GRCh38
rs777964215 Chr.16: 1829139-1829139 on Build GRCh38
rs1000056645 Chr.16: 1828977-1828977 on Build GRCh38
rs543244007 Chr.16: 1829196-1829196 on Build GRCh38
rs972904206 Chr.16: 1829112-1829112 on Build GRCh38
rs919198483 Chr.16: 1829190-1829190 on Build GRCh38
rs79681924 Chr.16: 1828882-1828882 on Build GRCh38
rs1054334032 Chr.16: 1829184-1829184 on Build GRCh38
rs1032449362 Chr.16: 1828836-1828836 on Build GRCh38
rs915738220 Chr.16: 1829116-1829116 on Build GRCh38
rs547980121 Chr.16: 1828838-1828838 on Build GRCh38
rs1044350283 Chr.16: 1828950-1828950 on Build GRCh38
rs932840253 Chr.16: 1828928-1828928 on Build GRCh38
rs238682 Chr.16: 1829096-1829096 on Build GRCh38
rs1028274251 Chr.16: 1829235-1829235 on Build GRCh38
rs779203757 Chr.16: 1829067-1829067 on Build GRCh38
rs901240424 Chr.16: 1829231-1829231 on Build GRCh38
rs1051460122 Chr.16: 1829232-1829232 on Build GRCh38
rs1021002789 Chr.16: 1829032-1829035 on Build GRCh38
rs190173299 Chr.16: 1828934-1828934 on Build GRCh38
rs1021142071 Chr.16: 1829017-1829017 on Build GRCh38
rs558219433 Chr.16: 1829057-1829057 on Build GRCh38
rs186928631 Chr.16: 1829046-1829046 on Build GRCh38
rs11643306 Chr.16: 1828923-1828923 on Build GRCh38
rs949774221 Chr.16: 1829121-1829122 on Build GRCh38
rs34277837 Chr.16: 1828861-1828861 on Build GRCh38
rs897507802 Chr.16: 1828929-1828929 on Build GRCh38
rs73488060 Chr.16: 1829012-1829012 on Build GRCh38

View Full Product Details