Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1993083-1993587 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYNGR3
Entrez Gene ID
9143
Gene Name
synaptogyrin 3
Gene Aliases
-
UniGene
Hs.435277
Gene Chromosome Location
Chr.16: 1989945-1994275 on Build GRCh38
Transcripts
NM_004209.5

SNP Detail

SNP ID Chromosome Location
rs780582269 Chr.16: 1993444-1993444 on Build GRCh38
rs768787609 Chr.16: 1993120-1993120 on Build GRCh38
rs770462749 Chr.16: 1993426-1993426 on Build GRCh38
rs772933013 Chr.16: 1993389-1993389 on Build GRCh38
rs936830640 Chr.16: 1993303-1993303 on Build GRCh38
rs781517053 Chr.16: 1993442-1993442 on Build GRCh38
rs908978238 Chr.16: 1993227-1993227 on Build GRCh38
rs138868351 Chr.16: 1993221-1993221 on Build GRCh38
rs939862583 Chr.16: 1993413-1993413 on Build GRCh38
rs777378482 Chr.16: 1993308-1993308 on Build GRCh38
rs529849509 Chr.16: 1993365-1993365 on Build GRCh38
rs554897915 Chr.16: 1993441-1993441 on Build GRCh38
rs1037386226 Chr.16: 1993443-1993443 on Build GRCh38
rs756325755 Chr.16: 1993455-1993455 on Build GRCh38
rs761833342 Chr.16: 1993138-1993138 on Build GRCh38
rs749526138 Chr.16: 1993517-1993517 on Build GRCh38
rs892569582 Chr.16: 1993352-1993352 on Build GRCh38
rs1009717312 Chr.16: 1993557-1993557 on Build GRCh38
rs910681945 Chr.16: 1993297-1993297 on Build GRCh38
rs142000283 Chr.16: 1993225-1993225 on Build GRCh38
rs933550277 Chr.16: 1993281-1993281 on Build GRCh38
rs536881320 Chr.16: 1993476-1993476 on Build GRCh38
rs751974515 Chr.16: 1993395-1993395 on Build GRCh38
rs182428676 Chr.16: 1993350-1993350 on Build GRCh38
rs1034237611 Chr.16: 1993379-1993379 on Build GRCh38
rs774097845 Chr.16: 1993121-1993121 on Build GRCh38
rs922957128 Chr.16: 1993268-1993268 on Build GRCh38
rs958228085 Chr.16: 1993259-1993259 on Build GRCh38
rs771723536 Chr.16: 1993142-1993142 on Build GRCh38
rs971326902 Chr.16: 1993161-1993161 on Build GRCh38
rs770053172 Chr.16: 1993103-1993103 on Build GRCh38
rs961992936 Chr.16: 1993231-1993231 on Build GRCh38
rs146005618 Chr.16: 1993372-1993372 on Build GRCh38
rs760379027 Chr.16: 1993178-1993178 on Build GRCh38
rs775536930 Chr.16: 1993110-1993110 on Build GRCh38
rs562240204 Chr.16: 1993200-1993200 on Build GRCh38
rs748003544 Chr.16: 1993558-1993558 on Build GRCh38
rs1016536806 Chr.16: 1993431-1993431 on Build GRCh38
rs989644309 Chr.16: 1993552-1993552 on Build GRCh38
rs905612846 Chr.16: 1993519-1993519 on Build GRCh38
rs374831237 Chr.16: 1993330-1993330 on Build GRCh38
rs914091954 Chr.16: 1993553-1993553 on Build GRCh38
rs921363430 Chr.16: 1993318-1993318 on Build GRCh38
rs917156291 Chr.16: 1993445-1993445 on Build GRCh38
rs906584026 Chr.16: 1993368-1993368 on Build GRCh38
rs971740509 Chr.16: 1993250-1993250 on Build GRCh38
rs773180697 Chr.16: 1993149-1993149 on Build GRCh38
rs746062578 Chr.16: 1993102-1993102 on Build GRCh38
rs1009582754 Chr.16: 1993147-1993147 on Build GRCh38
rs1002780657 Chr.16: 1993170-1993170 on Build GRCh38
rs1050702730 Chr.16: 1993287-1993287 on Build GRCh38
rs556757955 Chr.16: 1993522-1993522 on Build GRCh38
rs1013914480 Chr.16: 1993214-1993214 on Build GRCh38
rs534317433 Chr.16: 1993438-1993438 on Build GRCh38
rs764813194 Chr.16: 1993394-1993394 on Build GRCh38
rs945475163 Chr.16: 1993359-1993359 on Build GRCh38
rs548901746 Chr.16: 1993195-1993195 on Build GRCh38
rs894286094 Chr.16: 1993405-1993405 on Build GRCh38
rs1032885470 Chr.16: 1993254-1993254 on Build GRCh38
rs559699236 Chr.16: 1993174-1993174 on Build GRCh38
rs949713454 Chr.16: 1993515-1993516 on Build GRCh38
rs1045136174 Chr.16: 1993219-1993219 on Build GRCh38
rs573620752 Chr.16: 1993201-1993201 on Build GRCh38
rs568244807 Chr.16: 1993470-1993470 on Build GRCh38
rs892427093 Chr.16: 1993129-1993129 on Build GRCh38
rs749438329 Chr.16: 1993115-1993115 on Build GRCh38
rs779080863 Chr.16: 1993527-1993527 on Build GRCh38
rs972256406 Chr.16: 1993460-1993460 on Build GRCh38
rs772094111 Chr.16: 1993561-1993561 on Build GRCh38
rs1029993859 Chr.16: 1993498-1993498 on Build GRCh38
rs545862921 Chr.16: 1993157-1993157 on Build GRCh38
rs528433045 Chr.16: 1993193-1993193 on Build GRCh38
rs576581523 Chr.16: 1993546-1993546 on Build GRCh38

View Full Product Details