Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3597301-3597811 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLX4
Entrez Gene ID
84464
Gene Name
SLX4 structure-specific endonuclease subunit
Gene Aliases
BTBD12, FANCP, MUS312
UniGene
Hs.143681
Gene Chromosome Location
Chr.16: 3581181-3611598 on Build GRCh38
Transcripts
NM_032444.2 XM_011522715.2 XM_017023775.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
334032 c.1355G>C p.R452T Chr.16: 3597808-3597808 on Build GRCh38
6458442 c.1406delC p.P469fs*4 Chr.16: 3597656-3597656 on Build GRCh38
3387431 c.1518C>T p.S506S Chr.16: 3597544-3597544 on Build GRCh38
5823658 c.1574G>T p.R525L Chr.16: 3597488-3597488 on Build GRCh38
5882712 c.1640C>T p.T547M Chr.16: 3597422-3597422 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs77529764 Chr.16: 3597726-3597726 on Build GRCh38
rs369243098 Chr.16: 3597464-3597464 on Build GRCh38
rs982087433 Chr.16: 3597601-3597601 on Build GRCh38
rs766609103 Chr.16: 3597748-3597748 on Build GRCh38
rs181470615 Chr.16: 3597640-3597640 on Build GRCh38
rs888281987 Chr.16: 3597474-3597474 on Build GRCh38
rs763938852 Chr.16: 3597725-3597725 on Build GRCh38
rs372264472 Chr.16: 3597390-3597390 on Build GRCh38
rs764639716 Chr.16: 3597461-3597461 on Build GRCh38
rs761510940 Chr.16: 3597497-3597499 on Build GRCh38
rs572291726 Chr.16: 3597398-3597398 on Build GRCh38
rs746364844 Chr.16: 3597593-3597593 on Build GRCh38
rs376641868 Chr.16: 3597625-3597625 on Build GRCh38
rs74319927 Chr.16: 3597691-3597691 on Build GRCh38
rs768879506 Chr.16: 3597342-3597342 on Build GRCh38
rs774791210 Chr.16: 3597395-3597395 on Build GRCh38
rs200497436 Chr.16: 3597421-3597421 on Build GRCh38
rs200337039 Chr.16: 3597572-3597572 on Build GRCh38
rs766375138 Chr.16: 3597471-3597471 on Build GRCh38
rs757327408 Chr.16: 3597583-3597583 on Build GRCh38
rs200437227 Chr.16: 3597465-3597465 on Build GRCh38
rs376668879 Chr.16: 3597673-3597673 on Build GRCh38
rs139865448 Chr.16: 3597478-3597478 on Build GRCh38
rs758533390 Chr.16: 3597743-3597743 on Build GRCh38
rs774532876 Chr.16: 3597656-3597656 on Build GRCh38
rs776189589 Chr.16: 3597647-3597647 on Build GRCh38
rs896917989 Chr.16: 3597662-3597662 on Build GRCh38
rs746419738 Chr.16: 3597507-3597507 on Build GRCh38
rs750536193 Chr.16: 3597664-3597664 on Build GRCh38
rs748260629 Chr.16: 3597374-3597374 on Build GRCh38
rs374465569 Chr.16: 3597746-3597746 on Build GRCh38
rs1018886618 Chr.16: 3597666-3597666 on Build GRCh38
rs958404823 Chr.16: 3597412-3597412 on Build GRCh38
rs150547487 Chr.16: 3597425-3597425 on Build GRCh38
rs775295243 Chr.16: 3597747-3597747 on Build GRCh38
rs771618757 Chr.16: 3597713-3597713 on Build GRCh38
rs763869769 Chr.16: 3597399-3597399 on Build GRCh38
rs745404152 Chr.16: 3597703-3597703 on Build GRCh38
rs756369289 Chr.16: 3597488-3597488 on Build GRCh38
rs779857803 Chr.16: 3597434-3597434 on Build GRCh38
rs748957904 Chr.16: 3597633-3597633 on Build GRCh38
rs867235131 Chr.16: 3597661-3597661 on Build GRCh38
rs775465966 Chr.16: 3597714-3597714 on Build GRCh38
rs752089590 Chr.16: 3597756-3597756 on Build GRCh38
rs752899329 Chr.16: 3597479-3597479 on Build GRCh38
rs769495113 Chr.16: 3597525-3597525 on Build GRCh38
rs748407493 Chr.16: 3597424-3597424 on Build GRCh38
rs781416857 Chr.16: 3597423-3597423 on Build GRCh38
rs781465990 Chr.16: 3597680-3597680 on Build GRCh38
rs776626195 Chr.16: 3597359-3597359 on Build GRCh38
rs369137872 Chr.16: 3597411-3597411 on Build GRCh38
rs773558127 Chr.16: 3597389-3597389 on Build GRCh38
rs755156257 Chr.16: 3597422-3597422 on Build GRCh38
rs377440877 Chr.16: 3597563-3597563 on Build GRCh38
rs558236399 Chr.16: 3597605-3597605 on Build GRCh38
rs112133511 Chr.16: 3597677-3597677 on Build GRCh38
rs771292301 Chr.16: 3597498-3597498 on Build GRCh38
rs375996119 Chr.16: 3597400-3597400 on Build GRCh38
rs779660315 Chr.16: 3597416-3597416 on Build GRCh38
rs373381697 Chr.16: 3597614-3597614 on Build GRCh38
rs749696604 Chr.16: 3597433-3597433 on Build GRCh38
rs76350200 Chr.16: 3597786-3597786 on Build GRCh38
rs769908031 Chr.16: 3597388-3597388 on Build GRCh38
rs569591337 Chr.16: 3597551-3597551 on Build GRCh38
rs747522879 Chr.16: 3597674-3597674 on Build GRCh38
rs199683722 Chr.16: 3597515-3597515 on Build GRCh38
rs765859186 Chr.16: 3597545-3597545 on Build GRCh38
rs149126845 Chr.16: 3597690-3597690 on Build GRCh38
rs754055947 Chr.16: 3597577-3597577 on Build GRCh38
rs147090612 Chr.16: 3597735-3597735 on Build GRCh38
rs758419931 Chr.16: 3597413-3597413 on Build GRCh38
rs781549077 Chr.16: 3597783-3597783 on Build GRCh38
rs139766312 Chr.16: 3597592-3597592 on Build GRCh38
rs760594129 Chr.16: 3597720-3597720 on Build GRCh38
rs377148341 Chr.16: 3597435-3597435 on Build GRCh38
rs767473883 Chr.16: 3597651-3597651 on Build GRCh38
rs762550449 Chr.16: 3597542-3597542 on Build GRCh38
rs773861381 Chr.16: 3597643-3597643 on Build GRCh38
rs763833617 Chr.16: 3597657-3597657 on Build GRCh38
rs369994733 Chr.16: 3597683-3597683 on Build GRCh38
rs751308254 Chr.16: 3597477-3597477 on Build GRCh38
rs569518866 Chr.16: 3597667-3597667 on Build GRCh38
rs150053867 Chr.16: 3597607-3597607 on Build GRCh38
rs761314760 Chr.16: 3597524-3597524 on Build GRCh38
rs143831403 Chr.16: 3597489-3597489 on Build GRCh38
rs772612094 Chr.16: 3597447-3597447 on Build GRCh38
rs772560570 Chr.16: 3597532-3597532 on Build GRCh38
rs747219368 Chr.16: 3597335-3597335 on Build GRCh38
rs200793851 Chr.16: 3597768-3597768 on Build GRCh38
rs765058222 Chr.16: 3597606-3597607 on Build GRCh38
rs146360238 Chr.16: 3597499-3597499 on Build GRCh38
rs145194745 Chr.16: 3597620-3597620 on Build GRCh38
rs776394340 Chr.16: 3597459-3597459 on Build GRCh38
rs762061457 Chr.16: 3597729-3597729 on Build GRCh38
rs778213300 Chr.16: 3597541-3597541 on Build GRCh38
rs914008567 Chr.16: 3597568-3597568 on Build GRCh38
rs773395054 Chr.16: 3597432-3597432 on Build GRCh38
rs914165558 Chr.16: 3597780-3597780 on Build GRCh38
rs772562892 Chr.16: 3597512-3597512 on Build GRCh38
rs574659618 Chr.16: 3597654-3597654 on Build GRCh38
rs765351696 Chr.16: 3597658-3597658 on Build GRCh38
rs759525267 Chr.16: 3597560-3597560 on Build GRCh38
rs760494841 Chr.16: 3597656-3597656 on Build GRCh38
rs760000532 Chr.16: 3597442-3597442 on Build GRCh38
rs760336038 Chr.16: 3597397-3597397 on Build GRCh38
rs185962726 Chr.16: 3597739-3597739 on Build GRCh38
rs369280911 Chr.16: 3597566-3597566 on Build GRCh38
rs201754095 Chr.16: 3597460-3597460 on Build GRCh38
rs747483922 Chr.16: 3597562-3597562 on Build GRCh38
rs954947904 Chr.16: 3597509-3597509 on Build GRCh38
rs372128800 Chr.16: 3597668-3597668 on Build GRCh38
rs562774038 Chr.16: 3597402-3597402 on Build GRCh38

View Full Product Details