Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30703914-30704374 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SRCAP
Entrez Gene ID
10847
Gene Name
Snf2-related CREBBP activator protein
Gene Aliases
DOMO1, EAF1, FLHS, SWR1
UniGene
Hs.620916
Gene Chromosome Location
Chr.16: 30699141-30740129 on Build GRCh38
Transcripts
NM_006662.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4393492 c.99C>T p.A33A Chr.16: 30704108-30704108 on Build GRCh38
417047 c.134G>A p.G45D Chr.16: 30704143-30704143 on Build GRCh38
3112801 c.261G>A p.P87P Chr.16: 30704270-30704270 on Build GRCh38
703266 c.63G>A p.S21S Chr.16: 30704072-30704072 on Build GRCh38
3370396 c.257G>A p.G86D Chr.16: 30704266-30704266 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781397603 Chr.16: 30704197-30704197 on Build GRCh38
rs555507140 Chr.16: 30704287-30704287 on Build GRCh38
rs747077712 Chr.16: 30704026-30704026 on Build GRCh38
rs770123385 Chr.16: 30704043-30704045 on Build GRCh38
rs374297760 Chr.16: 30704308-30704308 on Build GRCh38
rs771901170 Chr.16: 30704211-30704211 on Build GRCh38
rs925955060 Chr.16: 30704090-30704090 on Build GRCh38
rs774214549 Chr.16: 30704062-30704062 on Build GRCh38
rs374164117 Chr.16: 30704052-30704052 on Build GRCh38
rs751863851 Chr.16: 30704233-30704233 on Build GRCh38
rs563981058 Chr.16: 30704000-30704000 on Build GRCh38
rs201001345 Chr.16: 30704065-30704065 on Build GRCh38
rs759822774 Chr.16: 30704330-30704330 on Build GRCh38
rs1046251918 Chr.16: 30704156-30704156 on Build GRCh38
rs760443794 Chr.16: 30704164-30704164 on Build GRCh38
rs375952582 Chr.16: 30704270-30704270 on Build GRCh38
rs200145878 Chr.16: 30704259-30704259 on Build GRCh38
rs867691569 Chr.16: 30704119-30704119 on Build GRCh38
rs914848937 Chr.16: 30704066-30704066 on Build GRCh38
rs763828860 Chr.16: 30704172-30704172 on Build GRCh38
rs183497403 Chr.16: 30704071-30704071 on Build GRCh38
rs964786712 Chr.16: 30704209-30704209 on Build GRCh38
rs370329618 Chr.16: 30704291-30704291 on Build GRCh38
rs766290034 Chr.16: 30704055-30704055 on Build GRCh38
rs1046499798 Chr.16: 30704261-30704261 on Build GRCh38
rs914038371 Chr.16: 30704252-30704252 on Build GRCh38
rs780722635 Chr.16: 30704087-30704087 on Build GRCh38
rs759036317 Chr.16: 30704079-30704079 on Build GRCh38
rs892314073 Chr.16: 30703942-30703942 on Build GRCh38
rs774834223 Chr.16: 30704161-30704161 on Build GRCh38
rs890262782 Chr.16: 30704334-30704334 on Build GRCh38
rs759343194 Chr.16: 30704155-30704155 on Build GRCh38
rs149628651 Chr.16: 30704141-30704141 on Build GRCh38
rs1002367639 Chr.16: 30704182-30704182 on Build GRCh38
rs963362269 Chr.16: 30704043-30704043 on Build GRCh38
rs765737673 Chr.16: 30704075-30704075 on Build GRCh38
rs748266967 Chr.16: 30704040-30704040 on Build GRCh38
rs1033313786 Chr.16: 30704190-30704190 on Build GRCh38
rs755700498 Chr.16: 30704072-30704072 on Build GRCh38
rs772860042 Chr.16: 30704325-30704325 on Build GRCh38
rs753165963 Chr.16: 30704336-30704336 on Build GRCh38
rs758841424 Chr.16: 30704194-30704194 on Build GRCh38
rs777780785 Chr.16: 30704272-30704272 on Build GRCh38
rs1054131466 Chr.16: 30704123-30704123 on Build GRCh38
rs776246377 Chr.16: 30704314-30704314 on Build GRCh38
rs757440326 Chr.16: 30704279-30704279 on Build GRCh38
rs545982078 Chr.16: 30704303-30704303 on Build GRCh38
rs766898996 Chr.16: 30704068-30704068 on Build GRCh38
rs138333984 Chr.16: 30704076-30704076 on Build GRCh38
rs201959277 Chr.16: 30704232-30704232 on Build GRCh38
rs754108503 Chr.16: 30704274-30704274 on Build GRCh38
rs192581483 Chr.16: 30703956-30703956 on Build GRCh38
rs771609780 Chr.16: 30704341-30704341 on Build GRCh38
rs769995512 Chr.16: 30704118-30704118 on Build GRCh38
rs545543568 Chr.16: 30703988-30703988 on Build GRCh38
rs150494351 Chr.16: 30703966-30703966 on Build GRCh38
rs779951980 Chr.16: 30704016-30704017 on Build GRCh38
rs769315687 Chr.16: 30704324-30704324 on Build GRCh38
rs767921710 Chr.16: 30704268-30704268 on Build GRCh38
rs561701697 Chr.16: 30704102-30704102 on Build GRCh38
rs772955745 Chr.16: 30704050-30704050 on Build GRCh38
rs565926724 Chr.16: 30704153-30704153 on Build GRCh38
rs769676996 Chr.16: 30704046-30704046 on Build GRCh38
rs539594195 Chr.16: 30704176-30704176 on Build GRCh38
rs760145023 Chr.16: 30704245-30704245 on Build GRCh38
rs779862738 Chr.16: 30704292-30704292 on Build GRCh38
rs748307883 Chr.16: 30704042-30704042 on Build GRCh38
rs753912923 Chr.16: 30704351-30704351 on Build GRCh38
rs570017914 Chr.16: 30704228-30704228 on Build GRCh38
rs748439863 Chr.16: 30704106-30704106 on Build GRCh38
rs993395791 Chr.16: 30704315-30704315 on Build GRCh38
rs1002234310 Chr.16: 30703989-30703989 on Build GRCh38
rs746908083 Chr.16: 30704304-30704304 on Build GRCh38
rs776920211 Chr.16: 30704035-30704035 on Build GRCh38
rs752152893 Chr.16: 30704195-30704195 on Build GRCh38
rs760119570 Chr.16: 30704217-30704217 on Build GRCh38
rs767882138 Chr.16: 30704335-30704335 on Build GRCh38
rs746426119 Chr.16: 30704024-30704024 on Build GRCh38
rs933169755 Chr.16: 30703950-30703973 on Build GRCh38
rs768219091 Chr.16: 30704220-30704220 on Build GRCh38
rs772677313 Chr.16: 30704025-30704025 on Build GRCh38
rs747410723 Chr.16: 30704095-30704095 on Build GRCh38
rs767103262 Chr.16: 30704160-30704160 on Build GRCh38
rs370333025 Chr.16: 30704202-30704202 on Build GRCh38
rs79656879 Chr.16: 30704198-30704198 on Build GRCh38
rs775126389 Chr.16: 30704215-30704215 on Build GRCh38
rs777975632 Chr.16: 30704199-30704199 on Build GRCh38
rs778837387 Chr.16: 30704286-30704286 on Build GRCh38
rs753179022 Chr.16: 30704269-30704269 on Build GRCh38
rs776458047 Chr.16: 30704223-30704223 on Build GRCh38
rs770735806 Chr.16: 30704200-30704200 on Build GRCh38
rs1001871335 Chr.16: 30703997-30703997 on Build GRCh38
rs906292383 Chr.16: 30704180-30704180 on Build GRCh38
rs58723456 Chr.16: 30704319-30704319 on Build GRCh38
rs577807397 Chr.16: 30703971-30703971 on Build GRCh38
rs56209507 Chr.16: 30703955-30703955 on Build GRCh38
rs773047519 Chr.16: 30704129-30704129 on Build GRCh38
rs188276509 Chr.16: 30704142-30704142 on Build GRCh38
rs181109795 Chr.16: 30704344-30704344 on Build GRCh38
rs905306931 Chr.16: 30704267-30704267 on Build GRCh38
rs886051899 Chr.16: 30704293-30704293 on Build GRCh38
rs369999790 Chr.16: 30704133-30704133 on Build GRCh38
rs369716264 Chr.16: 30704019-30704019 on Build GRCh38
rs768563614 Chr.16: 30704031-30704031 on Build GRCh38

View Full Product Details