Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30760549-30761071 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC189
Entrez Gene ID
90835
Gene Name
coiled-coil domain containing 189
Gene Aliases
C16orf93
UniGene
Hs.724010
Gene Chromosome Location
Chr.16: 30757226-30762277 on Build GRCh38
Transcripts
NM_001014979.2 NM_001195620.1 XM_011545976.1 XM_011545978.1 XM_017023852.1 XM_017023853.1 XM_017023854.1 XM_017023855.1 XM_017023856.1 XM_017023857.1 XM_017023858.1 XM_017023859.1 XM_017023861.1 XM_017023862.1 NM_000294.2 NM_001172432.1

Gene Symbol
PHKG2
Entrez Gene ID
5261
Gene Name
phosphorylase kinase catalytic subunit gamma 2
Gene Aliases
GSD9C
UniGene
Hs.65735
Gene Chromosome Location
Chr.16: 30748299-30761176 on Build GRCh38
Transcripts
NM_001014979.2 NM_001195620.1 XM_011545976.1 XM_011545978.1 XM_017023852.1 XM_017023853.1 XM_017023854.1 XM_017023855.1 XM_017023856.1 XM_017023857.1 XM_017023858.1 XM_017023859.1 XM_017023861.1 XM_017023862.1 NM_000294.2 NM_001172432.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6851791 c.281C>A p.T94N Chr.16: 30760624-30760624 on Build GRCh38
6851792 c.170C>A p.T57N Chr.16: 30760624-30760624 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs750024690 Chr.16: 30760703-30760703 on Build GRCh38
rs972970982 Chr.16: 30760673-30760674 on Build GRCh38
rs909661514 Chr.16: 30760839-30760839 on Build GRCh38
rs1053042824 Chr.16: 30760796-30760796 on Build GRCh38
rs767551827 Chr.16: 30760641-30760641 on Build GRCh38
rs959964853 Chr.16: 30760618-30760618 on Build GRCh38
rs771388927 Chr.16: 30760621-30760621 on Build GRCh38
rs546197463 Chr.16: 30760960-30760960 on Build GRCh38
rs953471947 Chr.16: 30760587-30760587 on Build GRCh38
rs796303385 Chr.16: 30760687-30760688 on Build GRCh38
rs1030488205 Chr.16: 30760580-30760580 on Build GRCh38
rs879153174 Chr.16: 30760911-30760911 on Build GRCh38
rs368771114 Chr.16: 30760645-30760645 on Build GRCh38
rs201882666 Chr.16: 30760688-30760689 on Build GRCh38
rs35446957 Chr.16: 30760684-30760685 on Build GRCh38
rs886051921 Chr.16: 30760976-30760976 on Build GRCh38
rs1036696279 Chr.16: 30760662-30760662 on Build GRCh38
rs750112532 Chr.16: 30760944-30760944 on Build GRCh38
rs756999045 Chr.16: 30760675-30760675 on Build GRCh38
rs979864947 Chr.16: 30760759-30760759 on Build GRCh38
rs868054576 Chr.16: 30760809-30760809 on Build GRCh38
rs992680995 Chr.16: 30760787-30760787 on Build GRCh38
rs561244435 Chr.16: 30760674-30760674 on Build GRCh38
rs1008389917 Chr.16: 30761040-30761040 on Build GRCh38
rs766717522 Chr.16: 30760580-30760581 on Build GRCh38
rs963317103 Chr.16: 30760666-30760666 on Build GRCh38
rs969084392 Chr.16: 30760595-30760595 on Build GRCh38
rs569691884 Chr.16: 30760854-30760854 on Build GRCh38
rs1043228593 Chr.16: 30760972-30760972 on Build GRCh38
rs773581097 Chr.16: 30760597-30760597 on Build GRCh38
rs150382749 Chr.16: 30760655-30760655 on Build GRCh38
rs753602877 Chr.16: 30760660-30760660 on Build GRCh38
rs755274906 Chr.16: 30760594-30760595 on Build GRCh38
rs200933996 Chr.16: 30760607-30760607 on Build GRCh38
rs774418752 Chr.16: 30760627-30760627 on Build GRCh38
rs997755706 Chr.16: 30760688-30760688 on Build GRCh38
rs201327122 Chr.16: 30760683-30760683 on Build GRCh38
rs922265843 Chr.16: 30760933-30760933 on Build GRCh38
rs1022094366 Chr.16: 30760737-30760737 on Build GRCh38
rs759799433 Chr.16: 30760635-30760635 on Build GRCh38
rs554975918 Chr.16: 30761045-30761045 on Build GRCh38
rs190683100 Chr.16: 30760808-30760808 on Build GRCh38
rs749885797 Chr.16: 30760730-30760730 on Build GRCh38
rs147660704 Chr.16: 30760969-30760969 on Build GRCh38
rs935888964 Chr.16: 30760763-30760763 on Build GRCh38
rs1021648423 Chr.16: 30760594-30760594 on Build GRCh38
rs370274490 Chr.16: 30760642-30760642 on Build GRCh38
rs905649960 Chr.16: 30760859-30760859 on Build GRCh38
rs898176899 Chr.16: 30760906-30760906 on Build GRCh38
rs545271753 Chr.16: 30760762-30760762 on Build GRCh38
rs1051979942 Chr.16: 30760953-30760953 on Build GRCh38
rs1007651425 Chr.16: 30760732-30760732 on Build GRCh38
rs939765952 Chr.16: 30760866-30760866 on Build GRCh38
rs767189076 Chr.16: 30760883-30760883 on Build GRCh38
rs775190630 Chr.16: 30760573-30760573 on Build GRCh38
rs538862364 Chr.16: 30760804-30760804 on Build GRCh38
rs758821993 Chr.16: 30761000-30761001 on Build GRCh38
rs563676045 Chr.16: 30760589-30760589 on Build GRCh38
rs968825152 Chr.16: 30760738-30760738 on Build GRCh38
rs374671822 Chr.16: 30760652-30760652 on Build GRCh38
rs889296413 Chr.16: 30760959-30760959 on Build GRCh38
rs779673765 Chr.16: 30760712-30760712 on Build GRCh38
rs758167193 Chr.16: 30760709-30760709 on Build GRCh38
rs905495571 Chr.16: 30760629-30760629 on Build GRCh38
rs994337282 Chr.16: 30760908-30760908 on Build GRCh38
rs1031658278 Chr.16: 30760778-30760778 on Build GRCh38
rs959959467 Chr.16: 30760784-30760784 on Build GRCh38
rs149984900 Chr.16: 30760801-30760801 on Build GRCh38
rs1001191906 Chr.16: 30760863-30760863 on Build GRCh38
rs778827493 Chr.16: 30760681-30760681 on Build GRCh38
rs898096322 Chr.16: 30760668-30760668 on Build GRCh38
rs547331495 Chr.16: 30760722-30760722 on Build GRCh38
rs761527381 Chr.16: 30760830-30760830 on Build GRCh38
rs759824996 Chr.16: 30760679-30760686 on Build GRCh38
rs4889505 Chr.16: 30760719-30760719 on Build GRCh38
rs748637602 Chr.16: 30760588-30760588 on Build GRCh38
rs909555780 Chr.16: 30760658-30760658 on Build GRCh38

View Full Product Details