Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 52026860-52027371 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf97
Entrez Gene ID
388276
Gene Name
chromosome 16 open reading frame 97
Gene Aliases
-
UniGene
Hs.534669
Gene Chromosome Location
Chr.16: 52026352-52078517 on Build GRCh38
Transcripts
NM_001242473.1 XM_011523081.1

SNP Detail

SNP ID Chromosome Location
rs912564197 Chr.16: 52027119-52027119 on Build GRCh38
rs533446455 Chr.16: 52027264-52027264 on Build GRCh38
rs913978885 Chr.16: 52027276-52027276 on Build GRCh38
rs1005668810 Chr.16: 52027220-52027220 on Build GRCh38
rs988405769 Chr.16: 52027289-52027289 on Build GRCh38
rs184930335 Chr.16: 52027144-52027144 on Build GRCh38
rs748705719 Chr.16: 52026941-52026941 on Build GRCh38
rs113845775 Chr.16: 52027131-52027131 on Build GRCh38
rs144400974 Chr.16: 52027217-52027217 on Build GRCh38
rs964240941 Chr.16: 52027017-52027017 on Build GRCh38
rs568494137 Chr.16: 52027034-52027034 on Build GRCh38
rs995995803 Chr.16: 52026926-52026926 on Build GRCh38
rs752653011 Chr.16: 52027243-52027243 on Build GRCh38
rs1040235321 Chr.16: 52026912-52026912 on Build GRCh38
rs1039779117 Chr.16: 52027132-52027132 on Build GRCh38
rs111610611 Chr.16: 52027177-52027177 on Build GRCh38
rs757248099 Chr.16: 52027308-52027308 on Build GRCh38
rs1048895889 Chr.16: 52027158-52027158 on Build GRCh38
rs984354822 Chr.16: 52026982-52026982 on Build GRCh38
rs377442950 Chr.16: 52027190-52027190 on Build GRCh38
rs922742221 Chr.16: 52027240-52027240 on Build GRCh38
rs561811185 Chr.16: 52027187-52027187 on Build GRCh38
rs891168145 Chr.16: 52026909-52026909 on Build GRCh38
rs925494405 Chr.16: 52027093-52027093 on Build GRCh38
rs975838972 Chr.16: 52027348-52027348 on Build GRCh38
rs1001778918 Chr.16: 52026932-52026932 on Build GRCh38
rs1055575453 Chr.16: 52027145-52027145 on Build GRCh38
rs12149256 Chr.16: 52027135-52027135 on Build GRCh38
rs765060264 Chr.16: 52027116-52027116 on Build GRCh38
rs925872658 Chr.16: 52027309-52027309 on Build GRCh38
rs1027512162 Chr.16: 52026929-52026929 on Build GRCh38
rs767307719 Chr.16: 52027325-52027325 on Build GRCh38
rs778938897 Chr.16: 52027256-52027256 on Build GRCh38
rs550094189 Chr.16: 52026914-52026914 on Build GRCh38
rs910127388 Chr.16: 52026998-52026998 on Build GRCh38
rs181779361 Chr.16: 52027013-52027013 on Build GRCh38
rs565787553 Chr.16: 52026970-52026970 on Build GRCh38
rs557755917 Chr.16: 52027088-52027088 on Build GRCh38
rs895618635 Chr.16: 52027324-52027324 on Build GRCh38
rs867285541 Chr.16: 52027238-52027238 on Build GRCh38
rs1032513068 Chr.16: 52027258-52027258 on Build GRCh38
rs975560512 Chr.16: 52027239-52027239 on Build GRCh38
rs1055649213 Chr.16: 52027319-52027319 on Build GRCh38
rs979560373 Chr.16: 52027257-52027257 on Build GRCh38
rs969525983 Chr.16: 52027054-52027054 on Build GRCh38
rs949890158 Chr.16: 52027326-52027326 on Build GRCh38
rs1025238314 Chr.16: 52026964-52026964 on Build GRCh38
rs1017178567 Chr.16: 52027225-52027225 on Build GRCh38
rs535539479 Chr.16: 52026957-52026957 on Build GRCh38
rs944135800 Chr.16: 52026910-52026910 on Build GRCh38
rs573105674 Chr.16: 52027218-52027218 on Build GRCh38
rs550499398 Chr.16: 52027327-52027327 on Build GRCh38
rs541042310 Chr.16: 52027246-52027246 on Build GRCh38
rs922669290 Chr.16: 52027053-52027053 on Build GRCh38
rs181083823 Chr.16: 52027262-52027262 on Build GRCh38
rs975614151 Chr.16: 52027044-52027044 on Build GRCh38
rs555453351 Chr.16: 52026969-52026969 on Build GRCh38
rs759070903 Chr.16: 52026977-52026977 on Build GRCh38
rs764820222 Chr.16: 52027075-52027075 on Build GRCh38
rs991914566 Chr.16: 52026986-52026986 on Build GRCh38
rs538785637 Chr.16: 52027241-52027241 on Build GRCh38
rs969137920 Chr.16: 52027251-52027251 on Build GRCh38
rs965390853 Chr.16: 52027318-52027318 on Build GRCh38
rs935574922 Chr.16: 52027115-52027115 on Build GRCh38
rs959809076 Chr.16: 52026948-52026948 on Build GRCh38
rs960691844 Chr.16: 52026984-52026984 on Build GRCh38
rs534589979 Chr.16: 52027039-52027039 on Build GRCh38
rs899940419 Chr.16: 52027134-52027134 on Build GRCh38
rs1023832762 Chr.16: 52027003-52027003 on Build GRCh38
rs964676257 Chr.16: 52027235-52027235 on Build GRCh38
rs763963345 Chr.16: 52027171-52027171 on Build GRCh38
rs117092817 Chr.16: 52027186-52027186 on Build GRCh38
rs1014600202 Chr.16: 52026980-52026980 on Build GRCh38
rs570300018 Chr.16: 52026915-52026915 on Build GRCh38
rs912837392 Chr.16: 52027304-52027304 on Build GRCh38
rs1052540687 Chr.16: 52027104-52027104 on Build GRCh38
rs550299137 Chr.16: 52026971-52026971 on Build GRCh38
rs34419501 Chr.16: 52027330-52027331 on Build GRCh38

View Full Product Details