Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 57025665-57026153 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NLRC5
Entrez Gene ID
84166
Gene Name
NLR family CARD domain containing 5
Gene Aliases
CLR16.1, NOD27, NOD4
UniGene
Hs.528836
Gene Chromosome Location
Chr.16: 56989547-57083524 on Build GRCh38
Transcripts
NM_032206.4 XM_005256193.2 XM_005256194.3 XM_005256195.2 XM_005256196.2 XM_005256197.2 XM_005256199.3 XM_005256201.3 XM_006721297.2 XM_006721298.2 XM_006721300.2 XM_011523373.2 XM_011523375.2 XM_017023766.1 XM_017023767.1 XM_017023768.1 XM_017023769.1 XM_017023770.1 XM_017023771.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5882734 c.948G>C p.M316I Chr.16: 57025891-57025891 on Build GRCh38
6456937 c.1089C>A p.H363Q Chr.16: 57026032-57026032 on Build GRCh38
971765 c.1062C>T p.C354C Chr.16: 57026005-57026005 on Build GRCh38
4460460 c.1167C>T p.A389A Chr.16: 57026110-57026110 on Build GRCh38
6225894 c.784C>T p.R262C Chr.16: 57025727-57025727 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200816547 Chr.16: 57025728-57025728 on Build GRCh38
rs765031527 Chr.16: 57025751-57025751 on Build GRCh38
rs184144107 Chr.16: 57025837-57025837 on Build GRCh38
rs762040833 Chr.16: 57025862-57025862 on Build GRCh38
rs199475973 Chr.16: 57025799-57025799 on Build GRCh38
rs758068265 Chr.16: 57025948-57025948 on Build GRCh38
rs528175085 Chr.16: 57025807-57025807 on Build GRCh38
rs768908242 Chr.16: 57026049-57026049 on Build GRCh38
rs757860782 Chr.16: 57025691-57025691 on Build GRCh38
rs769667893 Chr.16: 57025986-57025986 on Build GRCh38
rs756691502 Chr.16: 57025966-57025966 on Build GRCh38
rs746592436 Chr.16: 57025811-57025811 on Build GRCh38
rs778879120 Chr.16: 57026119-57026119 on Build GRCh38
rs149835370 Chr.16: 57025747-57025747 on Build GRCh38
rs745812080 Chr.16: 57025832-57025832 on Build GRCh38
rs780713848 Chr.16: 57025974-57025974 on Build GRCh38
rs780658848 Chr.16: 57026099-57026099 on Build GRCh38
rs749975403 Chr.16: 57026100-57026100 on Build GRCh38
rs201944671 Chr.16: 57025765-57025765 on Build GRCh38
rs148962065 Chr.16: 57025810-57025810 on Build GRCh38
rs140127152 Chr.16: 57025699-57025699 on Build GRCh38
rs151040563 Chr.16: 57025978-57025978 on Build GRCh38
rs143641044 Chr.16: 57025965-57025965 on Build GRCh38
rs141391037 Chr.16: 57026086-57026086 on Build GRCh38
rs768417275 Chr.16: 57025726-57025726 on Build GRCh38
rs775715527 Chr.16: 57025760-57025760 on Build GRCh38
rs370758328 Chr.16: 57025777-57025777 on Build GRCh38
rs776496498 Chr.16: 57025831-57025831 on Build GRCh38
rs758040701 Chr.16: 57025801-57025801 on Build GRCh38
rs772623989 Chr.16: 57025741-57025741 on Build GRCh38
rs1043168817 Chr.16: 57026034-57026034 on Build GRCh38
rs752066689 Chr.16: 57025685-57025685 on Build GRCh38
rs780283637 Chr.16: 57025797-57025797 on Build GRCh38
rs769539344 Chr.16: 57025833-57025833 on Build GRCh38
rs777351415 Chr.16: 57025952-57025952 on Build GRCh38
rs757807193 Chr.16: 57025768-57025768 on Build GRCh38
rs370396132 Chr.16: 57026098-57026098 on Build GRCh38
rs756485667 Chr.16: 57025791-57025791 on Build GRCh38
rs759550372 Chr.16: 57026073-57026073 on Build GRCh38
rs756074401 Chr.16: 57026102-57026102 on Build GRCh38
rs770844231 Chr.16: 57025821-57025821 on Build GRCh38
rs368949786 Chr.16: 57026104-57026104 on Build GRCh38
rs766712762 Chr.16: 57025899-57025899 on Build GRCh38
rs929168133 Chr.16: 57025808-57025808 on Build GRCh38
rs373616538 Chr.16: 57026054-57026054 on Build GRCh38
rs939141396 Chr.16: 57025873-57025873 on Build GRCh38
rs371370138 Chr.16: 57025861-57025861 on Build GRCh38
rs746274418 Chr.16: 57025702-57025702 on Build GRCh38
rs865847577 Chr.16: 57025772-57025772 on Build GRCh38
rs759386496 Chr.16: 57025749-57025749 on Build GRCh38
rs763116144 Chr.16: 57025761-57025761 on Build GRCh38
rs749102247 Chr.16: 57026021-57026021 on Build GRCh38
rs750802020 Chr.16: 57025785-57025785 on Build GRCh38
rs760643795 Chr.16: 57025743-57025743 on Build GRCh38
rs528088895 Chr.16: 57026092-57026092 on Build GRCh38
rs145752799 Chr.16: 57025762-57025762 on Build GRCh38
rs200058658 Chr.16: 57026087-57026087 on Build GRCh38
rs775323140 Chr.16: 57026000-57026000 on Build GRCh38
rs761237528 Chr.16: 57026075-57026075 on Build GRCh38
rs201169198 Chr.16: 57025736-57025736 on Build GRCh38
rs575248573 Chr.16: 57026035-57026035 on Build GRCh38
rs763212603 Chr.16: 57025855-57025855 on Build GRCh38
rs773443195 Chr.16: 57026072-57026072 on Build GRCh38
rs756716483 Chr.16: 57025881-57025881 on Build GRCh38
rs199937881 Chr.16: 57025746-57025746 on Build GRCh38
rs987948170 Chr.16: 57025745-57025745 on Build GRCh38
rs754199282 Chr.16: 57025921-57025921 on Build GRCh38
rs776190500 Chr.16: 57025993-57025994 on Build GRCh38
rs368643974 Chr.16: 57026059-57026059 on Build GRCh38
rs199475972 Chr.16: 57025790-57025790 on Build GRCh38
rs562351862 Chr.16: 57026123-57026123 on Build GRCh38
rs376197946 Chr.16: 57026113-57026113 on Build GRCh38
rs147147126 Chr.16: 57025800-57025800 on Build GRCh38
rs769045410 Chr.16: 57025994-57025997 on Build GRCh38
rs755931672 Chr.16: 57026134-57026134 on Build GRCh38
rs764261540 Chr.16: 57025860-57025860 on Build GRCh38
rs762127136 Chr.16: 57026055-57026055 on Build GRCh38
rs747765853 Chr.16: 57025735-57025735 on Build GRCh38
rs199565452 Chr.16: 57026097-57026097 on Build GRCh38
rs767715827 Chr.16: 57025869-57025869 on Build GRCh38
rs750890230 Chr.16: 57025877-57025877 on Build GRCh38

View Full Product Details