Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 58197214-58197706 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC113
Entrez Gene ID
29070
Gene Name
coiled-coil domain containing 113
Gene Aliases
HSPC065
UniGene
Hs.11614
Gene Chromosome Location
Chr.16: 58196863-58283836 on Build GRCh38
Transcripts
XM_011523039.2 NM_001896.2 XM_005255800.3 XM_005255801.3

Gene Symbol
CSNK2A2
Entrez Gene ID
1459
Gene Name
casein kinase 2 alpha 2
Gene Aliases
CK2A2, CK2alpha', CSNK2A1
UniGene
Hs.82201
Gene Chromosome Location
Chr.16: 58157907-58198116 on Build GRCh38
Transcripts
XM_011523039.2 NM_001896.2 XM_005255800.3 XM_005255801.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6522894 c.65G>T p.R22L Chr.16: 58197672-58197672 on Build GRCh38
6522893 c.88C>T p.H30Y Chr.16: 58197649-58197649 on Build GRCh38
1189284 c.75G>T p.W25C Chr.16: 58197662-58197662 on Build GRCh38
5479151 c.85G>T p.A29S Chr.16: 58197652-58197652 on Build GRCh38
6322116 c.31C>A p.R11R Chr.16: 58197706-58197706 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs928958238 Chr.16: 58197552-58197552 on Build GRCh38
rs1006084794 Chr.16: 58197248-58197248 on Build GRCh38
rs969665804 Chr.16: 58197575-58197575 on Build GRCh38
rs959179462 Chr.16: 58197278-58197278 on Build GRCh38
rs867795522 Chr.16: 58197403-58197403 on Build GRCh38
rs535777438 Chr.16: 58197469-58197469 on Build GRCh38
rs1018544700 Chr.16: 58197532-58197532 on Build GRCh38
rs111959151 Chr.16: 58197557-58197557 on Build GRCh38
rs912871180 Chr.16: 58197680-58197680 on Build GRCh38
rs576968089 Chr.16: 58197683-58197683 on Build GRCh38
rs988986368 Chr.16: 58197608-58197608 on Build GRCh38
rs773955015 Chr.16: 58197531-58197531 on Build GRCh38
rs570571472 Chr.16: 58197365-58197365 on Build GRCh38
rs941975179 Chr.16: 58197445-58197445 on Build GRCh38
rs906650236 Chr.16: 58197486-58197486 on Build GRCh38
rs1049142753 Chr.16: 58197326-58197326 on Build GRCh38
rs988480925 Chr.16: 58197465-58197468 on Build GRCh38
rs78279459 Chr.16: 58197439-58197439 on Build GRCh38
rs891702031 Chr.16: 58197255-58197255 on Build GRCh38
rs1016078003 Chr.16: 58197263-58197263 on Build GRCh38
rs556857717 Chr.16: 58197560-58197560 on Build GRCh38
rs999609162 Chr.16: 58197503-58197503 on Build GRCh38
rs942858668 Chr.16: 58197372-58197372 on Build GRCh38
rs983201321 Chr.16: 58197574-58197574 on Build GRCh38
rs534552306 Chr.16: 58197389-58197389 on Build GRCh38
rs564988972 Chr.16: 58197665-58197665 on Build GRCh38
rs989451331 Chr.16: 58197528-58197528 on Build GRCh38
rs1014271355 Chr.16: 58197390-58197390 on Build GRCh38
rs747933430 Chr.16: 58197413-58197414 on Build GRCh38
rs745740318 Chr.16: 58197661-58197661 on Build GRCh38
rs746514351 Chr.16: 58197625-58197625 on Build GRCh38
rs1047882594 Chr.16: 58197438-58197438 on Build GRCh38
rs375935688 Chr.16: 58197408-58197408 on Build GRCh38
rs1012059889 Chr.16: 58197291-58197291 on Build GRCh38
rs751637921 Chr.16: 58197582-58197582 on Build GRCh38
rs867887217 Chr.16: 58197662-58197662 on Build GRCh38
rs933079873 Chr.16: 58197417-58197417 on Build GRCh38
rs776712843 Chr.16: 58197647-58197647 on Build GRCh38
rs1030998777 Chr.16: 58197340-58197340 on Build GRCh38
rs976912799 Chr.16: 58197384-58197384 on Build GRCh38
rs1004883982 Chr.16: 58197354-58197354 on Build GRCh38
rs896490888 Chr.16: 58197558-58197558 on Build GRCh38
rs1025495175 Chr.16: 58197315-58197315 on Build GRCh38
rs775297340 Chr.16: 58197608-58197617 on Build GRCh38
rs970805667 Chr.16: 58197327-58197327 on Build GRCh38
rs977364425 Chr.16: 58197553-58197553 on Build GRCh38
rs761228953 Chr.16: 58197573-58197573 on Build GRCh38
rs1012503030 Chr.16: 58197471-58197471 on Build GRCh38
rs921738238 Chr.16: 58197524-58197524 on Build GRCh38
rs957127507 Chr.16: 58197590-58197590 on Build GRCh38
rs922785303 Chr.16: 58197561-58197561 on Build GRCh38
rs747826682 Chr.16: 58197611-58197611 on Build GRCh38
rs74019837 Chr.16: 58197362-58197362 on Build GRCh38
rs746593736 Chr.16: 58197611-58197612 on Build GRCh38
rs887855637 Chr.16: 58197547-58197547 on Build GRCh38
rs978502324 Chr.16: 58197337-58197337 on Build GRCh38
rs1037600932 Chr.16: 58197446-58197446 on Build GRCh38
rs772553045 Chr.16: 58197582-58197583 on Build GRCh38
rs958008364 Chr.16: 58197526-58197526 on Build GRCh38
rs781722523 Chr.16: 58197587-58197587 on Build GRCh38
rs149021498 Chr.16: 58197583-58197583 on Build GRCh38
rs545600875 Chr.16: 58197613-58197613 on Build GRCh38
rs907605439 Chr.16: 58197576-58197576 on Build GRCh38
rs969553226 Chr.16: 58197433-58197433 on Build GRCh38
rs777306084 Chr.16: 58197346-58197346 on Build GRCh38
rs1023779545 Chr.16: 58197423-58197423 on Build GRCh38
rs1024772642 Chr.16: 58197480-58197480 on Build GRCh38
rs964288297 Chr.16: 58197542-58197542 on Build GRCh38
rs756487076 Chr.16: 58197591-58197591 on Build GRCh38
rs996190773 Chr.16: 58197308-58197308 on Build GRCh38
rs771594630 Chr.16: 58197413-58197413 on Build GRCh38
rs537665757 Chr.16: 58197242-58197242 on Build GRCh38
rs944617512 Chr.16: 58197490-58197490 on Build GRCh38
rs867066879 Chr.16: 58197516-58197516 on Build GRCh38
rs1031038629 Chr.16: 58197510-58197510 on Build GRCh38
rs770840395 Chr.16: 58197641-58197641 on Build GRCh38
rs958310536 Chr.16: 58197358-58197358 on Build GRCh38
rs780344159 Chr.16: 58197609-58197609 on Build GRCh38

View Full Product Details